Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19968825-19969317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
UniGene
Hs.713616
Gene Chromosome Location
Chr.22: 19966491-20016802 on Build GRCh38
Transcripts
XM_005261242.2 XM_006724243.2 XM_006724246.3 XM_011530179.2 XM_011530182.2 NM_000754.3 NM_001135161.1 NM_001135162.1 NM_007310.2 XM_011529886.1 XM_017028594.1 XM_017028595.1

Gene Symbol
COMT
Entrez Gene ID
1312
Gene Name
catechol-O-methyltransferase
Gene Aliases
HEL-S-98n
UniGene
Hs.370408
Gene Chromosome Location
Chr.22: 19941740-19969975 on Build GRCh38
Transcripts
XM_005261242.2 XM_006724243.2 XM_006724246.3 XM_011530179.2 XM_011530182.2 NM_000754.3 NM_001135161.1 NM_001135162.1 NM_007310.2 XM_011529886.1 XM_017028594.1 XM_017028595.1

SNP Detail

SNP ID Chromosome Location
rs954133053 Chr.22: 19969162-19969162 on Build GRCh38
rs776181324 Chr.22: 19969264-19969264 on Build GRCh38
rs927576287 Chr.22: 19968982-19968982 on Build GRCh38
rs529947354 Chr.22: 19968928-19968928 on Build GRCh38
rs986733835 Chr.22: 19969231-19969231 on Build GRCh38
rs934034534 Chr.22: 19969057-19969057 on Build GRCh38
rs988745083 Chr.22: 19969248-19969248 on Build GRCh38
rs1034674434 Chr.22: 19968853-19968853 on Build GRCh38
rs1026534163 Chr.22: 19969250-19969250 on Build GRCh38
rs1044521483 Chr.22: 19969039-19969039 on Build GRCh38
rs1003115872 Chr.22: 19969077-19969077 on Build GRCh38
rs1053135372 Chr.22: 19968992-19968992 on Build GRCh38
rs937680650 Chr.22: 19968990-19968990 on Build GRCh38
rs138918504 Chr.22: 19969111-19969111 on Build GRCh38
rs896155299 Chr.22: 19969114-19969114 on Build GRCh38
rs531680429 Chr.22: 19968902-19968902 on Build GRCh38
rs372414281 Chr.22: 19968979-19968979 on Build GRCh38
rs1003844883 Chr.22: 19969120-19969120 on Build GRCh38
rs9332380 Chr.22: 19968894-19968894 on Build GRCh38
rs545915950 Chr.22: 19968857-19968857 on Build GRCh38
rs973872583 Chr.22: 19968875-19968875 on Build GRCh38
rs995362599 Chr.22: 19969130-19969130 on Build GRCh38
rs553561747 Chr.22: 19969054-19969054 on Build GRCh38
rs165599 Chr.22: 19969258-19969258 on Build GRCh38
rs9332381 Chr.22: 19969030-19969030 on Build GRCh38
rs934782448 Chr.22: 19968976-19968976 on Build GRCh38
rs112696498 Chr.22: 19969244-19969244 on Build GRCh38
rs879060530 Chr.22: 19968897-19968897 on Build GRCh38
rs947572832 Chr.22: 19969032-19969032 on Build GRCh38
rs746218496 Chr.22: 19969121-19969121 on Build GRCh38
rs569006363 Chr.22: 19969175-19969175 on Build GRCh38
rs561135670 Chr.22: 19968927-19968927 on Build GRCh38
rs962542027 Chr.22: 19968867-19968867 on Build GRCh38
rs1002298934 Chr.22: 19968847-19968847 on Build GRCh38
rs981893856 Chr.22: 19968980-19968980 on Build GRCh38
rs749822167 Chr.22: 19968866-19968866 on Build GRCh38
rs1003960733 Chr.22: 19968864-19968864 on Build GRCh38
rs889790482 Chr.22: 19968882-19968882 on Build GRCh38
rs987233753 Chr.22: 19968925-19968925 on Build GRCh38
rs1051190392 Chr.22: 19969083-19969083 on Build GRCh38
rs14968 Chr.22: 19968910-19968911 on Build GRCh38
rs1028143667 Chr.22: 19969149-19969149 on Build GRCh38
rs960769182 Chr.22: 19968858-19968858 on Build GRCh38
rs760848078 Chr.22: 19969286-19969286 on Build GRCh38
rs1034781128 Chr.22: 19969269-19969269 on Build GRCh38
rs1042989000 Chr.22: 19969040-19969040 on Build GRCh38
rs1007390072 Chr.22: 19968890-19968890 on Build GRCh38
rs772365005 Chr.22: 19969129-19969129 on Build GRCh38
rs145166266 Chr.22: 19968946-19968946 on Build GRCh38
rs956047560 Chr.22: 19969245-19969245 on Build GRCh38
rs550545168 Chr.22: 19968903-19968903 on Build GRCh38
rs915526088 Chr.22: 19969027-19969027 on Build GRCh38
rs973669692 Chr.22: 19968905-19968905 on Build GRCh38
rs879893042 Chr.22: 19968911-19968911 on Build GRCh38
rs552180875 Chr.22: 19969163-19969163 on Build GRCh38

View Full Product Details