Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19971796-19972302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
UniGene
Hs.713616
Gene Chromosome Location
Chr.22: 19966491-20016802 on Build GRCh38
Transcripts
NM_001670.2 XM_005261242.2 XM_005261243.3 XM_005261244.3 XM_006724243.2 XM_006724245.2 XM_006724246.3 XM_006724247.3 XM_006724248.3 XM_011530179.2 XM_011530180.1 XM_011530182.2 XM_011530183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356524 c.2734C>T p.R912W Chr.22: 19971933-19971933 on Build GRCh38
1190388 c.2699G>A p.G900E Chr.22: 19971968-19971968 on Build GRCh38
7356525 c.2726G>A p.R909Q Chr.22: 19971941-19971941 on Build GRCh38
725589 c.2725C>T p.R909W Chr.22: 19971942-19971942 on Build GRCh38
6262224 c.2742C>A p.A914A Chr.22: 19971925-19971925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774174976 Chr.22: 19971972-19971972 on Build GRCh38
rs951595515 Chr.22: 19972062-19972062 on Build GRCh38
rs201215289 Chr.22: 19971927-19971927 on Build GRCh38
rs370567245 Chr.22: 19971997-19971997 on Build GRCh38
rs755467214 Chr.22: 19972004-19972004 on Build GRCh38
rs776172743 Chr.22: 19971891-19971891 on Build GRCh38
rs1051530226 Chr.22: 19972187-19972187 on Build GRCh38
rs933175175 Chr.22: 19972178-19972178 on Build GRCh38
rs777526976 Chr.22: 19971874-19971874 on Build GRCh38
rs371273554 Chr.22: 19971946-19971946 on Build GRCh38
rs771156379 Chr.22: 19971963-19971963 on Build GRCh38
rs573444040 Chr.22: 19972119-19972119 on Build GRCh38
rs937066674 Chr.22: 19971978-19971978 on Build GRCh38
rs755596040 Chr.22: 19972000-19972000 on Build GRCh38
rs1045624489 Chr.22: 19971955-19971955 on Build GRCh38
rs377706812 Chr.22: 19971938-19971938 on Build GRCh38
rs921658965 Chr.22: 19972159-19972159 on Build GRCh38
rs5993891 Chr.22: 19972223-19972223 on Build GRCh38
rs757443161 Chr.22: 19972250-19972250 on Build GRCh38
rs972096562 Chr.22: 19972212-19972212 on Build GRCh38
rs753654167 Chr.22: 19971856-19971856 on Build GRCh38
rs905649206 Chr.22: 19971847-19971847 on Build GRCh38
rs374705390 Chr.22: 19971969-19971969 on Build GRCh38
rs779851910 Chr.22: 19971958-19971959 on Build GRCh38
rs34445280 Chr.22: 19971933-19971933 on Build GRCh38
rs775821127 Chr.22: 19971841-19971841 on Build GRCh38
rs202000927 Chr.22: 19971968-19971968 on Build GRCh38
rs773343199 Chr.22: 19971962-19971962 on Build GRCh38
rs1054631432 Chr.22: 19972008-19972008 on Build GRCh38
rs761554205 Chr.22: 19971977-19971977 on Build GRCh38
rs1051646967 Chr.22: 19971975-19971975 on Build GRCh38
rs34638476 Chr.22: 19971941-19971941 on Build GRCh38
rs542894098 Chr.22: 19971819-19971819 on Build GRCh38
rs966691516 Chr.22: 19971835-19971835 on Build GRCh38
rs372852955 Chr.22: 19971854-19971854 on Build GRCh38
rs138284380 Chr.22: 19971865-19971865 on Build GRCh38
rs765187569 Chr.22: 19971922-19971922 on Build GRCh38
rs376935202 Chr.22: 19971828-19971828 on Build GRCh38
rs916729982 Chr.22: 19971864-19971864 on Build GRCh38
rs534613187 Chr.22: 19971970-19971970 on Build GRCh38
rs748522405 Chr.22: 19971958-19971958 on Build GRCh38
rs774502315 Chr.22: 19971903-19971903 on Build GRCh38
rs772312656 Chr.22: 19971959-19971959 on Build GRCh38
rs927125181 Chr.22: 19972249-19972252 on Build GRCh38
rs759510236 Chr.22: 19971857-19971857 on Build GRCh38
rs763994428 Chr.22: 19971928-19971928 on Build GRCh38
rs35072297 Chr.22: 19972044-19972044 on Build GRCh38
rs999508441 Chr.22: 19971988-19971988 on Build GRCh38
rs202105687 Chr.22: 19971934-19971934 on Build GRCh38
rs542660635 Chr.22: 19972109-19972109 on Build GRCh38
rs752515297 Chr.22: 19971867-19971867 on Build GRCh38
rs200856269 Chr.22: 19971853-19971853 on Build GRCh38
rs553616292 Chr.22: 19972117-19972117 on Build GRCh38
rs902470370 Chr.22: 19972216-19972216 on Build GRCh38
rs748409278 Chr.22: 19971907-19971907 on Build GRCh38
rs1053668914 Chr.22: 19972245-19972245 on Build GRCh38
rs200129576 Chr.22: 19971932-19971932 on Build GRCh38
rs773325652 Chr.22: 19971909-19971909 on Build GRCh38
rs751278797 Chr.22: 19971877-19971877 on Build GRCh38
rs908868351 Chr.22: 19972091-19972091 on Build GRCh38
rs1009885709 Chr.22: 19972037-19972037 on Build GRCh38
rs902139362 Chr.22: 19972045-19972045 on Build GRCh38
rs780756406 Chr.22: 19971883-19971883 on Build GRCh38
rs753076529 Chr.22: 19972021-19972021 on Build GRCh38
rs573701681 Chr.22: 19972061-19972061 on Build GRCh38
rs758304166 Chr.22: 19971872-19971872 on Build GRCh38
rs982251633 Chr.22: 19972057-19972057 on Build GRCh38
rs902416547 Chr.22: 19971981-19971981 on Build GRCh38
rs970897948 Chr.22: 19972051-19972051 on Build GRCh38
rs374701376 Chr.22: 19971917-19971917 on Build GRCh38
rs756473032 Chr.22: 19972188-19972188 on Build GRCh38
rs775469355 Chr.22: 19971834-19971834 on Build GRCh38
rs1054196761 Chr.22: 19972255-19972255 on Build GRCh38
rs34687532 Chr.22: 19971942-19971942 on Build GRCh38
rs772252724 Chr.22: 19971908-19971908 on Build GRCh38
rs165815 Chr.22: 19971950-19971950 on Build GRCh38
rs368459939 Chr.22: 19971918-19971918 on Build GRCh38
rs545740094 Chr.22: 19972220-19972220 on Build GRCh38
rs754698792 Chr.22: 19971979-19971980 on Build GRCh38
rs745429036 Chr.22: 19971893-19971893 on Build GRCh38
rs200354896 Chr.22: 19972016-19972016 on Build GRCh38
rs918865248 Chr.22: 19972059-19972059 on Build GRCh38
rs945708332 Chr.22: 19972257-19972257 on Build GRCh38
rs377217563 Chr.22: 19971899-19971899 on Build GRCh38
rs779001051 Chr.22: 19972256-19972256 on Build GRCh38
rs1012218794 Chr.22: 19972249-19972249 on Build GRCh38
rs1012746830 Chr.22: 19972033-19972033 on Build GRCh38
rs200903789 Chr.22: 19971980-19971980 on Build GRCh38
rs754046547 Chr.22: 19972248-19972248 on Build GRCh38
rs983172209 Chr.22: 19972068-19972068 on Build GRCh38
rs557595858 Chr.22: 19971979-19971979 on Build GRCh38
rs765882627 Chr.22: 19972002-19972002 on Build GRCh38
rs953693305 Chr.22: 19972093-19972093 on Build GRCh38
rs753415069 Chr.22: 19972003-19972003 on Build GRCh38
rs1025810013 Chr.22: 19972272-19972272 on Build GRCh38
rs958185621 Chr.22: 19972025-19972025 on Build GRCh38
rs1041331920 Chr.22: 19972266-19972266 on Build GRCh38
rs1003741710 Chr.22: 19972261-19972261 on Build GRCh38
rs749814206 Chr.22: 19971943-19971943 on Build GRCh38
rs780579245 Chr.22: 19971894-19971894 on Build GRCh38
rs769983739 Chr.22: 19971836-19971836 on Build GRCh38
rs779441840 Chr.22: 19972014-19972014 on Build GRCh38
rs760715340 Chr.22: 19971911-19971911 on Build GRCh38
rs165824 Chr.22: 19971843-19971843 on Build GRCh38
rs768836282 Chr.22: 19971844-19971844 on Build GRCh38
rs775595848 Chr.22: 19971926-19971926 on Build GRCh38
rs373912180 Chr.22: 19972001-19972001 on Build GRCh38
rs375713594 Chr.22: 19971846-19971846 on Build GRCh38

View Full Product Details