Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19972280-19972779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
UniGene
Hs.713616
Gene Chromosome Location
Chr.22: 19966491-20016802 on Build GRCh38
Transcripts
NM_001670.2 XM_005261242.2 XM_005261243.3 XM_005261244.3 XM_006724243.2 XM_006724245.2 XM_006724246.3 XM_006724247.3 XM_006724248.3 XM_011530179.2 XM_011530180.1 XM_011530182.2 XM_011530183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3800036 c.2605G>A p.D869N Chr.22: 19972773-19972773 on Build GRCh38
1414919 c.2616G>A p.T872T Chr.22: 19972762-19972762 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572954360 Chr.22: 19972647-19972647 on Build GRCh38
rs776274170 Chr.22: 19972572-19972572 on Build GRCh38
rs1002639278 Chr.22: 19972588-19972588 on Build GRCh38
rs763364178 Chr.22: 19972440-19972440 on Build GRCh38
rs771030950 Chr.22: 19972757-19972757 on Build GRCh38
rs764469804 Chr.22: 19972721-19972721 on Build GRCh38
rs1029822587 Chr.22: 19972645-19972645 on Build GRCh38
rs958839856 Chr.22: 19972612-19972612 on Build GRCh38
rs2073744 Chr.22: 19972432-19972432 on Build GRCh38
rs192537896 Chr.22: 19972320-19972320 on Build GRCh38
rs765688298 Chr.22: 19972374-19972374 on Build GRCh38
rs753834271 Chr.22: 19972752-19972752 on Build GRCh38
rs747208340 Chr.22: 19972321-19972321 on Build GRCh38
rs770731874 Chr.22: 19972713-19972713 on Build GRCh38
rs34854812 Chr.22: 19972306-19972307 on Build GRCh38
rs551139321 Chr.22: 19972480-19972480 on Build GRCh38
rs762210783 Chr.22: 19972735-19972735 on Build GRCh38
rs1028704736 Chr.22: 19972512-19972512 on Build GRCh38
rs923639219 Chr.22: 19972530-19972530 on Build GRCh38
rs753180718 Chr.22: 19972436-19972436 on Build GRCh38
rs747611902 Chr.22: 19972739-19972740 on Build GRCh38
rs114219025 Chr.22: 19972396-19972396 on Build GRCh38
rs117249364 Chr.22: 19972535-19972535 on Build GRCh38
rs184260036 Chr.22: 19972571-19972571 on Build GRCh38
rs781607527 Chr.22: 19972393-19972393 on Build GRCh38
rs948112678 Chr.22: 19972415-19972415 on Build GRCh38
rs1017500702 Chr.22: 19972473-19972473 on Build GRCh38
rs781415499 Chr.22: 19972337-19972337 on Build GRCh38
rs954503442 Chr.22: 19972520-19972520 on Build GRCh38
rs915031564 Chr.22: 19972544-19972544 on Build GRCh38
rs199508558 Chr.22: 19972395-19972395 on Build GRCh38
rs778737856 Chr.22: 19972313-19972314 on Build GRCh38
rs778421058 Chr.22: 19972452-19972452 on Build GRCh38
rs766080914 Chr.22: 19972361-19972361 on Build GRCh38
rs1034264022 Chr.22: 19972594-19972594 on Build GRCh38
rs1047080875 Chr.22: 19972591-19972591 on Build GRCh38
rs188157323 Chr.22: 19972639-19972639 on Build GRCh38
rs933018405 Chr.22: 19972489-19972489 on Build GRCh38
rs746001701 Chr.22: 19972346-19972346 on Build GRCh38
rs767998510 Chr.22: 19972740-19972740 on Build GRCh38
rs746764007 Chr.22: 19972700-19972700 on Build GRCh38
rs750888016 Chr.22: 19972742-19972742 on Build GRCh38
rs974868547 Chr.22: 19972511-19972511 on Build GRCh38
rs369146419 Chr.22: 19972570-19972570 on Build GRCh38
rs113043239 Chr.22: 19972673-19972673 on Build GRCh38
rs555041511 Chr.22: 19972315-19972315 on Build GRCh38
rs539273029 Chr.22: 19972652-19972652 on Build GRCh38
rs768684700 Chr.22: 19972347-19972347 on Build GRCh38
rs62223685 Chr.22: 19972617-19972617 on Build GRCh38
rs753326944 Chr.22: 19972368-19972368 on Build GRCh38
rs752026069 Chr.22: 19972391-19972391 on Build GRCh38
rs571249027 Chr.22: 19972492-19972492 on Build GRCh38
rs371911365 Chr.22: 19972407-19972407 on Build GRCh38
rs760516494 Chr.22: 19972423-19972423 on Build GRCh38
rs1054016102 Chr.22: 19972399-19972399 on Build GRCh38
rs762571181 Chr.22: 19972718-19972718 on Build GRCh38
rs758249307 Chr.22: 19972687-19972687 on Build GRCh38
rs889407010 Chr.22: 19972513-19972513 on Build GRCh38
rs1016345751 Chr.22: 19972369-19972369 on Build GRCh38
rs201801179 Chr.22: 19972392-19972392 on Build GRCh38
rs200210011 Chr.22: 19972357-19972357 on Build GRCh38
rs760320413 Chr.22: 19972360-19972360 on Build GRCh38
rs771068133 Chr.22: 19972322-19972322 on Build GRCh38
rs375556189 Chr.22: 19972450-19972450 on Build GRCh38
rs759083877 Chr.22: 19972373-19972373 on Build GRCh38
rs1038628360 Chr.22: 19972619-19972619 on Build GRCh38
rs899963226 Chr.22: 19972633-19972633 on Build GRCh38
rs200572444 Chr.22: 19972697-19972697 on Build GRCh38
rs766675717 Chr.22: 19972750-19972750 on Build GRCh38
rs71313931 Chr.22: 19972661-19972661 on Build GRCh38
rs573735064 Chr.22: 19972507-19972510 on Build GRCh38
rs756281840 Chr.22: 19972744-19972744 on Build GRCh38
rs747611625 Chr.22: 19972324-19972337 on Build GRCh38
rs772873012 Chr.22: 19972422-19972422 on Build GRCh38
rs776204587 Chr.22: 19972714-19972714 on Build GRCh38
rs887137911 Chr.22: 19972598-19972598 on Build GRCh38
rs774187739 Chr.22: 19972354-19972354 on Build GRCh38
rs935228748 Chr.22: 19972534-19972534 on Build GRCh38
rs75475026 Chr.22: 19972715-19972715 on Build GRCh38
rs897966498 Chr.22: 19972736-19972736 on Build GRCh38
rs368737055 Chr.22: 19972461-19972461 on Build GRCh38
rs544318821 Chr.22: 19972303-19972303 on Build GRCh38
rs753163806 Chr.22: 19972377-19972377 on Build GRCh38
rs774749996 Chr.22: 19972601-19972601 on Build GRCh38
rs767801438 Chr.22: 19972457-19972457 on Build GRCh38
rs748044245 Chr.22: 19972409-19972409 on Build GRCh38
rs528241767 Chr.22: 19972434-19972434 on Build GRCh38
rs751949219 Chr.22: 19972454-19972454 on Build GRCh38
rs71313932 Chr.22: 19972675-19972675 on Build GRCh38
rs987092348 Chr.22: 19972527-19972527 on Build GRCh38
rs62223684 Chr.22: 19972390-19972390 on Build GRCh38
rs368871636 Chr.22: 19972458-19972458 on Build GRCh38
rs2518823 Chr.22: 19972665-19972665 on Build GRCh38
rs771720447 Chr.22: 19972430-19972432 on Build GRCh38
rs749447027 Chr.22: 19972462-19972462 on Build GRCh38
rs1008626742 Chr.22: 19972701-19972701 on Build GRCh38
rs761419496 Chr.22: 19972614-19972614 on Build GRCh38
rs775022925 Chr.22: 19972725-19972725 on Build GRCh38
rs887349099 Chr.22: 19972438-19972438 on Build GRCh38
rs35790506 Chr.22: 19972306-19972306 on Build GRCh38
rs942203423 Chr.22: 19972521-19972521 on Build GRCh38
rs375174769 Chr.22: 19972430-19972430 on Build GRCh38
rs758993813 Chr.22: 19972433-19972433 on Build GRCh38
rs755112997 Chr.22: 19972753-19972753 on Build GRCh38
rs1021390839 Chr.22: 19972635-19972635 on Build GRCh38
rs761620841 Chr.22: 19972356-19972356 on Build GRCh38
rs771247117 Chr.22: 19972557-19972557 on Build GRCh38
rs758731278 Chr.22: 19972385-19972385 on Build GRCh38
rs772118624 Chr.22: 19972412-19972412 on Build GRCh38

View Full Product Details