Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19973975-19974516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
UniGene
Hs.713616
Gene Chromosome Location
Chr.22: 19966491-20016802 on Build GRCh38
Transcripts
NM_001670.2 XM_005261242.2 XM_005261243.3 XM_005261244.3 XM_006724243.2 XM_006724245.2 XM_006724246.3 XM_006724247.3 XM_006724248.3 XM_006724249.3 XM_006724250.3 XM_011530179.2 XM_011530180.1 XM_011530182.2 XM_011530183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344722 c.1981C>A p.P661T Chr.22: 19974219-19974219 on Build GRCh38
4769613 c.2024G>A p.R675Q Chr.22: 19974176-19974176 on Build GRCh38
5344031 c.2016G>A p.T672T Chr.22: 19974184-19974184 on Build GRCh38
3842202 c.1966G>C p.E656Q Chr.22: 19974234-19974234 on Build GRCh38
53510 c.2006C>G p.S669C Chr.22: 19974194-19974194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972528321 Chr.22: 19974213-19974213 on Build GRCh38
rs550149998 Chr.22: 19974119-19974119 on Build GRCh38
rs527910876 Chr.22: 19974016-19974016 on Build GRCh38
rs376063274 Chr.22: 19974103-19974103 on Build GRCh38
rs73387719 Chr.22: 19974040-19974040 on Build GRCh38
rs373944893 Chr.22: 19974093-19974093 on Build GRCh38
rs946097653 Chr.22: 19974356-19974356 on Build GRCh38
rs904146825 Chr.22: 19974038-19974038 on Build GRCh38
rs1052635852 Chr.22: 19974496-19974496 on Build GRCh38
rs750747299 Chr.22: 19974082-19974082 on Build GRCh38
rs752977311 Chr.22: 19974244-19974244 on Build GRCh38
rs757741348 Chr.22: 19974070-19974070 on Build GRCh38
rs924936894 Chr.22: 19974483-19974483 on Build GRCh38
rs767021296 Chr.22: 19974247-19974247 on Build GRCh38
rs887199 Chr.22: 19974432-19974432 on Build GRCh38
rs943989761 Chr.22: 19974344-19974344 on Build GRCh38
rs539743675 Chr.22: 19974096-19974096 on Build GRCh38
rs947755953 Chr.22: 19974469-19974469 on Build GRCh38
rs781213545 Chr.22: 19974171-19974171 on Build GRCh38
rs919627057 Chr.22: 19974006-19974006 on Build GRCh38
rs148872323 Chr.22: 19974169-19974169 on Build GRCh38
rs2239394 Chr.22: 19974403-19974403 on Build GRCh38
rs755388828 Chr.22: 19974222-19974222 on Build GRCh38
rs550169842 Chr.22: 19974492-19974492 on Build GRCh38
rs575921465 Chr.22: 19974232-19974232 on Build GRCh38
rs943875243 Chr.22: 19974085-19974085 on Build GRCh38
rs780072614 Chr.22: 19974266-19974266 on Build GRCh38
rs747558161 Chr.22: 19974406-19974406 on Build GRCh38
rs889580906 Chr.22: 19974057-19974057 on Build GRCh38
rs919933905 Chr.22: 19974144-19974144 on Build GRCh38
rs115025697 Chr.22: 19974145-19974145 on Build GRCh38
rs749565212 Chr.22: 19974259-19974268 on Build GRCh38
rs1043951946 Chr.22: 19974037-19974037 on Build GRCh38
rs185600338 Chr.22: 19974251-19974251 on Build GRCh38
rs535959125 Chr.22: 19974136-19974136 on Build GRCh38
rs544772347 Chr.22: 19974446-19974446 on Build GRCh38
rs761344502 Chr.22: 19974141-19974141 on Build GRCh38
rs768154752 Chr.22: 19974194-19974194 on Build GRCh38
rs553999690 Chr.22: 19974179-19974179 on Build GRCh38
rs547571518 Chr.22: 19974032-19974032 on Build GRCh38
rs762502292 Chr.22: 19974130-19974130 on Build GRCh38
rs748988551 Chr.22: 19974280-19974280 on Build GRCh38
rs564297048 Chr.22: 19974407-19974407 on Build GRCh38
rs755121911 Chr.22: 19974156-19974156 on Build GRCh38
rs1026738058 Chr.22: 19974470-19974470 on Build GRCh38
rs960566816 Chr.22: 19974442-19974442 on Build GRCh38
rs952502941 Chr.22: 19974487-19974487 on Build GRCh38
rs150166166 Chr.22: 19974124-19974124 on Build GRCh38
rs775425001 Chr.22: 19974126-19974126 on Build GRCh38
rs972797152 Chr.22: 19973999-19973999 on Build GRCh38
rs983352892 Chr.22: 19974411-19974411 on Build GRCh38
rs370344734 Chr.22: 19974068-19974068 on Build GRCh38
rs200370781 Chr.22: 19974177-19974177 on Build GRCh38
rs994287630 Chr.22: 19974456-19974456 on Build GRCh38
rs781694266 Chr.22: 19974081-19974081 on Build GRCh38
rs1052666149 Chr.22: 19974346-19974346 on Build GRCh38
rs78866590 Chr.22: 19974217-19974217 on Build GRCh38
rs1043157050 Chr.22: 19974375-19974375 on Build GRCh38
rs1018353539 Chr.22: 19974371-19974371 on Build GRCh38
rs146960902 Chr.22: 19974207-19974207 on Build GRCh38
rs907678586 Chr.22: 19974192-19974192 on Build GRCh38
rs1001535372 Chr.22: 19974386-19974386 on Build GRCh38
rs557355440 Chr.22: 19974290-19974290 on Build GRCh38
rs758763375 Chr.22: 19974249-19974249 on Build GRCh38
rs770947787 Chr.22: 19974114-19974114 on Build GRCh38
rs911134297 Chr.22: 19974318-19974318 on Build GRCh38
rs138605057 Chr.22: 19974150-19974150 on Build GRCh38
rs879900237 Chr.22: 19974399-19974399 on Build GRCh38
rs776735612 Chr.22: 19974118-19974118 on Build GRCh38
rs767954651 Chr.22: 19974143-19974143 on Build GRCh38
rs527378110 Chr.22: 19974351-19974351 on Build GRCh38
rs899883061 Chr.22: 19974373-19974373 on Build GRCh38
rs904485080 Chr.22: 19974380-19974380 on Build GRCh38
rs774329687 Chr.22: 19974160-19974160 on Build GRCh38
rs745799425 Chr.22: 19974176-19974176 on Build GRCh38
rs1041262789 Chr.22: 19974349-19974349 on Build GRCh38
rs755976851 Chr.22: 19974265-19974265 on Build GRCh38
rs916424902 Chr.22: 19974447-19974447 on Build GRCh38
rs376804470 Chr.22: 19974166-19974166 on Build GRCh38
rs761063363 Chr.22: 19974210-19974210 on Build GRCh38
rs777990697 Chr.22: 19974257-19974257 on Build GRCh38
rs748185873 Chr.22: 19974168-19974168 on Build GRCh38
rs748275533 Chr.22: 19974097-19974097 on Build GRCh38
rs749983318 Chr.22: 19974305-19974305 on Build GRCh38
rs768977913 Chr.22: 19974095-19974095 on Build GRCh38
rs1031089115 Chr.22: 19974053-19974053 on Build GRCh38
rs766638279 Chr.22: 19974151-19974151 on Build GRCh38
rs992989364 Chr.22: 19974172-19974172 on Build GRCh38
rs770497354 Chr.22: 19974279-19974279 on Build GRCh38
rs756484797 Chr.22: 19974087-19974087 on Build GRCh38
rs541788300 Chr.22: 19974262-19974262 on Build GRCh38
rs202240748 Chr.22: 19974198-19974198 on Build GRCh38
rs985303245 Chr.22: 19974308-19974308 on Build GRCh38
rs772764526 Chr.22: 19974209-19974209 on Build GRCh38
rs569906702 Chr.22: 19974132-19974132 on Build GRCh38
rs955551807 Chr.22: 19974063-19974063 on Build GRCh38
rs145699486 Chr.22: 19974493-19974493 on Build GRCh38
rs754276278 Chr.22: 19974155-19974155 on Build GRCh38
rs749150029 Chr.22: 19974193-19974193 on Build GRCh38
rs1026966598 Chr.22: 19974396-19974396 on Build GRCh38
rs1049645386 Chr.22: 19974056-19974056 on Build GRCh38

View Full Product Details