Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49922532-49923035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRELD2
Entrez Gene ID
79174
Gene Name
cysteine rich with EGF like domains 2
Gene Aliases
-
UniGene
Hs.211282
Gene Chromosome Location
Chr.22: 49918630-49927540 on Build GRCh38
Transcripts
NM_001135101.2 NM_001284317.1 NM_001284318.1 NM_024324.4 NR_104295.1 XM_005261737.2 XM_005261738.4 XM_017028938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5494249 c.819C>T p.D273D Chr.22: 49922691-49922691 on Build GRCh38
5494250 c.672C>T p.D224D Chr.22: 49922691-49922691 on Build GRCh38
1714512 c.593_594CC>TT p.A198V Chr.22: 49922612-49922613 on Build GRCh38
6283581 c.683G>A p.C228Y Chr.22: 49922702-49922702 on Build GRCh38
3939678 c.820G>C p.E274Q Chr.22: 49922692-49922692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988931290 Chr.22: 49922952-49922952 on Build GRCh38
rs751565719 Chr.22: 49922636-49922636 on Build GRCh38
rs386822609 Chr.22: 49922891-49922949 on Build GRCh38
rs964685257 Chr.22: 49922976-49922976 on Build GRCh38
rs763325653 Chr.22: 49922731-49922731 on Build GRCh38
rs952319406 Chr.22: 49922633-49922633 on Build GRCh38
rs575510807 Chr.22: 49922557-49922557 on Build GRCh38
rs748837257 Chr.22: 49922691-49922691 on Build GRCh38
rs528199585 Chr.22: 49922900-49922900 on Build GRCh38
rs1005660745 Chr.22: 49922703-49922703 on Build GRCh38
rs530397079 Chr.22: 49922752-49922752 on Build GRCh38
rs56351880 Chr.22: 49922949-49922949 on Build GRCh38
rs371852940 Chr.22: 49922698-49922698 on Build GRCh38
rs200309486 Chr.22: 49922716-49922716 on Build GRCh38
rs12152125 Chr.22: 49922796-49922796 on Build GRCh38
rs111257439 Chr.22: 49922561-49922561 on Build GRCh38
rs890325060 Chr.22: 49922979-49922979 on Build GRCh38
rs747719941 Chr.22: 49922661-49922661 on Build GRCh38
rs774822691 Chr.22: 49922697-49922697 on Build GRCh38
rs371632070 Chr.22: 49922596-49922596 on Build GRCh38
rs58253304 Chr.22: 49922921-49922921 on Build GRCh38
rs58312473 Chr.22: 49922836-49922836 on Build GRCh38
rs564007722 Chr.22: 49922780-49922780 on Build GRCh38
rs867638334 Chr.22: 49922644-49922644 on Build GRCh38
rs12171243 Chr.22: 49922726-49922726 on Build GRCh38
rs964798863 Chr.22: 49922611-49922611 on Build GRCh38
rs760113072 Chr.22: 49922589-49922589 on Build GRCh38
rs929242167 Chr.22: 49922937-49922937 on Build GRCh38
rs911193977 Chr.22: 49923000-49923000 on Build GRCh38
rs1001765080 Chr.22: 49922750-49922750 on Build GRCh38
rs766204075 Chr.22: 49922744-49922744 on Build GRCh38
rs58912478 Chr.22: 49922893-49922894 on Build GRCh38
rs61470167 Chr.22: 49922794-49922795 on Build GRCh38
rs758223623 Chr.22: 49922613-49922613 on Build GRCh38
rs373662275 Chr.22: 49922620-49922620 on Build GRCh38
rs375697289 Chr.22: 49923004-49923004 on Build GRCh38
rs1018940866 Chr.22: 49922956-49922956 on Build GRCh38
rs939563682 Chr.22: 49922649-49922649 on Build GRCh38
rs777489583 Chr.22: 49922664-49922664 on Build GRCh38
rs57978026 Chr.22: 49922826-49922826 on Build GRCh38
rs867013755 Chr.22: 49922681-49922681 on Build GRCh38
rs61510346 Chr.22: 49922891-49922891 on Build GRCh38
rs61443265 Chr.22: 49922825-49922825 on Build GRCh38
rs138811682 Chr.22: 49922640-49922640 on Build GRCh38
rs59946573 Chr.22: 49922811-49922811 on Build GRCh38
rs375930739 Chr.22: 49922751-49922751 on Build GRCh38
rs201724572 Chr.22: 49922579-49922579 on Build GRCh38
rs373405906 Chr.22: 49922825-49922825 on Build GRCh38
rs60207958 Chr.22: 49922806-49922806 on Build GRCh38
rs752416765 Chr.22: 49922685-49922685 on Build GRCh38
rs552163946 Chr.22: 49922773-49922773 on Build GRCh38
rs1035828588 Chr.22: 49922928-49922928 on Build GRCh38
rs140599118 Chr.22: 49922924-49922927 on Build GRCh38
rs371586711 Chr.22: 49922737-49922737 on Build GRCh38
rs1028165476 Chr.22: 49922984-49922984 on Build GRCh38
rs753940500 Chr.22: 49922597-49922597 on Build GRCh38
rs775053612 Chr.22: 49922582-49922582 on Build GRCh38
rs942607669 Chr.22: 49922628-49922628 on Build GRCh38
rs13053205 Chr.22: 49922812-49922812 on Build GRCh38
rs61046250 Chr.22: 49922881-49922881 on Build GRCh38
rs867948033 Chr.22: 49922612-49922612 on Build GRCh38
rs771487243 Chr.22: 49922692-49922692 on Build GRCh38
rs60822170 Chr.22: 49922805-49922805 on Build GRCh38
rs60375258 Chr.22: 49922846-49922846 on Build GRCh38
rs569206113 Chr.22: 49922556-49922556 on Build GRCh38
rs9616209 Chr.22: 49922721-49922721 on Build GRCh38
rs60305440 Chr.22: 49922818-49922818 on Build GRCh38
rs60440825 Chr.22: 49922853-49922853 on Build GRCh38
rs759098096 Chr.22: 49922619-49922619 on Build GRCh38
rs947423350 Chr.22: 49922572-49922572 on Build GRCh38
rs868106914 Chr.22: 49922639-49922639 on Build GRCh38
rs760622469 Chr.22: 49922594-49922594 on Build GRCh38
rs1040930363 Chr.22: 49923013-49923013 on Build GRCh38
rs888190670 Chr.22: 49922695-49922695 on Build GRCh38
rs572981290 Chr.22: 49922722-49922722 on Build GRCh38
rs59013246 Chr.22: 49922839-49922840 on Build GRCh38
rs767742220 Chr.22: 49922590-49922590 on Build GRCh38
rs751334595 Chr.22: 49922746-49922746 on Build GRCh38
rs190968664 Chr.22: 49922823-49922823 on Build GRCh38
rs947827524 Chr.22: 49922665-49922665 on Build GRCh38
rs755543544 Chr.22: 49922662-49922662 on Build GRCh38
rs148937327 Chr.22: 49922995-49922995 on Build GRCh38
rs908136766 Chr.22: 49922643-49922643 on Build GRCh38
rs202024363 Chr.22: 49922653-49922653 on Build GRCh38

View Full Product Details