Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50739192-50739686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACR
Entrez Gene ID
49
Gene Name
acrosin
Gene Aliases
-
UniGene
Hs.370870
Gene Chromosome Location
Chr.22: 50738224-50745299 on Build GRCh38
Transcripts
NM_001097.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354007 c.167G>T p.W56L Chr.22: 50739360-50739360 on Build GRCh38
5764261 c.94C>T p.R32W Chr.22: 50739287-50739287 on Build GRCh38
1244143 c.133G>A p.G45S Chr.22: 50739326-50739326 on Build GRCh38
5772021 c.78-6C>A p.? Chr.22: 50739265-50739265 on Build GRCh38
6005052 c.281+4C>T p.? Chr.22: 50739478-50739478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34962697 Chr.22: 50739534-50739535 on Build GRCh38
rs775250607 Chr.22: 50739440-50739440 on Build GRCh38
rs926344766 Chr.22: 50739642-50739642 on Build GRCh38
rs377766496 Chr.22: 50739257-50739257 on Build GRCh38
rs769870090 Chr.22: 50739434-50739434 on Build GRCh38
rs769015823 Chr.22: 50739397-50739397 on Build GRCh38
rs200217266 Chr.22: 50739649-50739649 on Build GRCh38
rs903875449 Chr.22: 50739265-50739265 on Build GRCh38
rs1036469476 Chr.22: 50739617-50739617 on Build GRCh38
rs77313877 Chr.22: 50739500-50739500 on Build GRCh38
rs759273323 Chr.22: 50739352-50739352 on Build GRCh38
rs766256979 Chr.22: 50739292-50739292 on Build GRCh38
rs963837122 Chr.22: 50739321-50739321 on Build GRCh38
rs775336676 Chr.22: 50739512-50739512 on Build GRCh38
rs1015936880 Chr.22: 50739624-50739624 on Build GRCh38
rs756717420 Chr.22: 50739381-50739381 on Build GRCh38
rs746332641 Chr.22: 50739385-50739385 on Build GRCh38
rs767026641 Chr.22: 50739656-50739656 on Build GRCh38
rs896561196 Chr.22: 50739467-50739467 on Build GRCh38
rs537267372 Chr.22: 50739593-50739593 on Build GRCh38
rs780466256 Chr.22: 50739388-50739388 on Build GRCh38
rs376377088 Chr.22: 50739335-50739335 on Build GRCh38
rs191165317 Chr.22: 50739323-50739323 on Build GRCh38
rs771782584 Chr.22: 50739535-50739535 on Build GRCh38
rs975094965 Chr.22: 50739356-50739356 on Build GRCh38
rs772193783 Chr.22: 50739249-50739249 on Build GRCh38
rs150273293 Chr.22: 50739463-50739463 on Build GRCh38
rs764251990 Chr.22: 50739264-50739264 on Build GRCh38
rs979796868 Chr.22: 50739289-50739289 on Build GRCh38
rs865888697 Chr.22: 50739468-50739468 on Build GRCh38
rs377757183 Chr.22: 50739481-50739481 on Build GRCh38
rs768927642 Chr.22: 50739334-50739334 on Build GRCh38
rs761928464 Chr.22: 50739346-50739346 on Build GRCh38
rs764777015 Chr.22: 50739464-50739464 on Build GRCh38
rs745849346 Chr.22: 50739506-50739506 on Build GRCh38
rs373285057 Chr.22: 50739511-50739511 on Build GRCh38
rs763738146 Chr.22: 50739456-50739456 on Build GRCh38
rs760865181 Chr.22: 50739258-50739258 on Build GRCh38
rs75385660 Chr.22: 50739570-50739570 on Build GRCh38
rs965107563 Chr.22: 50739554-50739554 on Build GRCh38
rs781006970 Chr.22: 50739558-50739558 on Build GRCh38
rs780653208 Chr.22: 50739470-50739470 on Build GRCh38
rs372139399 Chr.22: 50739326-50739326 on Build GRCh38
rs750430072 Chr.22: 50739268-50739268 on Build GRCh38
rs748269754 Chr.22: 50739408-50739408 on Build GRCh38
rs550216074 Chr.22: 50739637-50739637 on Build GRCh38
rs752344889 Chr.22: 50739314-50739314 on Build GRCh38
rs983819730 Chr.22: 50739572-50739572 on Build GRCh38
rs975609850 Chr.22: 50739564-50739564 on Build GRCh38
rs554850342 Chr.22: 50739539-50739539 on Build GRCh38
rs748359489 Chr.22: 50739489-50739489 on Build GRCh38
rs769803264 Chr.22: 50739496-50739496 on Build GRCh38
rs774203834 Chr.22: 50739435-50739435 on Build GRCh38
rs956628467 Chr.22: 50739538-50739538 on Build GRCh38
rs1024084186 Chr.22: 50739245-50739245 on Build GRCh38
rs1064733 Chr.22: 50739384-50739384 on Build GRCh38
rs553429158 Chr.22: 50739653-50739653 on Build GRCh38
rs777321406 Chr.22: 50739372-50739372 on Build GRCh38
rs755668971 Chr.22: 50739316-50739316 on Build GRCh38
rs576896841 Chr.22: 50739317-50739317 on Build GRCh38
rs147948077 Chr.22: 50739594-50739594 on Build GRCh38
rs531697145 Chr.22: 50739529-50739529 on Build GRCh38
rs750963565 Chr.22: 50739465-50739465 on Build GRCh38
rs760402111 Chr.22: 50739446-50739446 on Build GRCh38
rs185375060 Chr.22: 50739213-50739213 on Build GRCh38
rs758733691 Chr.22: 50739386-50739386 on Build GRCh38
rs140628153 Chr.22: 50739328-50739328 on Build GRCh38
rs370903765 Chr.22: 50739303-50739303 on Build GRCh38
rs749966445 Chr.22: 50739650-50739650 on Build GRCh38
rs774572935 Chr.22: 50739223-50739223 on Build GRCh38
rs751325618 Chr.22: 50739297-50739297 on Build GRCh38
rs753355839 Chr.22: 50739377-50739377 on Build GRCh38
rs145756479 Chr.22: 50739370-50739370 on Build GRCh38
rs749856990 Chr.22: 50739322-50739322 on Build GRCh38
rs762982880 Chr.22: 50739287-50739287 on Build GRCh38
rs773101273 Chr.22: 50739619-50739619 on Build GRCh38
rs558691256 Chr.22: 50739598-50739598 on Build GRCh38
rs374344475 Chr.22: 50739478-50739478 on Build GRCh38
rs369405326 Chr.22: 50739325-50739325 on Build GRCh38
rs767321569 Chr.22: 50739367-50739367 on Build GRCh38
rs565587762 Chr.22: 50739522-50739522 on Build GRCh38
rs755756962 Chr.22: 50739371-50739371 on Build GRCh38
rs753445401 Chr.22: 50739457-50739457 on Build GRCh38
rs1013510465 Chr.22: 50739218-50739218 on Build GRCh38
rs760951346 Chr.22: 50739329-50739329 on Build GRCh38
rs759858900 Chr.22: 50739242-50739242 on Build GRCh38
rs752065858 Chr.22: 50739661-50739661 on Build GRCh38
rs920934156 Chr.22: 50739567-50739567 on Build GRCh38
rs766233441 Chr.22: 50739348-50739348 on Build GRCh38
rs900602481 Chr.22: 50739536-50739536 on Build GRCh38
rs76866150 Chr.22: 50739390-50739390 on Build GRCh38
rs189214490 Chr.22: 50739513-50739513 on Build GRCh38
rs1019489427 Chr.22: 50739553-50739553 on Build GRCh38
rs1045521116 Chr.22: 50739636-50739636 on Build GRCh38
rs776310766 Chr.22: 50739647-50739647 on Build GRCh38
rs2285395 Chr.22: 50739662-50739662 on Build GRCh38
rs114634361 Chr.22: 50739436-50739436 on Build GRCh38
rs1009953090 Chr.22: 50739238-50739238 on Build GRCh38
rs868658360 Chr.22: 50739416-50739416 on Build GRCh38
rs748800676 Chr.22: 50739466-50739466 on Build GRCh38
rs368884202 Chr.22: 50739349-50739349 on Build GRCh38
rs949489029 Chr.22: 50739623-50739623 on Build GRCh38
rs561127825 Chr.22: 50739401-50739401 on Build GRCh38

View Full Product Details