Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196587292-196587804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO45
Entrez Gene ID
200933
Gene Name
F-box protein 45
Gene Aliases
Fbx45
UniGene
Hs.169815
Gene Chromosome Location
Chr.3: 196568854-196589059 on Build GRCh38
Transcripts
NM_001105573.1

SNP Detail

SNP ID Chromosome Location
rs977672841 Chr.3: 196587597-196587597 on Build GRCh38
rs57643210 Chr.3: 196587742-196587743 on Build GRCh38
rs1057244594 Chr.3: 196587317-196587317 on Build GRCh38
rs4916523 Chr.3: 196587463-196587463 on Build GRCh38
rs77592178 Chr.3: 196587537-196587537 on Build GRCh38
rs893162744 Chr.3: 196587732-196587732 on Build GRCh38
rs9829370 Chr.3: 196587733-196587733 on Build GRCh38
rs568317745 Chr.3: 196587723-196587723 on Build GRCh38
rs560776480 Chr.3: 196587432-196587432 on Build GRCh38
rs780226536 Chr.3: 196587778-196587778 on Build GRCh38
rs1026685606 Chr.3: 196587496-196587497 on Build GRCh38
rs1006455876 Chr.3: 196587734-196587734 on Build GRCh38
rs550146383 Chr.3: 196587705-196587705 on Build GRCh38
rs145304112 Chr.3: 196587351-196587351 on Build GRCh38
rs916029938 Chr.3: 196587704-196587704 on Build GRCh38
rs768037726 Chr.3: 196587547-196587547 on Build GRCh38
rs919450204 Chr.3: 196587775-196587775 on Build GRCh38
rs111836612 Chr.3: 196587733-196587734 on Build GRCh38
rs928709587 Chr.3: 196587599-196587599 on Build GRCh38
rs775811955 Chr.3: 196587592-196587592 on Build GRCh38
rs142170058 Chr.3: 196587612-196587612 on Build GRCh38
rs1057150574 Chr.3: 196587703-196587703 on Build GRCh38
rs9825103 Chr.3: 196587340-196587340 on Build GRCh38
rs961768521 Chr.3: 196587514-196587514 on Build GRCh38
rs997575416 Chr.3: 196587519-196587519 on Build GRCh38
rs914703745 Chr.3: 196587579-196587579 on Build GRCh38
rs972620257 Chr.3: 196587737-196587737 on Build GRCh38
rs924844371 Chr.3: 196587629-196587629 on Build GRCh38
rs777843814 Chr.3: 196587627-196587628 on Build GRCh38
rs1056186935 Chr.3: 196587685-196587685 on Build GRCh38
rs1007905114 Chr.3: 196587350-196587350 on Build GRCh38
rs773296756 Chr.3: 196587337-196587337 on Build GRCh38
rs537987359 Chr.3: 196587665-196587665 on Build GRCh38
rs982717286 Chr.3: 196587515-196587515 on Build GRCh38
rs548551913 Chr.3: 196587383-196587383 on Build GRCh38
rs1020391311 Chr.3: 196587322-196587322 on Build GRCh38
rs771211881 Chr.3: 196587376-196587376 on Build GRCh38
rs559368539 Chr.3: 196587398-196587398 on Build GRCh38
rs774686316 Chr.3: 196587391-196587391 on Build GRCh38
rs897736860 Chr.3: 196587774-196587774 on Build GRCh38
rs1044920692 Chr.3: 196587721-196587721 on Build GRCh38
rs764771564 Chr.3: 196587746-196587746 on Build GRCh38
rs746337849 Chr.3: 196587724-196587724 on Build GRCh38
rs986271292 Chr.3: 196587575-196587575 on Build GRCh38
rs948747695 Chr.3: 196587716-196587716 on Build GRCh38

View Full Product Details