Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1395503-1396009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIPAK
Entrez Gene ID
285464
Gene Name
cysteine rich PAK1 inhibitor
Gene Aliases
-
UniGene
Hs.26410
Gene Chromosome Location
Chr.4: 1391552-1395994 on Build GRCh38
Transcripts
NM_175918.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3011048 c.1043T>C p.M348T Chr.4: 1395554-1395554 on Build GRCh38
5865766 c.1117C>T p.P373S Chr.4: 1395628-1395628 on Build GRCh38
391039 c.1093_1094delCA p.R367fs*40 Chr.4: 1395604-1395605 on Build GRCh38
3011046 c.1024G>A p.A342T Chr.4: 1395535-1395535 on Build GRCh38
7362687 c.1258G>A p.V420I Chr.4: 1395769-1395769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757986011 Chr.4: 1395587-1395616 on Build GRCh38
rs780420861 Chr.4: 1395862-1395862 on Build GRCh38
rs750083721 Chr.4: 1395542-1395543 on Build GRCh38
rs1140011 Chr.4: 1395535-1395535 on Build GRCh38
rs368420407 Chr.4: 1395817-1395817 on Build GRCh38
rs763579027 Chr.4: 1395524-1395526 on Build GRCh38
rs370690293 Chr.4: 1395760-1395760 on Build GRCh38
rs990315238 Chr.4: 1395795-1395795 on Build GRCh38
rs763344201 Chr.4: 1395542-1395542 on Build GRCh38
rs34525526 Chr.4: 1395609-1395610 on Build GRCh38
rs771051554 Chr.4: 1395900-1395900 on Build GRCh38
rs374867542 Chr.4: 1395597-1395597 on Build GRCh38
rs780660443 Chr.4: 1395803-1395803 on Build GRCh38
rs1027654191 Chr.4: 1395739-1395739 on Build GRCh38
rs756850760 Chr.4: 1395858-1395858 on Build GRCh38
rs753266415 Chr.4: 1395853-1395853 on Build GRCh38
rs767780297 Chr.4: 1395691-1395691 on Build GRCh38
rs1031718553 Chr.4: 1395935-1395935 on Build GRCh38
rs143447149 Chr.4: 1395745-1395745 on Build GRCh38
rs761645889 Chr.4: 1395661-1395693 on Build GRCh38
rs748502553 Chr.4: 1395597-1395598 on Build GRCh38
rs145208075 Chr.4: 1395645-1395645 on Build GRCh38
rs762933937 Chr.4: 1395899-1395899 on Build GRCh38
rs746378277 Chr.4: 1395742-1395742 on Build GRCh38
rs541093494 Chr.4: 1395622-1395622 on Build GRCh38
rs757377224 Chr.4: 1395701-1395701 on Build GRCh38
rs1007279033 Chr.4: 1395692-1395692 on Build GRCh38
rs774281558 Chr.4: 1395610-1395611 on Build GRCh38
rs759616963 Chr.4: 1395552-1395554 on Build GRCh38
rs970076496 Chr.4: 1395983-1395983 on Build GRCh38
rs749745146 Chr.4: 1395606-1395636 on Build GRCh38
rs763834025 Chr.4: 1395655-1395655 on Build GRCh38
rs778224829 Chr.4: 1395878-1395879 on Build GRCh38
rs778232013 Chr.4: 1395585-1395611 on Build GRCh38
rs768959166 Chr.4: 1395873-1395873 on Build GRCh38
rs13137870 Chr.4: 1395552-1395552 on Build GRCh38
rs752794724 Chr.4: 1395553-1395553 on Build GRCh38
rs1022333363 Chr.4: 1395963-1395963 on Build GRCh38
rs764280082 Chr.4: 1395747-1395747 on Build GRCh38
rs746348588 Chr.4: 1395788-1395788 on Build GRCh38
rs771433280 Chr.4: 1395534-1395534 on Build GRCh38
rs781567667 Chr.4: 1395883-1395883 on Build GRCh38
rs757247658 Chr.4: 1395583-1395585 on Build GRCh38
rs571164794 Chr.4: 1395573-1395574 on Build GRCh38
rs747644053 Chr.4: 1395728-1395728 on Build GRCh38
rs765289932 Chr.4: 1395668-1395671 on Build GRCh38
rs746652550 Chr.4: 1395674-1395674 on Build GRCh38
rs771538516 Chr.4: 1395680-1395680 on Build GRCh38
rs754552902 Chr.4: 1395724-1395724 on Build GRCh38
rs779800691 Chr.4: 1395594-1395594 on Build GRCh38
rs779690250 Chr.4: 1395672-1395672 on Build GRCh38
rs748550646 Chr.4: 1395884-1395884 on Build GRCh38
rs370222193 Chr.4: 1395736-1395736 on Build GRCh38
rs765762425 Chr.4: 1395537-1395537 on Build GRCh38
rs769822302 Chr.4: 1395819-1395819 on Build GRCh38
rs776896441 Chr.4: 1395749-1395749 on Build GRCh38
rs35061485 Chr.4: 1395583-1395583 on Build GRCh38
rs766010772 Chr.4: 1395755-1395756 on Build GRCh38
rs946731419 Chr.4: 1395592-1395592 on Build GRCh38
rs767837312 Chr.4: 1395565-1395572 on Build GRCh38
rs981690648 Chr.4: 1395776-1395776 on Build GRCh38
rs1003290305 Chr.4: 1395934-1395934 on Build GRCh38
rs970466516 Chr.4: 1395758-1395758 on Build GRCh38
rs530576391 Chr.4: 1395660-1395660 on Build GRCh38
rs78222806 Chr.4: 1395578-1395578 on Build GRCh38
rs141444125 Chr.4: 1395630-1395630 on Build GRCh38
rs775063341 Chr.4: 1395581-1395581 on Build GRCh38
rs781037370 Chr.4: 1395726-1395726 on Build GRCh38
rs1012982964 Chr.4: 1395867-1395867 on Build GRCh38
rs779733970 Chr.4: 1395717-1395717 on Build GRCh38
rs546704857 Chr.4: 1395923-1395923 on Build GRCh38
rs372615048 Chr.4: 1395712-1395712 on Build GRCh38
rs35263928 Chr.4: 1395587-1395587 on Build GRCh38
rs367966601 Chr.4: 1395759-1395759 on Build GRCh38
rs757976126 Chr.4: 1395669-1395670 on Build GRCh38
rs775733514 Chr.4: 1395637-1395671 on Build GRCh38
rs113374471 Chr.4: 1395620-1395620 on Build GRCh38
rs780992323 Chr.4: 1395773-1395773 on Build GRCh38
rs760907068 Chr.4: 1395647-1395652 on Build GRCh38
rs374903316 Chr.4: 1395730-1395730 on Build GRCh38
rs149222149 Chr.4: 1395693-1395693 on Build GRCh38
rs770234609 Chr.4: 1395604-1395605 on Build GRCh38
rs773132222 Chr.4: 1395832-1395832 on Build GRCh38
rs201846658 Chr.4: 1395647-1395647 on Build GRCh38
rs142260088 Chr.4: 1395783-1395783 on Build GRCh38
rs956215287 Chr.4: 1395907-1395907 on Build GRCh38
rs753884610 Chr.4: 1395699-1395699 on Build GRCh38
rs576663931 Chr.4: 1395605-1395605 on Build GRCh38
rs146895743 Chr.4: 1395840-1395840 on Build GRCh38
rs750331205 Chr.4: 1395714-1395714 on Build GRCh38
rs572687139 Chr.4: 1395546-1395546 on Build GRCh38
rs759198422 Chr.4: 1395531-1395531 on Build GRCh38
rs778701454 Chr.4: 1395569-1395569 on Build GRCh38
rs929181219 Chr.4: 1395830-1395830 on Build GRCh38
rs750009695 Chr.4: 1395586-1395588 on Build GRCh38
rs138375760 Chr.4: 1395694-1395694 on Build GRCh38
rs760500173 Chr.4: 1395574-1395578 on Build GRCh38
rs115895472 Chr.4: 1395750-1395750 on Build GRCh38
rs777524225 Chr.4: 1395852-1395852 on Build GRCh38
rs776775196 Chr.4: 1395813-1395813 on Build GRCh38
rs890772672 Chr.4: 1395820-1395820 on Build GRCh38
rs749873551 Chr.4: 1395668-1395669 on Build GRCh38
rs766807026 Chr.4: 1395614-1395614 on Build GRCh38
rs146370789 Chr.4: 1395566-1395566 on Build GRCh38
rs770877308 Chr.4: 1395534-1395541 on Build GRCh38
rs544074983 Chr.4: 1395811-1395811 on Build GRCh38
rs35321918 Chr.4: 1395651-1395651 on Build GRCh38
rs1015944818 Chr.4: 1395718-1395718 on Build GRCh38
rs773538135 Chr.4: 1395898-1395898 on Build GRCh38
rs777730447 Chr.4: 1395642-1395642 on Build GRCh38
rs868619481 Chr.4: 1395595-1395596 on Build GRCh38
rs557440144 Chr.4: 1395637-1395638 on Build GRCh38
rs770653730 Chr.4: 1395567-1395567 on Build GRCh38
rs763139171 Chr.4: 1395836-1395836 on Build GRCh38
rs753723250 Chr.4: 1395580-1395605 on Build GRCh38
rs766210583 Chr.4: 1395838-1395838 on Build GRCh38
rs746025473 Chr.4: 1395630-1395631 on Build GRCh38
rs142095345 Chr.4: 1395565-1395565 on Build GRCh38
rs956268654 Chr.4: 1395937-1395937 on Build GRCh38
rs754684035 Chr.4: 1395782-1395784 on Build GRCh38
rs759944860 Chr.4: 1395687-1395687 on Build GRCh38
rs751786166 Chr.4: 1395547-1395547 on Build GRCh38
rs375014471 Chr.4: 1395806-1395806 on Build GRCh38
rs750959612 Chr.4: 1395540-1395540 on Build GRCh38
rs752405363 Chr.4: 1395781-1395781 on Build GRCh38
rs777449463 Chr.4: 1395786-1395786 on Build GRCh38
rs139000482 Chr.4: 1395769-1395769 on Build GRCh38
rs13136287 Chr.4: 1395521-1395521 on Build GRCh38
rs773924694 Chr.4: 1395530-1395530 on Build GRCh38
rs375041221 Chr.4: 1395746-1395746 on Build GRCh38
rs778315091 Chr.4: 1395604-1395638 on Build GRCh38
rs142215664 Chr.4: 1395611-1395611 on Build GRCh38
rs543944569 Chr.4: 1395610-1395610 on Build GRCh38
rs763993360 Chr.4: 1395577-1395582 on Build GRCh38
rs202094706 Chr.4: 1395696-1395696 on Build GRCh38
rs138439520 Chr.4: 1395849-1395849 on Build GRCh38
rs774492471 Chr.4: 1395547-1395559 on Build GRCh38
rs879384799 Chr.4: 1395740-1395740 on Build GRCh38
rs745481624 Chr.4: 1395577-1395577 on Build GRCh38
rs771480735 Chr.4: 1395607-1395608 on Build GRCh38
rs758148371 Chr.4: 1395668-1395668 on Build GRCh38
rs550303044 Chr.4: 1395768-1395768 on Build GRCh38
rs747696999 Chr.4: 1395678-1395678 on Build GRCh38
rs760142417 Chr.4: 1395536-1395536 on Build GRCh38
rs552712734 Chr.4: 1395715-1395715 on Build GRCh38
rs147839649 Chr.4: 1395616-1395616 on Build GRCh38
rs776426370 Chr.4: 1395659-1395684 on Build GRCh38
rs767865987 Chr.4: 1395841-1395841 on Build GRCh38
rs758194114 Chr.4: 1395663-1395663 on Build GRCh38
rs201430636 Chr.4: 1395848-1395848 on Build GRCh38
rs766865493 Chr.4: 1395545-1395545 on Build GRCh38
rs148729919 Chr.4: 1395596-1395596 on Build GRCh38
rs751219434 Chr.4: 1395756-1395757 on Build GRCh38
rs13118336 Chr.4: 1395556-1395556 on Build GRCh38
rs115679864 Chr.4: 1395800-1395800 on Build GRCh38
rs769979052 Chr.4: 1395896-1395896 on Build GRCh38
rs993146357 Chr.4: 1395675-1395675 on Build GRCh38
rs569514802 Chr.4: 1395655-1395655 on Build GRCh38
rs770776568 Chr.4: 1395528-1395528 on Build GRCh38
rs773794823 Chr.4: 1395601-1395601 on Build GRCh38
rs534369529 Chr.4: 1395523-1395523 on Build GRCh38
rs776309615 Chr.4: 1395677-1395677 on Build GRCh38
rs745640575 Chr.4: 1395658-1395658 on Build GRCh38
rs199706071 Chr.4: 1395641-1395641 on Build GRCh38
rs376958256 Chr.4: 1395863-1395863 on Build GRCh38

View Full Product Details