Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73075442-73075990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD17
Entrez Gene ID
26057
Gene Name
ankyrin repeat domain 17
Gene Aliases
GTAR, MASK2, NY-BR-16
UniGene
Hs.601206
Gene Chromosome Location
Chr.4: 73073966-73259142 on Build GRCh38
Transcripts
NM_001286771.2 NM_015574.1 NM_032217.4 NM_198889.2 XM_005265671.3 XM_017008011.1 XM_017008012.1 XM_017008013.1

SNP Detail

SNP ID Chromosome Location
rs545108673 Chr.4: 73075501-73075501 on Build GRCh38
rs911645568 Chr.4: 73075653-73075653 on Build GRCh38
rs571885262 Chr.4: 73075911-73075911 on Build GRCh38
rs905826005 Chr.4: 73075807-73075807 on Build GRCh38
rs561102472 Chr.4: 73075767-73075767 on Build GRCh38
rs1040965017 Chr.4: 73075573-73075573 on Build GRCh38
rs113470841 Chr.4: 73075601-73075601 on Build GRCh38
rs972302493 Chr.4: 73075702-73075702 on Build GRCh38
rs758705431 Chr.4: 73075583-73075583 on Build GRCh38
rs937325904 Chr.4: 73075855-73075855 on Build GRCh38
rs755750639 Chr.4: 73075685-73075685 on Build GRCh38
rs987148008 Chr.4: 73075648-73075648 on Build GRCh38
rs528678191 Chr.4: 73075842-73075842 on Build GRCh38
rs563421918 Chr.4: 73075520-73075520 on Build GRCh38
rs778434008 Chr.4: 73075865-73075865 on Build GRCh38
rs926333253 Chr.4: 73075484-73075484 on Build GRCh38
rs1035783213 Chr.4: 73075524-73075524 on Build GRCh38
rs920471276 Chr.4: 73075765-73075765 on Build GRCh38
rs557819151 Chr.4: 73075676-73075676 on Build GRCh38
rs192713686 Chr.4: 73075634-73075634 on Build GRCh38
rs993513280 Chr.4: 73075806-73075806 on Build GRCh38
rs1032220014 Chr.4: 73075724-73075724 on Build GRCh38
rs745338634 Chr.4: 73075927-73075927 on Build GRCh38
rs926364184 Chr.4: 73075960-73075960 on Build GRCh38
rs931153763 Chr.4: 73075487-73075487 on Build GRCh38
rs980411532 Chr.4: 73075955-73075955 on Build GRCh38
rs780400703 Chr.4: 73075620-73075620 on Build GRCh38
rs183765814 Chr.4: 73075915-73075915 on Build GRCh38
rs547266388 Chr.4: 73075904-73075904 on Build GRCh38
rs1020485606 Chr.4: 73075597-73075598 on Build GRCh38
rs151299808 Chr.4: 73075560-73075560 on Build GRCh38
rs917497036 Chr.4: 73075821-73075821 on Build GRCh38
rs750895338 Chr.4: 73075488-73075488 on Build GRCh38
rs777315116 Chr.4: 73075799-73075799 on Build GRCh38
rs901205246 Chr.4: 73075579-73075579 on Build GRCh38
rs930540415 Chr.4: 73075803-73075803 on Build GRCh38
rs747864098 Chr.4: 73075631-73075631 on Build GRCh38
rs770468315 Chr.4: 73075591-73075591 on Build GRCh38
rs956223094 Chr.4: 73075735-73075735 on Build GRCh38
rs930614311 Chr.4: 73075483-73075483 on Build GRCh38

View Full Product Details