Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73741860-73742367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL8
Entrez Gene ID
3576
Gene Name
C-X-C motif chemokine ligand 8
Gene Aliases
GCP-1, GCP1, IL8, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1
UniGene
Hs.624
Gene Chromosome Location
Chr.4: 73740506-73743716 on Build GRCh38
Transcripts
NM_000584.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820796 c.232C>A p.L78M Chr.4: 73741980-73741980 on Build GRCh38
6224433 c.239C>T p.P80L Chr.4: 73741987-73741987 on Build GRCh38
4585176 c.231T>A p.C77* Chr.4: 73741979-73741979 on Build GRCh38
2831359 c.214G>A p.D72N Chr.4: 73741962-73741962 on Build GRCh38
4446223 c.201-3C>T p.? Chr.4: 73741946-73741946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770716928 Chr.4: 73741935-73741935 on Build GRCh38
rs966747236 Chr.4: 73742284-73742284 on Build GRCh38
rs913548486 Chr.4: 73741925-73741925 on Build GRCh38
rs759066024 Chr.4: 73742054-73742054 on Build GRCh38
rs941595131 Chr.4: 73742116-73742116 on Build GRCh38
rs769138009 Chr.4: 73742058-73742058 on Build GRCh38
rs773747524 Chr.4: 73741922-73741922 on Build GRCh38
rs942468226 Chr.4: 73742162-73742162 on Build GRCh38
rs957928491 Chr.4: 73742213-73742213 on Build GRCh38
rs952593231 Chr.4: 73741930-73741930 on Build GRCh38
rs776188727 Chr.4: 73742045-73742045 on Build GRCh38
rs552848872 Chr.4: 73742166-73742166 on Build GRCh38
rs780209935 Chr.4: 73742000-73742000 on Build GRCh38
rs897145927 Chr.4: 73742157-73742157 on Build GRCh38
rs759032011 Chr.4: 73741955-73741955 on Build GRCh38
rs1046708263 Chr.4: 73742304-73742304 on Build GRCh38
rs990598523 Chr.4: 73742318-73742318 on Build GRCh38
rs2227543 Chr.4: 73742193-73742193 on Build GRCh38
rs866758378 Chr.4: 73742125-73742125 on Build GRCh38
rs200662278 Chr.4: 73742026-73742026 on Build GRCh38
rs1020875203 Chr.4: 73742236-73742236 on Build GRCh38
rs201577790 Chr.4: 73742052-73742052 on Build GRCh38
rs573569793 Chr.4: 73742065-73742065 on Build GRCh38
rs748504611 Chr.4: 73741919-73741919 on Build GRCh38
rs998204273 Chr.4: 73742173-73742173 on Build GRCh38
rs1037655202 Chr.4: 73742330-73742330 on Build GRCh38
rs1034185009 Chr.4: 73742328-73742328 on Build GRCh38
rs199753152 Chr.4: 73742152-73742152 on Build GRCh38
rs201751726 Chr.4: 73742150-73742150 on Build GRCh38
rs199793724 Chr.4: 73742059-73742059 on Build GRCh38
rs201374695 Chr.4: 73741931-73741931 on Build GRCh38
rs531811301 Chr.4: 73742210-73742210 on Build GRCh38
rs747439661 Chr.4: 73742040-73742040 on Build GRCh38
rs796293296 Chr.4: 73742141-73742141 on Build GRCh38
rs746753139 Chr.4: 73741923-73741937 on Build GRCh38
rs1005457503 Chr.4: 73742321-73742321 on Build GRCh38
rs752874546 Chr.4: 73741929-73741929 on Build GRCh38
rs981788462 Chr.4: 73742285-73742285 on Build GRCh38
rs747764679 Chr.4: 73741999-73741999 on Build GRCh38
rs747217394 Chr.4: 73742071-73742071 on Build GRCh38
rs373408845 Chr.4: 73741997-73741997 on Build GRCh38
rs188973626 Chr.4: 73742248-73742248 on Build GRCh38
rs200382511 Chr.4: 73741916-73741916 on Build GRCh38
rs772520102 Chr.4: 73741920-73741920 on Build GRCh38
rs911387481 Chr.4: 73742135-73742135 on Build GRCh38
rs751369405 Chr.4: 73741973-73741973 on Build GRCh38
rs753921688 Chr.4: 73742008-73742008 on Build GRCh38
rs371104442 Chr.4: 73741964-73741964 on Build GRCh38
rs1029673722 Chr.4: 73742180-73742180 on Build GRCh38
rs1055321050 Chr.4: 73742048-73742048 on Build GRCh38
rs772706294 Chr.4: 73742030-73742030 on Build GRCh38
rs762899923 Chr.4: 73741957-73741957 on Build GRCh38
rs989799301 Chr.4: 73742214-73742214 on Build GRCh38
rs185040023 Chr.4: 73742029-73742029 on Build GRCh38
rs201799579 Chr.4: 73742161-73742161 on Build GRCh38
rs756294837 Chr.4: 73742009-73742009 on Build GRCh38
rs910033622 Chr.4: 73742320-73742320 on Build GRCh38
rs778229177 Chr.4: 73742033-73742033 on Build GRCh38
rs540883353 Chr.4: 73742126-73742126 on Build GRCh38
rs770615996 Chr.4: 73742042-73742042 on Build GRCh38
rs369608699 Chr.4: 73742077-73742077 on Build GRCh38
rs768729285 Chr.4: 73741972-73741973 on Build GRCh38
rs927676317 Chr.4: 73742287-73742287 on Build GRCh38
rs917918028 Chr.4: 73741968-73741968 on Build GRCh38
rs779068762 Chr.4: 73742011-73742011 on Build GRCh38
rs888645849 Chr.4: 73742113-73742113 on Build GRCh38
rs780925576 Chr.4: 73741983-73741984 on Build GRCh38
rs199638674 Chr.4: 73741970-73741970 on Build GRCh38
rs990861106 Chr.4: 73741961-73741961 on Build GRCh38
rs562080412 Chr.4: 73742329-73742329 on Build GRCh38
rs375781159 Chr.4: 73742161-73742161 on Build GRCh38
rs888388655 Chr.4: 73742315-73742315 on Build GRCh38
rs970666841 Chr.4: 73742063-73742063 on Build GRCh38
rs370518457 Chr.4: 73742015-73742015 on Build GRCh38
rs369603475 Chr.4: 73742152-73742152 on Build GRCh38
rs181810989 Chr.4: 73741886-73741886 on Build GRCh38
rs147230462 Chr.4: 73742188-73742188 on Build GRCh38
rs563373543 Chr.4: 73742197-73742197 on Build GRCh38
rs937726590 Chr.4: 73742290-73742290 on Build GRCh38
rs536774132 Chr.4: 73741979-73741979 on Build GRCh38
rs776187785 Chr.4: 73741946-73741946 on Build GRCh38
rs764075530 Chr.4: 73742085-73742085 on Build GRCh38

View Full Product Details