Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76217259-76217756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47E
Entrez Gene ID
100129583
Gene Name
family with sequence similarity 47 member E
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76214040-76283783 on Build GRCh38
Transcripts
NM_001242936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6038746 c.3G>C p.M1I Chr.4: 76217610-76217610 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187578340 Chr.4: 76217445-76217445 on Build GRCh38
rs987966048 Chr.4: 76217458-76217458 on Build GRCh38
rs954785956 Chr.4: 76217427-76217427 on Build GRCh38
rs936575909 Chr.4: 76217535-76217535 on Build GRCh38
rs76357627 Chr.4: 76217545-76217545 on Build GRCh38
rs1022254907 Chr.4: 76217290-76217290 on Build GRCh38
rs892295383 Chr.4: 76217653-76217653 on Build GRCh38
rs759014095 Chr.4: 76217712-76217712 on Build GRCh38
rs768429191 Chr.4: 76217650-76217650 on Build GRCh38
rs576816603 Chr.4: 76217627-76217627 on Build GRCh38
rs34763038 Chr.4: 76217716-76217717 on Build GRCh38
rs1002367459 Chr.4: 76217674-76217674 on Build GRCh38
rs779089305 Chr.4: 76217682-76217682 on Build GRCh38
rs975848093 Chr.4: 76217331-76217331 on Build GRCh38
rs1866975 Chr.4: 76217505-76217505 on Build GRCh38
rs903806576 Chr.4: 76217529-76217529 on Build GRCh38
rs868001229 Chr.4: 76217470-76217470 on Build GRCh38
rs751406800 Chr.4: 76217606-76217606 on Build GRCh38
rs1015824149 Chr.4: 76217707-76217707 on Build GRCh38
rs922986880 Chr.4: 76217396-76217396 on Build GRCh38
rs576172910 Chr.4: 76217612-76217612 on Build GRCh38
rs948324473 Chr.4: 76217518-76217518 on Build GRCh38
rs569202794 Chr.4: 76217720-76217720 on Build GRCh38
rs776335160 Chr.4: 76217689-76217689 on Build GRCh38
rs766694425 Chr.4: 76217625-76217625 on Build GRCh38
rs983420973 Chr.4: 76217706-76217706 on Build GRCh38
rs556927579 Chr.4: 76217644-76217644 on Build GRCh38
rs908050308 Chr.4: 76217476-76217476 on Build GRCh38
rs562623143 Chr.4: 76217651-76217651 on Build GRCh38
rs920608 Chr.4: 76217307-76217307 on Build GRCh38
rs953021175 Chr.4: 76217691-76217691 on Build GRCh38
rs755124881 Chr.4: 76217665-76217665 on Build GRCh38
rs987567362 Chr.4: 76217728-76217730 on Build GRCh38
rs984508670 Chr.4: 76217305-76217305 on Build GRCh38
rs576496781 Chr.4: 76217721-76217721 on Build GRCh38
rs545838272 Chr.4: 76217629-76217629 on Build GRCh38
rs753290145 Chr.4: 76217701-76217701 on Build GRCh38
rs997177384 Chr.4: 76217715-76217715 on Build GRCh38
rs921943370 Chr.4: 76217415-76217415 on Build GRCh38
rs193290840 Chr.4: 76217482-76217482 on Build GRCh38
rs771929968 Chr.4: 76217319-76217319 on Build GRCh38
rs1052472702 Chr.4: 76217543-76217543 on Build GRCh38
rs920870749 Chr.4: 76217509-76217509 on Build GRCh38
rs1044707894 Chr.4: 76217568-76217568 on Build GRCh38
rs974792337 Chr.4: 76217364-76217364 on Build GRCh38
rs905248744 Chr.4: 76217655-76217655 on Build GRCh38
rs761503157 Chr.4: 76217624-76217624 on Build GRCh38

View Full Product Details