Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76271483-76271978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47E
Entrez Gene ID
100129583
Gene Name
family with sequence similarity 47 member E
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76214040-76283783 on Build GRCh38
Transcripts
NM_001136570.2 NM_001242936.1 NM_001242939.1

Gene Symbol
FAM47E-STBD1
Entrez Gene ID
100631383
Gene Name
FAM47E-STBD1 readthrough
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76251700-76311130 on Build GRCh38
Transcripts
NM_001136570.2 NM_001242936.1 NM_001242939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5425368 c.523G>A p.E175K Chr.4: 76271715-76271715 on Build GRCh38
5401572 c.390C>T p.I130I Chr.4: 76271582-76271582 on Build GRCh38
1431076 c.755C>T p.T252M Chr.4: 76271653-76271653 on Build GRCh38
1057652 c.397G>T p.E133* Chr.4: 76271589-76271589 on Build GRCh38
2832123 c.728C>T p.T243I Chr.4: 76271626-76271626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008094819 Chr.4: 76271576-76271576 on Build GRCh38
rs3733252 Chr.4: 76271575-76271575 on Build GRCh38
rs189574767 Chr.4: 76271942-76271942 on Build GRCh38
rs960123621 Chr.4: 76271683-76271683 on Build GRCh38
rs963871498 Chr.4: 76271582-76271582 on Build GRCh38
rs916083372 Chr.4: 76271689-76271689 on Build GRCh38
rs534203291 Chr.4: 76271764-76271764 on Build GRCh38
rs141636919 Chr.4: 76271854-76271854 on Build GRCh38
rs977254577 Chr.4: 76271660-76271660 on Build GRCh38
rs755809691 Chr.4: 76271693-76271693 on Build GRCh38
rs1039731373 Chr.4: 76271740-76271740 on Build GRCh38
rs774617220 Chr.4: 76271604-76271604 on Build GRCh38
rs971153868 Chr.4: 76271907-76271907 on Build GRCh38
rs187862162 Chr.4: 76271530-76271530 on Build GRCh38
rs577390613 Chr.4: 76271806-76271806 on Build GRCh38
rs772084828 Chr.4: 76271619-76271619 on Build GRCh38
rs546368201 Chr.4: 76271849-76271849 on Build GRCh38
rs924496883 Chr.4: 76271667-76271667 on Build GRCh38
rs768804874 Chr.4: 76271596-76271596 on Build GRCh38
rs56071515 Chr.4: 76271869-76271869 on Build GRCh38
rs529912787 Chr.4: 76271557-76271557 on Build GRCh38
rs958649133 Chr.4: 76271886-76271886 on Build GRCh38
rs77440835 Chr.4: 76271894-76271894 on Build GRCh38
rs949830253 Chr.4: 76271654-76271654 on Build GRCh38
rs183715787 Chr.4: 76271904-76271904 on Build GRCh38
rs191481797 Chr.4: 76271639-76271639 on Build GRCh38
rs777367803 Chr.4: 76271801-76271801 on Build GRCh38
rs576562392 Chr.4: 76271874-76271874 on Build GRCh38
rs371976589 Chr.4: 76271583-76271583 on Build GRCh38
rs202038650 Chr.4: 76271678-76271678 on Build GRCh38
rs3733253 Chr.4: 76271535-76271535 on Build GRCh38
rs995157575 Chr.4: 76271547-76271547 on Build GRCh38
rs761862662 Chr.4: 76271613-76271613 on Build GRCh38
rs977221464 Chr.4: 76271930-76271930 on Build GRCh38
rs3733250 Chr.4: 76271715-76271715 on Build GRCh38
rs1016597872 Chr.4: 76271856-76271856 on Build GRCh38
rs901269374 Chr.4: 76271775-76271775 on Build GRCh38
rs975752078 Chr.4: 76271600-76271600 on Build GRCh38
rs948886965 Chr.4: 76271697-76271697 on Build GRCh38
rs767566218 Chr.4: 76271881-76271881 on Build GRCh38
rs1008394481 Chr.4: 76271875-76271875 on Build GRCh38
rs751577886 Chr.4: 76271816-76271816 on Build GRCh38
rs1021539514 Chr.4: 76271902-76271902 on Build GRCh38
rs899675596 Chr.4: 76271885-76271885 on Build GRCh38
rs754205276 Chr.4: 76271688-76271688 on Build GRCh38
rs574751211 Chr.4: 76271723-76271723 on Build GRCh38
rs998287271 Chr.4: 76271778-76271778 on Build GRCh38
rs1019872926 Chr.4: 76271581-76271581 on Build GRCh38
rs977392038 Chr.4: 76271655-76271655 on Build GRCh38
rs983775701 Chr.4: 76271541-76271541 on Build GRCh38
rs554242487 Chr.4: 76271793-76271793 on Build GRCh38
rs769457260 Chr.4: 76271646-76271646 on Build GRCh38
rs3733251 Chr.4: 76271685-76271685 on Build GRCh38
rs542480339 Chr.4: 76271880-76271880 on Build GRCh38
rs375151279 Chr.4: 76271637-76271637 on Build GRCh38
rs538371670 Chr.4: 76271570-76271570 on Build GRCh38
rs757298712 Chr.4: 76271818-76271818 on Build GRCh38
rs868746214 Chr.4: 76271919-76271919 on Build GRCh38
rs752328576 Chr.4: 76271792-76271792 on Build GRCh38
rs1021511555 Chr.4: 76271609-76271609 on Build GRCh38
rs893820799 Chr.4: 76271711-76271711 on Build GRCh38
rs867217004 Chr.4: 76271653-76271653 on Build GRCh38

View Full Product Details