Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77725980-77726480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT6L
Entrez Gene ID
246175
Gene Name
CCR4-NOT transcription complex subunit 6 like
Gene Aliases
CCR4b
UniGene
Hs.592519
Gene Chromosome Location
Chr.4: 77713387-77820238 on Build GRCh38
Transcripts
NM_001286790.1 NM_144571.2 XM_011531806.2 XM_011531807.2 XM_011531808.2 XM_011531809.1 XM_011531810.1 XM_017007956.1 XM_017007957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219207 c.1305C>T p.F435F Chr.4: 77726317-77726317 on Build GRCh38
7429971 c.1403G>A p.R468K Chr.4: 77726306-77726306 on Build GRCh38
7429972 c.1316G>A p.R439K Chr.4: 77726306-77726306 on Build GRCh38
734613 c.1447G>T p.G483* Chr.4: 77726262-77726262 on Build GRCh38
6219208 c.1392C>T p.F464F Chr.4: 77726317-77726317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958736998 Chr.4: 77726055-77726055 on Build GRCh38
rs759094417 Chr.4: 77726116-77726116 on Build GRCh38
rs773682070 Chr.4: 77726095-77726095 on Build GRCh38
rs772685591 Chr.4: 77726258-77726258 on Build GRCh38
rs774836591 Chr.4: 77726308-77726308 on Build GRCh38
rs751375348 Chr.4: 77726144-77726144 on Build GRCh38
rs762247295 Chr.4: 77726090-77726091 on Build GRCh38
rs776386186 Chr.4: 77726115-77726115 on Build GRCh38
rs990254002 Chr.4: 77726417-77726417 on Build GRCh38
rs757003627 Chr.4: 77726156-77726156 on Build GRCh38
rs769417199 Chr.4: 77726302-77726302 on Build GRCh38
rs572736871 Chr.4: 77726288-77726288 on Build GRCh38
rs755124333 Chr.4: 77726395-77726395 on Build GRCh38
rs778853656 Chr.4: 77726209-77726209 on Build GRCh38
rs554703151 Chr.4: 77726034-77726034 on Build GRCh38
rs763896530 Chr.4: 77726335-77726335 on Build GRCh38
rs777177292 Chr.4: 77726071-77726073 on Build GRCh38
rs959649045 Chr.4: 77726433-77726433 on Build GRCh38
rs760454461 Chr.4: 77726102-77726102 on Build GRCh38
rs759101214 Chr.4: 77726300-77726300 on Build GRCh38
rs377046330 Chr.4: 77726202-77726202 on Build GRCh38
rs766154872 Chr.4: 77726104-77726104 on Build GRCh38
rs748589918 Chr.4: 77726085-77726085 on Build GRCh38
rs893910891 Chr.4: 77726148-77726148 on Build GRCh38
rs146657222 Chr.4: 77726440-77726440 on Build GRCh38
rs780258014 Chr.4: 77726182-77726182 on Build GRCh38
rs889130237 Chr.4: 77726089-77726089 on Build GRCh38
rs1016362193 Chr.4: 77726112-77726112 on Build GRCh38
rs562772355 Chr.4: 77726256-77726256 on Build GRCh38
rs778376366 Chr.4: 77726259-77726259 on Build GRCh38
rs767285973 Chr.4: 77726380-77726380 on Build GRCh38
rs751430489 Chr.4: 77726374-77726374 on Build GRCh38
rs143289121 Chr.4: 77726403-77726403 on Build GRCh38
rs774935909 Chr.4: 77726069-77726069 on Build GRCh38
rs761608331 Chr.4: 77726375-77726375 on Build GRCh38
rs559331159 Chr.4: 77726273-77726273 on Build GRCh38
rs573698776 Chr.4: 77726038-77726038 on Build GRCh38
rs763922103 Chr.4: 77726125-77726125 on Build GRCh38
rs764598455 Chr.4: 77726117-77726117 on Build GRCh38
rs752741620 Chr.4: 77726120-77726120 on Build GRCh38
rs778431465 Chr.4: 77726419-77726419 on Build GRCh38
rs752287597 Chr.4: 77726014-77726014 on Build GRCh38
rs755143372 Chr.4: 77726208-77726208 on Build GRCh38
rs542331649 Chr.4: 77726094-77726094 on Build GRCh38
rs755572143 Chr.4: 77726212-77726212 on Build GRCh38
rs779099390 Chr.4: 77726397-77726397 on Build GRCh38
rs754155884 Chr.4: 77726390-77726390 on Build GRCh38
rs942396808 Chr.4: 77726396-77726396 on Build GRCh38
rs923471294 Chr.4: 77726168-77726168 on Build GRCh38
rs747411145 Chr.4: 77726264-77726264 on Build GRCh38
rs758515070 Chr.4: 77726412-77726412 on Build GRCh38
rs765647806 Chr.4: 77726131-77726133 on Build GRCh38

View Full Product Details