Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79407072-79407572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK2
Entrez Gene ID
2712
Gene Name
glycerol kinase 2
Gene Aliases
GKP2, GKTA
UniGene
Hs.98008
Gene Chromosome Location
Chr.4: 79406353-79408218 on Build GRCh38
Transcripts
NM_033214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369499 c.1118A>T p.E373V Chr.4: 79407083-79407083 on Build GRCh38
2954984 c.871C>T p.R291C Chr.4: 79407330-79407330 on Build GRCh38
1058002 c.933G>T p.E311D Chr.4: 79407268-79407268 on Build GRCh38
2954982 c.958G>T p.G320C Chr.4: 79407243-79407243 on Build GRCh38
1058000 c.1040G>T p.R347I Chr.4: 79407161-79407161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866917714 Chr.4: 79407268-79407268 on Build GRCh38
rs773092299 Chr.4: 79407118-79407118 on Build GRCh38
rs764827524 Chr.4: 79407280-79407280 on Build GRCh38
rs551863127 Chr.4: 79407452-79407452 on Build GRCh38
rs747256111 Chr.4: 79407521-79407521 on Build GRCh38
rs7673905 Chr.4: 79407163-79407163 on Build GRCh38
rs747758992 Chr.4: 79407342-79407342 on Build GRCh38
rs201459558 Chr.4: 79407402-79407402 on Build GRCh38
rs757517049 Chr.4: 79407160-79407160 on Build GRCh38
rs888732535 Chr.4: 79407497-79407497 on Build GRCh38
rs114287210 Chr.4: 79407365-79407365 on Build GRCh38
rs776671840 Chr.4: 79407254-79407254 on Build GRCh38
rs764765869 Chr.4: 79407144-79407144 on Build GRCh38
rs776385662 Chr.4: 79407426-79407426 on Build GRCh38
rs137991824 Chr.4: 79407109-79407109 on Build GRCh38
rs777950818 Chr.4: 79407520-79407520 on Build GRCh38
rs770762499 Chr.4: 79407421-79407421 on Build GRCh38
rs200729434 Chr.4: 79407229-79407229 on Build GRCh38
rs773002678 Chr.4: 79407251-79407251 on Build GRCh38
rs774192509 Chr.4: 79407454-79407454 on Build GRCh38
rs753333072 Chr.4: 79407128-79407128 on Build GRCh38
rs755520714 Chr.4: 79407483-79407483 on Build GRCh38
rs749293008 Chr.4: 79407197-79407197 on Build GRCh38
rs772393135 Chr.4: 79407221-79407226 on Build GRCh38
rs200474130 Chr.4: 79407310-79407310 on Build GRCh38
rs767939060 Chr.4: 79407323-79407323 on Build GRCh38
rs144129402 Chr.4: 79407430-79407430 on Build GRCh38
rs1046513189 Chr.4: 79407394-79407394 on Build GRCh38
rs201151701 Chr.4: 79407446-79407446 on Build GRCh38
rs749473478 Chr.4: 79407096-79407096 on Build GRCh38
rs781716007 Chr.4: 79407167-79407167 on Build GRCh38
rs149474069 Chr.4: 79407212-79407212 on Build GRCh38
rs774967537 Chr.4: 79407308-79407308 on Build GRCh38
rs947564396 Chr.4: 79407155-79407155 on Build GRCh38
rs937575078 Chr.4: 79407224-79407224 on Build GRCh38
rs757007774 Chr.4: 79407522-79407522 on Build GRCh38
rs761488569 Chr.4: 79407107-79407107 on Build GRCh38
rs758948027 Chr.4: 79407265-79407265 on Build GRCh38
rs367710479 Chr.4: 79407357-79407357 on Build GRCh38
rs769121531 Chr.4: 79407428-79407428 on Build GRCh38
rs754388739 Chr.4: 79407335-79407335 on Build GRCh38
rs755276822 Chr.4: 79407179-79407179 on Build GRCh38
rs770593115 Chr.4: 79407288-79407289 on Build GRCh38
rs766967391 Chr.4: 79407334-79407334 on Build GRCh38
rs143875620 Chr.4: 79407329-79407329 on Build GRCh38
rs368611623 Chr.4: 79407406-79407406 on Build GRCh38
rs375896928 Chr.4: 79407537-79407537 on Build GRCh38
rs369528192 Chr.4: 79407303-79407303 on Build GRCh38
rs745851913 Chr.4: 79407427-79407427 on Build GRCh38
rs752444892 Chr.4: 79407145-79407145 on Build GRCh38
rs748431542 Chr.4: 79407215-79407215 on Build GRCh38
rs532948584 Chr.4: 79407124-79407124 on Build GRCh38
rs995315843 Chr.4: 79407418-79407418 on Build GRCh38
rs150499437 Chr.4: 79407383-79407383 on Build GRCh38
rs746776214 Chr.4: 79407408-79407408 on Build GRCh38
rs779052388 Chr.4: 79407355-79407355 on Build GRCh38
rs374732833 Chr.4: 79407181-79407181 on Build GRCh38
rs200048190 Chr.4: 79407487-79407487 on Build GRCh38
rs756583364 Chr.4: 79407331-79407331 on Build GRCh38
rs765816887 Chr.4: 79407127-79407127 on Build GRCh38
rs199687527 Chr.4: 79407306-79407306 on Build GRCh38
rs774670514 Chr.4: 79407098-79407098 on Build GRCh38
rs768751739 Chr.4: 79407097-79407097 on Build GRCh38
rs759152060 Chr.4: 79407138-79407138 on Build GRCh38
rs988949687 Chr.4: 79407091-79407091 on Build GRCh38
rs371750502 Chr.4: 79407135-79407135 on Build GRCh38
rs766794566 Chr.4: 79407463-79407463 on Build GRCh38
rs758512798 Chr.4: 79407515-79407515 on Build GRCh38
rs549791960 Chr.4: 79407202-79407202 on Build GRCh38
rs754229774 Chr.4: 79407472-79407472 on Build GRCh38
rs761068317 Chr.4: 79407457-79407457 on Build GRCh38
rs746864179 Chr.4: 79407253-79407253 on Build GRCh38
rs745737278 Chr.4: 79407544-79407544 on Build GRCh38
rs199938688 Chr.4: 79407223-79407224 on Build GRCh38
rs780767905 Chr.4: 79407185-79407185 on Build GRCh38

View Full Product Details