Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83269895-83270444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ2
Entrez Gene ID
27235
Gene Name
coenzyme Q2, polyprenyltransferase
Gene Aliases
CL640, COQ10D1, MSA1, PHB:PPT
UniGene
Hs.144304
Gene Chromosome Location
Chr.4: 83138568-83285129 on Build GRCh38
Transcripts
NM_015697.7 XM_011531855.2 XM_011531857.2 XM_011531859.2 XM_011531860.2 XM_011531862.2 XM_011531863.2 XM_011531866.2 XM_011531867.2 XM_017008031.1 XM_017008032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202086 c.800C>T p.A267V Chr.4: 83269972-83269972 on Build GRCh38
6634137 c.779G>A p.G260D Chr.4: 83269993-83269993 on Build GRCh38
6634136 c.830C>T p.S277F Chr.4: 83269942-83269942 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150559207 Chr.4: 83270067-83270067 on Build GRCh38
rs778664698 Chr.4: 83270017-83270017 on Build GRCh38
rs755258336 Chr.4: 83270010-83270010 on Build GRCh38
rs955658894 Chr.4: 83270145-83270145 on Build GRCh38
rs537545211 Chr.4: 83270399-83270399 on Build GRCh38
rs530261089 Chr.4: 83270286-83270286 on Build GRCh38
rs565282820 Chr.4: 83270148-83270148 on Build GRCh38
rs928256861 Chr.4: 83270407-83270407 on Build GRCh38
rs571717352 Chr.4: 83269980-83269980 on Build GRCh38
rs553681443 Chr.4: 83269946-83269946 on Build GRCh38
rs937288596 Chr.4: 83270418-83270418 on Build GRCh38
rs569982552 Chr.4: 83270300-83270300 on Build GRCh38
rs750974987 Chr.4: 83269973-83269973 on Build GRCh38
rs780750066 Chr.4: 83270344-83270345 on Build GRCh38
rs188166189 Chr.4: 83270197-83270197 on Build GRCh38
rs181399056 Chr.4: 83270081-83270081 on Build GRCh38
rs972262918 Chr.4: 83270381-83270381 on Build GRCh38
rs1040208483 Chr.4: 83270112-83270112 on Build GRCh38
rs759303117 Chr.4: 83269924-83269924 on Build GRCh38
rs368255651 Chr.4: 83270029-83270029 on Build GRCh38
rs945488145 Chr.4: 83270160-83270160 on Build GRCh38
rs774916508 Chr.4: 83269952-83269952 on Build GRCh38
rs183694694 Chr.4: 83270099-83270099 on Build GRCh38
rs756220409 Chr.4: 83270100-83270100 on Build GRCh38
rs532495119 Chr.4: 83270194-83270194 on Build GRCh38
rs762284326 Chr.4: 83269962-83269962 on Build GRCh38
rs566123221 Chr.4: 83270260-83270260 on Build GRCh38
rs981131032 Chr.4: 83270383-83270383 on Build GRCh38
rs1025174840 Chr.4: 83270303-83270303 on Build GRCh38
rs754108924 Chr.4: 83270264-83270264 on Build GRCh38
rs755309606 Chr.4: 83270265-83270265 on Build GRCh38
rs370821735 Chr.4: 83270030-83270030 on Build GRCh38
rs928606758 Chr.4: 83269992-83269992 on Build GRCh38
rs895415764 Chr.4: 83270296-83270296 on Build GRCh38
rs1026065799 Chr.4: 83270397-83270397 on Build GRCh38
rs542921054 Chr.4: 83270066-83270066 on Build GRCh38
rs767691112 Chr.4: 83270093-83270093 on Build GRCh38
rs773721095 Chr.4: 83270345-83270345 on Build GRCh38
rs180922948 Chr.4: 83270294-83270294 on Build GRCh38
rs961557971 Chr.4: 83270348-83270348 on Build GRCh38
rs766526545 Chr.4: 83269999-83269999 on Build GRCh38
rs984824029 Chr.4: 83270053-83270053 on Build GRCh38
rs779286987 Chr.4: 83270316-83270316 on Build GRCh38
rs777531990 Chr.4: 83270036-83270036 on Build GRCh38
rs930323146 Chr.4: 83270045-83270045 on Build GRCh38
rs549465544 Chr.4: 83270416-83270416 on Build GRCh38
rs754026311 Chr.4: 83270004-83270004 on Build GRCh38
rs1000883263 Chr.4: 83270283-83270283 on Build GRCh38
rs937215195 Chr.4: 83269993-83269996 on Build GRCh38
rs1005658089 Chr.4: 83270048-83270048 on Build GRCh38
rs1018465111 Chr.4: 83270244-83270244 on Build GRCh38
rs887578141 Chr.4: 83270046-83270046 on Build GRCh38
rs374610954 Chr.4: 83270028-83270028 on Build GRCh38
rs199581249 Chr.4: 83269971-83269971 on Build GRCh38
rs781009196 Chr.4: 83270012-83270012 on Build GRCh38
rs537360300 Chr.4: 83270314-83270314 on Build GRCh38

View Full Product Details