Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175443340-175443843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD1
Entrez Gene ID
1812
Gene Name
dopamine receptor D1
Gene Aliases
DADR, DRD1A
UniGene
Hs.2624
Gene Chromosome Location
Chr.5: 175440672-175444160 on Build GRCh38
Transcripts
NM_000794.3

SNP Detail

SNP ID Chromosome Location
rs916807984 Chr.5: 175443635-175443635 on Build GRCh38
rs977159937 Chr.5: 175443570-175443570 on Build GRCh38
rs557968071 Chr.5: 175443587-175443587 on Build GRCh38
rs994263551 Chr.5: 175443429-175443429 on Build GRCh38
rs1044050324 Chr.5: 175443664-175443664 on Build GRCh38
rs775070467 Chr.5: 175443469-175443469 on Build GRCh38
rs1027124229 Chr.5: 175443439-175443439 on Build GRCh38
rs568825406 Chr.5: 175443472-175443472 on Build GRCh38
rs912571196 Chr.5: 175443533-175443533 on Build GRCh38
rs533873033 Chr.5: 175443670-175443670 on Build GRCh38
rs577344141 Chr.5: 175443736-175443736 on Build GRCh38
rs185064274 Chr.5: 175443405-175443405 on Build GRCh38
rs749661152 Chr.5: 175443487-175443487 on Build GRCh38
rs936821390 Chr.5: 175443622-175443622 on Build GRCh38
rs929746769 Chr.5: 175443775-175443775 on Build GRCh38
rs1003030214 Chr.5: 175443365-175443365 on Build GRCh38
rs1026779714 Chr.5: 175443708-175443708 on Build GRCh38
rs11742764 Chr.5: 175443790-175443790 on Build GRCh38
rs1031049545 Chr.5: 175443614-175443614 on Build GRCh38
rs749920467 Chr.5: 175443550-175443550 on Build GRCh38
rs557207143 Chr.5: 175443433-175443433 on Build GRCh38
rs938420509 Chr.5: 175443380-175443380 on Build GRCh38
rs201089398 Chr.5: 175443802-175443802 on Build GRCh38
rs879891253 Chr.5: 175443637-175443637 on Build GRCh38
rs906911152 Chr.5: 175443362-175443362 on Build GRCh38
rs758917277 Chr.5: 175443804-175443804 on Build GRCh38
rs889667359 Chr.5: 175443729-175443729 on Build GRCh38
rs370742648 Chr.5: 175443656-175443656 on Build GRCh38
rs938386537 Chr.5: 175443678-175443678 on Build GRCh38
rs1037177608 Chr.5: 175443680-175443680 on Build GRCh38
rs1057211018 Chr.5: 175443688-175443688 on Build GRCh38
rs914430370 Chr.5: 175443636-175443636 on Build GRCh38
rs571417062 Chr.5: 175443402-175443402 on Build GRCh38
rs193082222 Chr.5: 175443683-175443683 on Build GRCh38
rs1019528241 Chr.5: 175443789-175443789 on Build GRCh38
rs554147045 Chr.5: 175443569-175443569 on Build GRCh38
rs995723721 Chr.5: 175443707-175443707 on Build GRCh38
rs965441557 Chr.5: 175443546-175443546 on Build GRCh38
rs535817442 Chr.5: 175443539-175443539 on Build GRCh38
rs190667188 Chr.5: 175443623-175443623 on Build GRCh38
rs544511033 Chr.5: 175443796-175443796 on Build GRCh38
rs1026830886 Chr.5: 175443478-175443478 on Build GRCh38
rs985389654 Chr.5: 175443496-175443496 on Build GRCh38

View Full Product Details