Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176642578-176643069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E1B
Entrez Gene ID
253314
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
-
UniGene
Hs.448362
Gene Chromosome Location
Chr.5: 176630613-176646644 on Build GRCh38
Transcripts
NM_001099408.1 XM_011534499.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470209 c.14A>T p.E5V Chr.5: 176642801-176642801 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868274689 Chr.5: 176642861-176642861 on Build GRCh38
rs375479409 Chr.5: 176642611-176642611 on Build GRCh38
rs924233097 Chr.5: 176642654-176642654 on Build GRCh38
rs963703253 Chr.5: 176642710-176642710 on Build GRCh38
rs759943190 Chr.5: 176642828-176642828 on Build GRCh38
rs1043121776 Chr.5: 176642855-176642855 on Build GRCh38
rs371120970 Chr.5: 176642651-176642651 on Build GRCh38
rs769851561 Chr.5: 176642824-176642824 on Build GRCh38
rs373982575 Chr.5: 176642612-176642612 on Build GRCh38
rs13176195 Chr.5: 176643003-176643003 on Build GRCh38
rs913849408 Chr.5: 176643014-176643014 on Build GRCh38
rs753258108 Chr.5: 176643036-176643036 on Build GRCh38
rs185021288 Chr.5: 176642827-176642827 on Build GRCh38
rs1053819950 Chr.5: 176642682-176642682 on Build GRCh38
rs757582071 Chr.5: 176642676-176642676 on Build GRCh38
rs200462680 Chr.5: 176642849-176642851 on Build GRCh38
rs535567370 Chr.5: 176642709-176642709 on Build GRCh38
rs938488341 Chr.5: 176642934-176642934 on Build GRCh38
rs965524592 Chr.5: 176642647-176642647 on Build GRCh38
rs901740 Chr.5: 176642830-176642830 on Build GRCh38
rs953864118 Chr.5: 176642831-176642831 on Build GRCh38
rs1036896933 Chr.5: 176642960-176642960 on Build GRCh38
rs189032522 Chr.5: 176642965-176642965 on Build GRCh38
rs776822137 Chr.5: 176642728-176642729 on Build GRCh38
rs912708236 Chr.5: 176642640-176642640 on Build GRCh38
rs937310536 Chr.5: 176642857-176642858 on Build GRCh38
rs558619523 Chr.5: 176642849-176642849 on Build GRCh38
rs752340680 Chr.5: 176642840-176642840 on Build GRCh38
rs55652220 Chr.5: 176642622-176642622 on Build GRCh38
rs371300385 Chr.5: 176642755-176642755 on Build GRCh38
rs575907394 Chr.5: 176642982-176642982 on Build GRCh38
rs963795476 Chr.5: 176642957-176642957 on Build GRCh38
rs1004846841 Chr.5: 176642716-176642716 on Build GRCh38
rs1052673753 Chr.5: 176642763-176642763 on Build GRCh38
rs924180848 Chr.5: 176642860-176642860 on Build GRCh38
rs577075780 Chr.5: 176642851-176642851 on Build GRCh38
rs551921260 Chr.5: 176642886-176642886 on Build GRCh38
rs916557023 Chr.5: 176642893-176642893 on Build GRCh38
rs1014801102 Chr.5: 176642643-176642643 on Build GRCh38
rs916633320 Chr.5: 176642698-176642698 on Build GRCh38
rs990156052 Chr.5: 176642885-176642885 on Build GRCh38
rs1014518974 Chr.5: 176642859-176642859 on Build GRCh38
rs780286530 Chr.5: 176642852-176642858 on Build GRCh38
rs757183634 Chr.5: 176642743-176642743 on Build GRCh38
rs868741478 Chr.5: 176642803-176642803 on Build GRCh38
rs781436374 Chr.5: 176642790-176642790 on Build GRCh38
rs897058439 Chr.5: 176642972-176642972 on Build GRCh38
rs866185731 Chr.5: 176642866-176642866 on Build GRCh38
rs772555913 Chr.5: 176642852-176642853 on Build GRCh38
rs988057678 Chr.5: 176643013-176643013 on Build GRCh38
rs756296646 Chr.5: 176642851-176642852 on Build GRCh38
rs778721946 Chr.5: 176642759-176642759 on Build GRCh38
rs948057052 Chr.5: 176642911-176642911 on Build GRCh38
rs904651321 Chr.5: 176642855-176642856 on Build GRCh38
rs374469410 Chr.5: 176642769-176642769 on Build GRCh38
rs948137646 Chr.5: 176642700-176642700 on Build GRCh38
rs553730978 Chr.5: 176642731-176642731 on Build GRCh38
rs560170677 Chr.5: 176643004-176643004 on Build GRCh38
rs773148617 Chr.5: 176642825-176642825 on Build GRCh38
rs760232166 Chr.5: 176642845-176642845 on Build GRCh38
rs56115420 Chr.5: 176642865-176642866 on Build GRCh38
rs960328907 Chr.5: 176642638-176642638 on Build GRCh38
rs544028654 Chr.5: 176642853-176642853 on Build GRCh38
rs951174160 Chr.5: 176642687-176642687 on Build GRCh38
rs757236490 Chr.5: 176643037-176643037 on Build GRCh38
rs5873545 Chr.5: 176642862-176642863 on Build GRCh38
rs781545350 Chr.5: 176642677-176642677 on Build GRCh38
rs1026342887 Chr.5: 176642860-176642861 on Build GRCh38
rs772941013 Chr.5: 176642712-176642712 on Build GRCh38
rs965513140 Chr.5: 176642856-176642856 on Build GRCh38
rs1055727224 Chr.5: 176642858-176642859 on Build GRCh38
rs1046436840 Chr.5: 176642914-176642914 on Build GRCh38
rs978262908 Chr.5: 176642857-176642857 on Build GRCh38
rs1048805926 Chr.5: 176642993-176642993 on Build GRCh38
rs367999309 Chr.5: 176642770-176642770 on Build GRCh38
rs928074305 Chr.5: 176642917-176642917 on Build GRCh38
rs748181057 Chr.5: 176642809-176642809 on Build GRCh38
rs4618446 Chr.5: 176642865-176642865 on Build GRCh38
rs958305034 Chr.5: 176642878-176642878 on Build GRCh38
rs975036344 Chr.5: 176642979-176642979 on Build GRCh38
rs745905318 Chr.5: 176642849-176642850 on Build GRCh38
rs896035849 Chr.5: 176642859-176642860 on Build GRCh38
rs983856230 Chr.5: 176642699-176642699 on Build GRCh38
rs562726741 Chr.5: 176642963-176642963 on Build GRCh38
rs867272495 Chr.5: 176642863-176642863 on Build GRCh38
rs373254210 Chr.5: 176642720-176642720 on Build GRCh38
rs1016684872 Chr.5: 176642956-176642956 on Build GRCh38
rs922476611 Chr.5: 176642744-176642744 on Build GRCh38
rs757227010 Chr.5: 176642780-176642781 on Build GRCh38

View Full Product Details