Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31356819-31357334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-B
Entrez Gene ID
3106
Gene Name
major histocompatibility complex, class I, B
Gene Aliases
AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
UniGene
Hs.654404
Gene Chromosome Location
Chr.6: 31353866-31357245 on Build GRCh38
Transcripts
NM_005514.7 XM_011514557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690644 c.44delC p.A15fs*5 Chr.6: 31357115-31357115 on Build GRCh38
5450232 c.72C>T p.A24A Chr.6: 31357087-31357087 on Build GRCh38
5830398 c.74-3delC p.? Chr.6: 31356960-31356960 on Build GRCh38
4592026 c.206A>G p.E69G Chr.6: 31356825-31356825 on Build GRCh38
3761869 c.103T>G p.S35A Chr.6: 31356928-31356928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9266184 Chr.6: 31356887-31356887 on Build GRCh38
rs964673339 Chr.6: 31357027-31357027 on Build GRCh38
rs41559921 Chr.6: 31356926-31356926 on Build GRCh38
rs766453064 Chr.6: 31357076-31357077 on Build GRCh38
rs772687832 Chr.6: 31357031-31357031 on Build GRCh38
rs760037174 Chr.6: 31357109-31357110 on Build GRCh38
rs151341095 Chr.6: 31356927-31356927 on Build GRCh38
rs1050529 Chr.6: 31356838-31356838 on Build GRCh38
rs961379087 Chr.6: 31357010-31357010 on Build GRCh38
rs151341069 Chr.6: 31357310-31357310 on Build GRCh38
rs137854636 Chr.6: 31356950-31356950 on Build GRCh38
rs9281380 Chr.6: 31357077-31357078 on Build GRCh38
rs780237727 Chr.6: 31357128-31357128 on Build GRCh38
rs796742686 Chr.6: 31357251-31357253 on Build GRCh38
rs151341114 Chr.6: 31356851-31356851 on Build GRCh38
rs112333881 Chr.6: 31357252-31357252 on Build GRCh38
rs151341094 Chr.6: 31356929-31356929 on Build GRCh38
rs1065378 Chr.6: 31356856-31356856 on Build GRCh38
rs137854659 Chr.6: 31356880-31356880 on Build GRCh38
rs151341108 Chr.6: 31356878-31356878 on Build GRCh38
rs749360820 Chr.6: 31357108-31357108 on Build GRCh38
rs151341106 Chr.6: 31356892-31356892 on Build GRCh38
rs151341088 Chr.6: 31356953-31356953 on Build GRCh38
rs949147688 Chr.6: 31357186-31357186 on Build GRCh38
rs151341093 Chr.6: 31356935-31356935 on Build GRCh38
rs749523872 Chr.6: 31357131-31357131 on Build GRCh38
rs45509993 Chr.6: 31356898-31356898 on Build GRCh38
rs768345769 Chr.6: 31356969-31356969 on Build GRCh38
rs796362505 Chr.6: 31357114-31357115 on Build GRCh38
rs11546719 Chr.6: 31356897-31356897 on Build GRCh38
rs1131159 Chr.6: 31357115-31357115 on Build GRCh38
rs151341107 Chr.6: 31356881-31356881 on Build GRCh38
rs1050459 Chr.6: 31357144-31357144 on Build GRCh38
rs45539837 Chr.6: 31357080-31357080 on Build GRCh38
rs114811870 Chr.6: 31357161-31357161 on Build GRCh38
rs796895272 Chr.6: 31357052-31357053 on Build GRCh38
rs281864593 Chr.6: 31356873-31356873 on Build GRCh38
rs77882874 Chr.6: 31356952-31356952 on Build GRCh38
rs151341087 Chr.6: 31356954-31356954 on Build GRCh38
rs151341105 Chr.6: 31356895-31356895 on Build GRCh38
rs151341092 Chr.6: 31356937-31356937 on Build GRCh38
rs79051037 Chr.6: 31356918-31356918 on Build GRCh38
rs41552714 Chr.6: 31356979-31356979 on Build GRCh38
rs202190710 Chr.6: 31356999-31356999 on Build GRCh38
rs2596490 Chr.6: 31357219-31357219 on Build GRCh38
rs151341084 Chr.6: 31356963-31356963 on Build GRCh38
rs41564319 Chr.6: 31356886-31356886 on Build GRCh38
rs771386725 Chr.6: 31357039-31357039 on Build GRCh38
rs9266195 Chr.6: 31357043-31357043 on Build GRCh38
rs281864594 Chr.6: 31356865-31356865 on Build GRCh38
rs771028948 Chr.6: 31356966-31356967 on Build GRCh38
rs865881157 Chr.6: 31357058-31357058 on Build GRCh38
rs1050462 Chr.6: 31357134-31357134 on Build GRCh38
rs281864592 Chr.6: 31356874-31356874 on Build GRCh38
rs45624240 Chr.6: 31357085-31357085 on Build GRCh38
rs114038515 Chr.6: 31357164-31357164 on Build GRCh38
rs41555918 Chr.6: 31356940-31356940 on Build GRCh38
rs713031 Chr.6: 31356889-31356889 on Build GRCh38
rs1131156 Chr.6: 31357118-31357118 on Build GRCh38
rs75402393 Chr.6: 31356974-31356974 on Build GRCh38
rs41542121 Chr.6: 31356885-31356885 on Build GRCh38
rs77592474 Chr.6: 31356917-31356917 on Build GRCh38
rs567524803 Chr.6: 31357230-31357230 on Build GRCh38
rs778385708 Chr.6: 31357034-31357034 on Build GRCh38
rs1050486 Chr.6: 31356925-31356925 on Build GRCh38
rs151341104 Chr.6: 31356899-31356899 on Build GRCh38
rs2596489 Chr.6: 31357251-31357251 on Build GRCh38
rs9266210 Chr.6: 31357246-31357246 on Build GRCh38
rs137854653 Chr.6: 31356904-31356904 on Build GRCh38
rs151341099 Chr.6: 31356916-31356916 on Build GRCh38
rs151341109 Chr.6: 31356876-31356876 on Build GRCh38
rs281864591 Chr.6: 31356877-31356877 on Build GRCh38
rs151341098 Chr.6: 31356920-31356920 on Build GRCh38
rs41563412 Chr.6: 31356966-31356966 on Build GRCh38
rs151341115 Chr.6: 31356847-31356847 on Build GRCh38
rs538011088 Chr.6: 31357242-31357242 on Build GRCh38
rs151341103 Chr.6: 31356908-31356908 on Build GRCh38
rs137854656 Chr.6: 31356893-31356893 on Build GRCh38
rs1023788662 Chr.6: 31357249-31357249 on Build GRCh38
rs748621222 Chr.6: 31357026-31357026 on Build GRCh38
rs41552717 Chr.6: 31356845-31356845 on Build GRCh38
rs201956837 Chr.6: 31356961-31356961 on Build GRCh38
rs765139348 Chr.6: 31357142-31357142 on Build GRCh38
rs151341097 Chr.6: 31356921-31356921 on Build GRCh38
rs764977894 Chr.6: 31357113-31357113 on Build GRCh38
rs41561613 Chr.6: 31356850-31356850 on Build GRCh38
rs796133251 Chr.6: 31356887-31356889 on Build GRCh38
rs9266194 Chr.6: 31357013-31357013 on Build GRCh38
rs45627838 Chr.6: 31357079-31357079 on Build GRCh38
rs41564222 Chr.6: 31356900-31356900 on Build GRCh38
rs45624436 Chr.6: 31357082-31357082 on Build GRCh38
rs151341116 Chr.6: 31356846-31356846 on Build GRCh38
rs151341102 Chr.6: 31356909-31356909 on Build GRCh38
rs281864595 Chr.6: 31356861-31356861 on Build GRCh38
rs765875917 Chr.6: 31357115-31357115 on Build GRCh38
rs761604346 Chr.6: 31357054-31357055 on Build GRCh38
rs41544413 Chr.6: 31356848-31356848 on Build GRCh38
rs9266198 Chr.6: 31357070-31357070 on Build GRCh38
rs41552116 Chr.6: 31357021-31357021 on Build GRCh38
rs137854633 Chr.6: 31357074-31357075 on Build GRCh38
rs889873191 Chr.6: 31356991-31356991 on Build GRCh38
rs41552516 Chr.6: 31356840-31356840 on Build GRCh38
rs41558918 Chr.6: 31356997-31356997 on Build GRCh38
rs151341117 Chr.6: 31356843-31356843 on Build GRCh38
rs41562812 Chr.6: 31356947-31356947 on Build GRCh38
rs773476018 Chr.6: 31356984-31356984 on Build GRCh38
rs955649905 Chr.6: 31357302-31357302 on Build GRCh38
rs2596488 Chr.6: 31357253-31357253 on Build GRCh38
rs761769875 Chr.6: 31357005-31357005 on Build GRCh38
rs774515074 Chr.6: 31357132-31357132 on Build GRCh38
rs56173567 Chr.6: 31356854-31356854 on Build GRCh38
rs137854671 Chr.6: 31356839-31356839 on Build GRCh38
rs45496696 Chr.6: 31356958-31356958 on Build GRCh38
rs371250843 Chr.6: 31356960-31356960 on Build GRCh38
rs151341073 Chr.6: 31357224-31357224 on Build GRCh38
rs747330852 Chr.6: 31357036-31357036 on Build GRCh38
rs868482646 Chr.6: 31357002-31357002 on Build GRCh38
rs151341091 Chr.6: 31356941-31356941 on Build GRCh38
rs41563012 Chr.6: 31356844-31356844 on Build GRCh38
rs41546814 Chr.6: 31356867-31356867 on Build GRCh38
rs774891894 Chr.6: 31356931-31356931 on Build GRCh38
rs281864585 Chr.6: 31356974-31356983 on Build GRCh38
rs779047853 Chr.6: 31357156-31357156 on Build GRCh38
rs762695896 Chr.6: 31357147-31357147 on Build GRCh38
rs9266207 Chr.6: 31357176-31357176 on Build GRCh38
rs45585233 Chr.6: 31357112-31357112 on Build GRCh38
rs2596487 Chr.6: 31357279-31357279 on Build GRCh38
rs770875504 Chr.6: 31357137-31357137 on Build GRCh38
rs778762328 Chr.6: 31357076-31357076 on Build GRCh38
rs41546213 Chr.6: 31356860-31356860 on Build GRCh38
rs45551433 Chr.6: 31357078-31357078 on Build GRCh38
rs771132803 Chr.6: 31357099-31357099 on Build GRCh38
rs766207552 Chr.6: 31357068-31357068 on Build GRCh38
rs753043104 Chr.6: 31357178-31357179 on Build GRCh38
rs199519399 Chr.6: 31356964-31356964 on Build GRCh38
rs41552318 Chr.6: 31356942-31356942 on Build GRCh38
rs151341090 Chr.6: 31356949-31356949 on Build GRCh38
rs1050502 Chr.6: 31356890-31356890 on Build GRCh38
rs151341085 Chr.6: 31356960-31356960 on Build GRCh38
rs755208749 Chr.6: 31357155-31357155 on Build GRCh38
rs76032827 Chr.6: 31356915-31356915 on Build GRCh38
rs796610180 Chr.6: 31357011-31357013 on Build GRCh38
rs151341071 Chr.6: 31357295-31357295 on Build GRCh38
rs749215209 Chr.6: 31357051-31357052 on Build GRCh38
rs45560938 Chr.6: 31357084-31357084 on Build GRCh38
rs200193749 Chr.6: 31356983-31356983 on Build GRCh38
rs9266196 Chr.6: 31357052-31357052 on Build GRCh38
rs775725340 Chr.6: 31357107-31357107 on Build GRCh38
rs9266214 Chr.6: 31357272-31357272 on Build GRCh38
rs151341118 Chr.6: 31356841-31356841 on Build GRCh38
rs1131165 Chr.6: 31357110-31357110 on Build GRCh38
rs1055758550 Chr.6: 31356958-31356958 on Build GRCh38
rs769822288 Chr.6: 31357046-31357046 on Build GRCh38
rs144862331 Chr.6: 31357123-31357123 on Build GRCh38
rs755972630 Chr.6: 31357009-31357009 on Build GRCh38
rs45550032 Chr.6: 31356901-31356901 on Build GRCh38
rs9266211 Chr.6: 31357248-31357248 on Build GRCh38
rs281864588 Chr.6: 31356891-31356891 on Build GRCh38
rs41540116 Chr.6: 31357103-31357103 on Build GRCh38
rs72558107 Chr.6: 31356907-31356907 on Build GRCh38
rs1131170 Chr.6: 31356928-31356928 on Build GRCh38
rs9266197 Chr.6: 31357053-31357053 on Build GRCh38
rs151341111 Chr.6: 31356863-31356863 on Build GRCh38
rs756409564 Chr.6: 31357153-31357153 on Build GRCh38
rs151341119 Chr.6: 31356837-31356837 on Build GRCh38
rs796986400 Chr.6: 31356864-31356866 on Build GRCh38
rs796100503 Chr.6: 31357246-31357253 on Build GRCh38
rs41548616 Chr.6: 31356872-31356872 on Build GRCh38
rs41560213 Chr.6: 31356912-31356912 on Build GRCh38
rs746200003 Chr.6: 31357125-31357125 on Build GRCh38
rs41556816 Chr.6: 31356842-31356842 on Build GRCh38
rs137854651 Chr.6: 31356910-31356910 on Build GRCh38
rs774095948 Chr.6: 31357053-31357054 on Build GRCh38
rs137854665 Chr.6: 31356855-31356855 on Build GRCh38
rs281864597 Chr.6: 31356852-31356852 on Build GRCh38
rs777942857 Chr.6: 31356963-31356965 on Build GRCh38
rs796503852 Chr.6: 31356965-31356966 on Build GRCh38
rs151341110 Chr.6: 31356869-31356869 on Build GRCh38
rs41560522 Chr.6: 31356987-31356987 on Build GRCh38
rs72558106 Chr.6: 31356945-31356945 on Build GRCh38
rs41546617 Chr.6: 31356938-31356938 on Build GRCh38
rs41558422 Chr.6: 31356883-31356883 on Build GRCh38
rs1050517 Chr.6: 31356866-31356866 on Build GRCh38
rs281864587 Chr.6: 31356915-31356915 on Build GRCh38
rs754702572 Chr.6: 31357075-31357075 on Build GRCh38
rs2596492 Chr.6: 31356934-31356934 on Build GRCh38
rs769980881 Chr.6: 31357106-31357106 on Build GRCh38
rs9266209 Chr.6: 31357227-31357227 on Build GRCh38
rs778131844 Chr.6: 31357086-31357086 on Build GRCh38
rs1050518 Chr.6: 31356864-31356864 on Build GRCh38
rs41557213 Chr.6: 31357042-31357042 on Build GRCh38
rs994131450 Chr.6: 31357223-31357223 on Build GRCh38
rs281864596 Chr.6: 31356859-31356859 on Build GRCh38
rs281864590 Chr.6: 31356879-31356879 on Build GRCh38
rs3177891 Chr.6: 31356914-31356914 on Build GRCh38
rs1131163 Chr.6: 31357111-31357111 on Build GRCh38
rs77234140 Chr.6: 31356933-31356933 on Build GRCh38
rs151341070 Chr.6: 31357309-31357309 on Build GRCh38
rs750982109 Chr.6: 31357152-31357152 on Build GRCh38
rs76827460 Chr.6: 31356868-31356868 on Build GRCh38
rs281864586 Chr.6: 31356939-31356939 on Build GRCh38
rs1050458 Chr.6: 31357148-31357148 on Build GRCh38
rs147324178 Chr.6: 31356965-31356965 on Build GRCh38
rs137854661 Chr.6: 31356871-31356871 on Build GRCh38
rs41558815 Chr.6: 31356973-31356973 on Build GRCh38
rs9266193 Chr.6: 31357011-31357011 on Build GRCh38
rs773942130 Chr.6: 31357184-31357184 on Build GRCh38
rs137854638 Chr.6: 31356946-31356946 on Build GRCh38
rs190427310 Chr.6: 31357162-31357162 on Build GRCh38
rs3177890 Chr.6: 31356905-31356905 on Build GRCh38
rs769674193 Chr.6: 31357168-31357168 on Build GRCh38
rs2596486 Chr.6: 31357298-31357298 on Build GRCh38
rs281864589 Chr.6: 31356882-31356882 on Build GRCh38
rs386699132 Chr.6: 31357176-31357178 on Build GRCh38
rs9266206 Chr.6: 31357154-31357154 on Build GRCh38
rs768502267 Chr.6: 31357052-31357053 on Build GRCh38
rs769331294 Chr.6: 31357030-31357031 on Build GRCh38
rs760874750 Chr.6: 31357075-31357076 on Build GRCh38
rs151341101 Chr.6: 31356911-31356911 on Build GRCh38
rs151341076 Chr.6: 31357087-31357087 on Build GRCh38
rs776780353 Chr.6: 31357140-31357140 on Build GRCh38
rs151341074 Chr.6: 31357154-31357154 on Build GRCh38
rs770087936 Chr.6: 31357299-31357299 on Build GRCh38
rs41543515 Chr.6: 31357145-31357145 on Build GRCh38
rs45607634 Chr.6: 31357077-31357077 on Build GRCh38
rs767251021 Chr.6: 31357006-31357006 on Build GRCh38
rs41555216 Chr.6: 31356948-31356948 on Build GRCh38
rs1131161 Chr.6: 31357114-31357114 on Build GRCh38
rs756929171 Chr.6: 31356924-31356925 on Build GRCh38
rs151341112 Chr.6: 31356858-31356858 on Build GRCh38
rs562677576 Chr.6: 31357206-31357207 on Build GRCh38
rs9266183 Chr.6: 31356870-31356870 on Build GRCh38
rs751970069 Chr.6: 31357117-31357117 on Build GRCh38
rs151341086 Chr.6: 31356959-31356959 on Build GRCh38
rs151341096 Chr.6: 31356923-31356923 on Build GRCh38
rs200199153 Chr.6: 31357000-31357000 on Build GRCh38
rs183692514 Chr.6: 31356972-31356972 on Build GRCh38
rs765313444 Chr.6: 31357003-31357003 on Build GRCh38
rs756534089 Chr.6: 31357127-31357127 on Build GRCh38
rs45452896 Chr.6: 31357081-31357081 on Build GRCh38
rs760371278 Chr.6: 31357062-31357062 on Build GRCh38
rs137854644 Chr.6: 31356924-31356924 on Build GRCh38
rs369681614 Chr.6: 31357174-31357174 on Build GRCh38
rs751430514 Chr.6: 31357004-31357004 on Build GRCh38
rs34437781 Chr.6: 31357178-31357178 on Build GRCh38
rs151341113 Chr.6: 31356853-31356853 on Build GRCh38
rs763666151 Chr.6: 31357116-31357116 on Build GRCh38
rs45566634 Chr.6: 31357083-31357083 on Build GRCh38
rs1019762428 Chr.6: 31356986-31356986 on Build GRCh38
rs41552612 Chr.6: 31356922-31356922 on Build GRCh38

View Full Product Details