Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109691130-109691630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
UniGene
Hs.205144
Gene Chromosome Location
Chr.6: 109492855-109691253 on Build GRCh38
Transcripts
NM_001145128.2 NM_145025.4 XM_006715376.3 XM_011535550.2 XM_011535552.2 XM_011535553.2 XM_011535554.2 XM_011535555.2 XM_011535556.2 XM_011535559.2 XM_017010384.1 XM_017010385.1 XM_017010386.1 XM_017010388.1 NM_014845.5 XM_017011591.1 XM_017011592.1 XM_017011593.1

Gene Symbol
FIG4
Entrez Gene ID
9896
Gene Name
FIG4 phosphoinositide 5-phosphatase
Gene Aliases
ALS11, BTOP, CMT4J, KIAA0274, SAC3, YVS, dJ249I4.1
UniGene
Hs.529959
Gene Chromosome Location
Chr.6: 109691216-109825431 on Build GRCh38
Transcripts
NM_001145128.2 NM_145025.4 XM_006715376.3 XM_011535550.2 XM_011535552.2 XM_011535553.2 XM_011535554.2 XM_011535555.2 XM_011535556.2 XM_011535559.2 XM_017010384.1 XM_017010385.1 XM_017010386.1 XM_017010388.1 NM_014845.5 XM_017011591.1 XM_017011592.1 XM_017011593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6313433 c.66+1G>T p.? Chr.6: 109691502-109691502 on Build GRCh38
5682131 c.48T>G p.V16V Chr.6: 109691483-109691483 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs386704757 Chr.6: 109691315-109691316 on Build GRCh38
rs765243012 Chr.6: 109691429-109691429 on Build GRCh38
rs997420128 Chr.6: 109691547-109691547 on Build GRCh38
rs572003905 Chr.6: 109691338-109691338 on Build GRCh38
rs974206216 Chr.6: 109691411-109691411 on Build GRCh38
rs973921106 Chr.6: 109691235-109691235 on Build GRCh38
rs984029577 Chr.6: 109691190-109691190 on Build GRCh38
rs917084760 Chr.6: 109691232-109691232 on Build GRCh38
rs939349755 Chr.6: 109691286-109691286 on Build GRCh38
rs572759000 Chr.6: 109691193-109691193 on Build GRCh38
rs992768596 Chr.6: 109691407-109691407 on Build GRCh38
rs961354652 Chr.6: 109691197-109691197 on Build GRCh38
rs544833165 Chr.6: 109691548-109691548 on Build GRCh38
rs764876093 Chr.6: 109691532-109691532 on Build GRCh38
rs559502057 Chr.6: 109691164-109691164 on Build GRCh38
rs761698877 Chr.6: 109691528-109691528 on Build GRCh38
rs574002828 Chr.6: 109691392-109691392 on Build GRCh38
rs926602970 Chr.6: 109691293-109691293 on Build GRCh38
rs557370159 Chr.6: 109691381-109691381 on Build GRCh38
rs55873083 Chr.6: 109691254-109691254 on Build GRCh38
rs1027956668 Chr.6: 109691186-109691186 on Build GRCh38
rs537038147 Chr.6: 109691380-109691380 on Build GRCh38
rs575191138 Chr.6: 109691565-109691565 on Build GRCh38
rs151120474 Chr.6: 109691320-109691320 on Build GRCh38
rs761202811 Chr.6: 109691225-109691225 on Build GRCh38
rs768404941 Chr.6: 109691509-109691509 on Build GRCh38
rs896832398 Chr.6: 109691589-109691589 on Build GRCh38
rs75509752 Chr.6: 109691315-109691315 on Build GRCh38
rs552856153 Chr.6: 109691477-109691477 on Build GRCh38
rs201985085 Chr.6: 109691550-109691550 on Build GRCh38
rs587777713 Chr.6: 109691485-109691485 on Build GRCh38
rs548471516 Chr.6: 109691304-109691304 on Build GRCh38
rs965122100 Chr.6: 109691154-109691154 on Build GRCh38
rs776460299 Chr.6: 109691397-109691397 on Build GRCh38
rs796189004 Chr.6: 109691319-109691319 on Build GRCh38
rs755637236 Chr.6: 109691513-109691514 on Build GRCh38
rs773216511 Chr.6: 109691539-109691539 on Build GRCh38
rs371338038 Chr.6: 109691575-109691575 on Build GRCh38
rs868101103 Chr.6: 109691451-109691451 on Build GRCh38
rs749639397 Chr.6: 109691471-109691471 on Build GRCh38
rs746307320 Chr.6: 109691489-109691489 on Build GRCh38
rs906553229 Chr.6: 109691270-109691270 on Build GRCh38
rs917052197 Chr.6: 109691450-109691450 on Build GRCh38
rs954990630 Chr.6: 109691170-109691170 on Build GRCh38
rs908434484 Chr.6: 109691192-109691192 on Build GRCh38
rs75045314 Chr.6: 109691316-109691316 on Build GRCh38
rs775137642 Chr.6: 109691498-109691498 on Build GRCh38
rs1053820039 Chr.6: 109691300-109691300 on Build GRCh38
rs144593588 Chr.6: 109691496-109691496 on Build GRCh38
rs543861827 Chr.6: 109691578-109691578 on Build GRCh38
rs1022477936 Chr.6: 109691306-109691306 on Build GRCh38
rs952368847 Chr.6: 109691188-109691188 on Build GRCh38
rs993069858 Chr.6: 109691208-109691208 on Build GRCh38
rs560945087 Chr.6: 109691587-109691587 on Build GRCh38
rs750085102 Chr.6: 109691509-109691510 on Build GRCh38
rs1046023701 Chr.6: 109691269-109691269 on Build GRCh38
rs200063827 Chr.6: 109691511-109691511 on Build GRCh38
rs1015300933 Chr.6: 109691362-109691362 on Build GRCh38
rs915749340 Chr.6: 109691449-109691449 on Build GRCh38
rs560811735 Chr.6: 109691302-109691302 on Build GRCh38
rs761707195 Chr.6: 109691417-109691417 on Build GRCh38
rs979929537 Chr.6: 109691464-109691464 on Build GRCh38
rs750289982 Chr.6: 109691431-109691431 on Build GRCh38
rs867674950 Chr.6: 109691591-109691591 on Build GRCh38
rs976044855 Chr.6: 109691163-109691163 on Build GRCh38
rs1045140165 Chr.6: 109691262-109691262 on Build GRCh38
rs964337692 Chr.6: 109691158-109691158 on Build GRCh38
rs141040807 Chr.6: 109691462-109691462 on Build GRCh38
rs962506527 Chr.6: 109691383-109691384 on Build GRCh38
rs544958043 Chr.6: 109691312-109691312 on Build GRCh38
rs891371495 Chr.6: 109691554-109691554 on Build GRCh38
rs757848529 Chr.6: 109691324-109691324 on Build GRCh38
rs527523781 Chr.6: 109691468-109691468 on Build GRCh38
rs1015287047 Chr.6: 109691577-109691577 on Build GRCh38
rs544916656 Chr.6: 109691217-109691217 on Build GRCh38
rs764845399 Chr.6: 109691352-109691352 on Build GRCh38
rs562558336 Chr.6: 109691206-109691206 on Build GRCh38
rs1041331138 Chr.6: 109691497-109691497 on Build GRCh38
rs1015381926 Chr.6: 109691377-109691377 on Build GRCh38
rs755056253 Chr.6: 109691459-109691459 on Build GRCh38
rs996996501 Chr.6: 109691512-109691512 on Build GRCh38
rs952462093 Chr.6: 109691328-109691328 on Build GRCh38
rs550585093 Chr.6: 109691298-109691298 on Build GRCh38
rs756325278 Chr.6: 109691467-109691467 on Build GRCh38
rs746703726 Chr.6: 109691499-109691499 on Build GRCh38
rs564513214 Chr.6: 109691238-109691238 on Build GRCh38

View Full Product Details