Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1091254-1091749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7269721 c.3G>T p.M1I Chr.7: 1091731-1091731 on Build GRCh38
4004259 c.14C>T p.S5F Chr.7: 1091742-1091742 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746729509 Chr.7: 1091697-1091697 on Build GRCh38
rs530277541 Chr.7: 1091571-1091571 on Build GRCh38
rs549941523 Chr.7: 1091581-1091581 on Build GRCh38
rs777450622 Chr.7: 1091692-1091692 on Build GRCh38
rs565108668 Chr.7: 1091475-1091475 on Build GRCh38
rs1014330407 Chr.7: 1091471-1091471 on Build GRCh38
rs200430968 Chr.7: 1091701-1091701 on Build GRCh38
rs545398867 Chr.7: 1091459-1091459 on Build GRCh38
rs935785912 Chr.7: 1091452-1091452 on Build GRCh38
rs1028034723 Chr.7: 1091696-1091696 on Build GRCh38
rs960305567 Chr.7: 1091554-1091554 on Build GRCh38
rs951932650 Chr.7: 1091487-1091487 on Build GRCh38
rs748799525 Chr.7: 1091548-1091548 on Build GRCh38
rs764601424 Chr.7: 1091486-1091486 on Build GRCh38
rs985757418 Chr.7: 1091711-1091711 on Build GRCh38
rs1027722777 Chr.7: 1091312-1091312 on Build GRCh38
rs1015414239 Chr.7: 1091510-1091510 on Build GRCh38
rs976064174 Chr.7: 1091543-1091543 on Build GRCh38
rs905774134 Chr.7: 1091474-1091474 on Build GRCh38
rs568647016 Chr.7: 1091635-1091635 on Build GRCh38
rs960212727 Chr.7: 1091431-1091431 on Build GRCh38
rs542837477 Chr.7: 1091373-1091373 on Build GRCh38
rs573921931 Chr.7: 1091372-1091372 on Build GRCh38
rs551358452 Chr.7: 1091647-1091647 on Build GRCh38
rs770412758 Chr.7: 1091700-1091700 on Build GRCh38
rs115385429 Chr.7: 1091484-1091484 on Build GRCh38
rs1027461481 Chr.7: 1091485-1091485 on Build GRCh38
rs1049674752 Chr.7: 1091468-1091468 on Build GRCh38
rs188551362 Chr.7: 1091644-1091644 on Build GRCh38
rs932162648 Chr.7: 1091414-1091414 on Build GRCh38
rs754967390 Chr.7: 1091681-1091681 on Build GRCh38
rs747994003 Chr.7: 1091689-1091689 on Build GRCh38
rs201353172 Chr.7: 1091683-1091683 on Build GRCh38
rs1001830540 Chr.7: 1091384-1091384 on Build GRCh38
rs1002166747 Chr.7: 1091506-1091506 on Build GRCh38
rs767636021 Chr.7: 1091721-1091721 on Build GRCh38
rs570765254 Chr.7: 1091329-1091329 on Build GRCh38
rs368800854 Chr.7: 1091717-1091717 on Build GRCh38
rs367781346 Chr.7: 1091453-1091453 on Build GRCh38
rs200264603 Chr.7: 1091718-1091718 on Build GRCh38
rs3802141 Chr.7: 1091720-1091720 on Build GRCh38
rs1010022163 Chr.7: 1091497-1091497 on Build GRCh38
rs907871800 Chr.7: 1091342-1091342 on Build GRCh38
rs779477770 Chr.7: 1091533-1091533 on Build GRCh38
rs756592402 Chr.7: 1091521-1091521 on Build GRCh38
rs966109862 Chr.7: 1091520-1091520 on Build GRCh38
rs138061097 Chr.7: 1091433-1091433 on Build GRCh38
rs771811389 Chr.7: 1091691-1091691 on Build GRCh38
rs750559019 Chr.7: 1091726-1091726 on Build GRCh38
rs922209111 Chr.7: 1091577-1091577 on Build GRCh38
rs891440043 Chr.7: 1091331-1091331 on Build GRCh38
rs962100232 Chr.7: 1091680-1091680 on Build GRCh38
rs759997674 Chr.7: 1091715-1091715 on Build GRCh38
rs73253760 Chr.7: 1091397-1091397 on Build GRCh38
rs1045646466 Chr.7: 1091472-1091472 on Build GRCh38
rs74871253 Chr.7: 1091505-1091505 on Build GRCh38
rs979656946 Chr.7: 1091613-1091613 on Build GRCh38
rs983915810 Chr.7: 1091341-1091341 on Build GRCh38
rs944670028 Chr.7: 1091352-1091352 on Build GRCh38
rs768259080 Chr.7: 1091602-1091602 on Build GRCh38
rs571114606 Chr.7: 1091659-1091659 on Build GRCh38
rs527992446 Chr.7: 1091478-1091478 on Build GRCh38
rs962130996 Chr.7: 1091445-1091445 on Build GRCh38
rs925413942 Chr.7: 1091446-1091446 on Build GRCh38
rs751991433 Chr.7: 1091334-1091334 on Build GRCh38

View Full Product Details