Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1092410-1092907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687794 c.751C>T p.R251W Chr.7: 1092479-1092479 on Build GRCh38
3632013 c.996G>C p.R332S Chr.7: 1092724-1092724 on Build GRCh38
7225906 c.1092C>T p.T364T Chr.7: 1092820-1092820 on Build GRCh38
6012602 c.769G>A p.A257T Chr.7: 1092497-1092497 on Build GRCh38
3256479 c.1010T>A p.L337Q Chr.7: 1092738-1092738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150960969 Chr.7: 1092624-1092624 on Build GRCh38
rs747159270 Chr.7: 1092787-1092787 on Build GRCh38
rs779367726 Chr.7: 1092653-1092653 on Build GRCh38
rs754326307 Chr.7: 1092646-1092646 on Build GRCh38
rs777852953 Chr.7: 1092662-1092662 on Build GRCh38
rs778212324 Chr.7: 1092785-1092785 on Build GRCh38
rs377670702 Chr.7: 1092481-1092481 on Build GRCh38
rs770631652 Chr.7: 1092541-1092541 on Build GRCh38
rs753418414 Chr.7: 1092767-1092767 on Build GRCh38
rs757659970 Chr.7: 1092856-1092856 on Build GRCh38
rs781390250 Chr.7: 1092860-1092860 on Build GRCh38
rs764618566 Chr.7: 1092698-1092698 on Build GRCh38
rs985035078 Chr.7: 1092825-1092825 on Build GRCh38
rs764896213 Chr.7: 1092846-1092846 on Build GRCh38
rs762883544 Chr.7: 1092611-1092611 on Build GRCh38
rs746767126 Chr.7: 1092881-1092881 on Build GRCh38
rs569932251 Chr.7: 1092585-1092585 on Build GRCh38
rs753789231 Chr.7: 1092444-1092444 on Build GRCh38
rs200314011 Chr.7: 1092556-1092556 on Build GRCh38
rs749872027 Chr.7: 1092588-1092588 on Build GRCh38
rs774415754 Chr.7: 1092460-1092460 on Build GRCh38
rs773531678 Chr.7: 1092600-1092600 on Build GRCh38
rs147941530 Chr.7: 1092832-1092832 on Build GRCh38
rs371278172 Chr.7: 1092464-1092464 on Build GRCh38
rs774706015 Chr.7: 1092528-1092528 on Build GRCh38
rs370415093 Chr.7: 1092634-1092634 on Build GRCh38
rs770844463 Chr.7: 1092768-1092768 on Build GRCh38
rs747835584 Chr.7: 1092480-1092480 on Build GRCh38
rs751714742 Chr.7: 1092436-1092436 on Build GRCh38
rs373485148 Chr.7: 1092664-1092664 on Build GRCh38
rs758976064 Chr.7: 1092850-1092850 on Build GRCh38
rs563630174 Chr.7: 1092488-1092488 on Build GRCh38
rs760376118 Chr.7: 1092766-1092766 on Build GRCh38
rs1054318571 Chr.7: 1092781-1092781 on Build GRCh38
rs757493670 Chr.7: 1092790-1092790 on Build GRCh38
rs142494393 Chr.7: 1092569-1092569 on Build GRCh38
rs139916786 Chr.7: 1092485-1092485 on Build GRCh38
rs770850498 Chr.7: 1092665-1092665 on Build GRCh38
rs10246354 Chr.7: 1092820-1092820 on Build GRCh38
rs781443203 Chr.7: 1092792-1092792 on Build GRCh38
rs3808352 Chr.7: 1092517-1092517 on Build GRCh38
rs986005400 Chr.7: 1092668-1092668 on Build GRCh38
rs373815808 Chr.7: 1092638-1092638 on Build GRCh38
rs749881809 Chr.7: 1092570-1092570 on Build GRCh38
rs936507527 Chr.7: 1092718-1092718 on Build GRCh38
rs762136910 Chr.7: 1092530-1092530 on Build GRCh38
rs748436640 Chr.7: 1092658-1092658 on Build GRCh38
rs776379860 Chr.7: 1092634-1092634 on Build GRCh38
rs913969847 Chr.7: 1092783-1092783 on Build GRCh38
rs201700404 Chr.7: 1092619-1092619 on Build GRCh38
rs376978032 Chr.7: 1092536-1092536 on Build GRCh38
rs755569333 Chr.7: 1092650-1092650 on Build GRCh38
rs766104171 Chr.7: 1092830-1092830 on Build GRCh38
rs780113437 Chr.7: 1092443-1092443 on Build GRCh38
rs370671788 Chr.7: 1092591-1092591 on Build GRCh38
rs138657567 Chr.7: 1092847-1092847 on Build GRCh38
rs780705424 Chr.7: 1092568-1092568 on Build GRCh38
rs375845162 Chr.7: 1092623-1092623 on Build GRCh38
rs958800961 Chr.7: 1092515-1092515 on Build GRCh38
rs746149188 Chr.7: 1092863-1092863 on Build GRCh38
rs766307448 Chr.7: 1092528-1092530 on Build GRCh38
rs140755092 Chr.7: 1092667-1092667 on Build GRCh38
rs543666614 Chr.7: 1092486-1092486 on Build GRCh38
rs760193133 Chr.7: 1092829-1092829 on Build GRCh38
rs773193769 Chr.7: 1092535-1092535 on Build GRCh38
rs373871922 Chr.7: 1092557-1092557 on Build GRCh38
rs367628125 Chr.7: 1092642-1092642 on Build GRCh38
rs993626351 Chr.7: 1092871-1092871 on Build GRCh38
rs2070118 Chr.7: 1092869-1092869 on Build GRCh38
rs369412581 Chr.7: 1092872-1092872 on Build GRCh38
rs745918553 Chr.7: 1092802-1092802 on Build GRCh38
rs771701149 Chr.7: 1092823-1092823 on Build GRCh38
rs943638671 Chr.7: 1092838-1092838 on Build GRCh38
rs148410177 Chr.7: 1092549-1092549 on Build GRCh38
rs775750584 Chr.7: 1092470-1092470 on Build GRCh38
rs758676506 Chr.7: 1092779-1092779 on Build GRCh38
rs543407810 Chr.7: 1092851-1092851 on Build GRCh38
rs758236330 Chr.7: 1092499-1092499 on Build GRCh38
rs11977449 Chr.7: 1092439-1092439 on Build GRCh38
rs758076694 Chr.7: 1092452-1092452 on Build GRCh38
rs774144814 Chr.7: 1092814-1092814 on Build GRCh38
rs190131612 Chr.7: 1092692-1092692 on Build GRCh38
rs776408692 Chr.7: 1092886-1092886 on Build GRCh38
rs774757548 Chr.7: 1092479-1092479 on Build GRCh38
rs74331861 Chr.7: 1092514-1092514 on Build GRCh38
rs756699715 Chr.7: 1092505-1092505 on Build GRCh38
rs768560550 Chr.7: 1092710-1092710 on Build GRCh38
rs771108834 Chr.7: 1092602-1092602 on Build GRCh38
rs763836425 Chr.7: 1092616-1092616 on Build GRCh38
rs772524763 Chr.7: 1092661-1092661 on Build GRCh38
rs35069667 Chr.7: 1092471-1092471 on Build GRCh38
rs201386398 Chr.7: 1092643-1092643 on Build GRCh38
rs754582018 Chr.7: 1092772-1092772 on Build GRCh38
rs761512963 Chr.7: 1092736-1092736 on Build GRCh38
rs759110202 Chr.7: 1092833-1092833 on Build GRCh38
rs1033361688 Chr.7: 1092730-1092730 on Build GRCh38
rs910392089 Chr.7: 1092703-1092703 on Build GRCh38
rs559603787 Chr.7: 1092532-1092532 on Build GRCh38
rs770914933 Chr.7: 1092885-1092885 on Build GRCh38
rs749990132 Chr.7: 1092765-1092765 on Build GRCh38
rs538484749 Chr.7: 1092589-1092589 on Build GRCh38
rs145985656 Chr.7: 1092459-1092459 on Build GRCh38
rs775392105 Chr.7: 1092605-1092605 on Build GRCh38
rs374724867 Chr.7: 1092489-1092489 on Build GRCh38
rs906521758 Chr.7: 1092803-1092803 on Build GRCh38
rs777183834 Chr.7: 1092463-1092463 on Build GRCh38
rs187037378 Chr.7: 1092576-1092576 on Build GRCh38
rs780572159 Chr.7: 1092512-1092512 on Build GRCh38
rs765806965 Chr.7: 1092776-1092776 on Build GRCh38
rs372252532 Chr.7: 1092744-1092744 on Build GRCh38
rs759274564 Chr.7: 1092888-1092888 on Build GRCh38
rs773068978 Chr.7: 1092874-1092874 on Build GRCh38
rs765060809 Chr.7: 1092547-1092547 on Build GRCh38
rs765275169 Chr.7: 1092823-1092824 on Build GRCh38
rs1053838604 Chr.7: 1092504-1092504 on Build GRCh38
rs747766901 Chr.7: 1092451-1092451 on Build GRCh38
rs952842968 Chr.7: 1092551-1092551 on Build GRCh38
rs375524389 Chr.7: 1092828-1092828 on Build GRCh38
rs777372818 Chr.7: 1092500-1092500 on Build GRCh38
rs181756759 Chr.7: 1092777-1092777 on Build GRCh38
rs1000090736 Chr.7: 1092673-1092673 on Build GRCh38
rs767547288 Chr.7: 1092482-1092482 on Build GRCh38
rs772203632 Chr.7: 1092599-1092599 on Build GRCh38
rs770131675 Chr.7: 1092465-1092465 on Build GRCh38
rs776724162 Chr.7: 1092681-1092681 on Build GRCh38
rs762429776 Chr.7: 1092837-1092837 on Build GRCh38
rs376780893 Chr.7: 1092584-1092584 on Build GRCh38
rs564861434 Chr.7: 1092516-1092516 on Build GRCh38
rs746729641 Chr.7: 1092503-1092503 on Build GRCh38
rs923438554 Chr.7: 1092868-1092868 on Build GRCh38
rs760587019 Chr.7: 1092522-1092524 on Build GRCh38
rs774314914 Chr.7: 1092733-1092733 on Build GRCh38
rs763891231 Chr.7: 1092552-1092552 on Build GRCh38
rs374017708 Chr.7: 1092601-1092601 on Build GRCh38
rs748338923 Chr.7: 1092597-1092597 on Build GRCh38
rs369986460 Chr.7: 1092550-1092550 on Build GRCh38
rs367713059 Chr.7: 1092445-1092445 on Build GRCh38
rs146041906 Chr.7: 1092519-1092519 on Build GRCh38
rs767048392 Chr.7: 1092628-1092628 on Build GRCh38

View Full Product Details