Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1093190-1093693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

Gene Symbol
GPER1
Entrez Gene ID
2852
Gene Name
G protein-coupled estrogen receptor 1
Gene Aliases
CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
UniGene
Hs.20961
Gene Chromosome Location
Chr.7: 1086807-1093815 on Build GRCh38
Transcripts
NM_001134395.1 NM_001134396.1 NM_001318252.1 NM_032350.5 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 XM_017012722.1 NM_001039966.1 NM_001098201.1 NM_001505.2

SNP Detail

SNP ID Chromosome Location
rs771262064 Chr.7: 1093459-1093459 on Build GRCh38
rs1046402505 Chr.7: 1093326-1093326 on Build GRCh38
rs148746582 Chr.7: 1093429-1093429 on Build GRCh38
rs868831301 Chr.7: 1093606-1093606 on Build GRCh38
rs1012355489 Chr.7: 1093608-1093608 on Build GRCh38
rs1017047119 Chr.7: 1093625-1093625 on Build GRCh38
rs1133043 Chr.7: 1093669-1093669 on Build GRCh38
rs555778461 Chr.7: 1093420-1093420 on Build GRCh38
rs563267613 Chr.7: 1093331-1093331 on Build GRCh38
rs376635062 Chr.7: 1093542-1093542 on Build GRCh38
rs545459626 Chr.7: 1093541-1093541 on Build GRCh38
rs1010985650 Chr.7: 1093414-1093414 on Build GRCh38
rs1012364301 Chr.7: 1093447-1093447 on Build GRCh38
rs3808353 Chr.7: 1093336-1093336 on Build GRCh38
rs539441913 Chr.7: 1093410-1093410 on Build GRCh38
rs750442253 Chr.7: 1093377-1093377 on Build GRCh38
rs763787350 Chr.7: 1093385-1093386 on Build GRCh38
rs951433494 Chr.7: 1093632-1093632 on Build GRCh38
rs894070716 Chr.7: 1093590-1093590 on Build GRCh38
rs146081211 Chr.7: 1093380-1093380 on Build GRCh38
rs573558576 Chr.7: 1093425-1093425 on Build GRCh38
rs528297839 Chr.7: 1093356-1093356 on Build GRCh38
rs756034146 Chr.7: 1093534-1093534 on Build GRCh38
rs10235195 Chr.7: 1093241-1093241 on Build GRCh38
rs994239420 Chr.7: 1093668-1093668 on Build GRCh38
rs185131562 Chr.7: 1093381-1093381 on Build GRCh38
rs781747318 Chr.7: 1093639-1093639 on Build GRCh38
rs747511882 Chr.7: 1093613-1093613 on Build GRCh38
rs746842139 Chr.7: 1093242-1093242 on Build GRCh38
rs1017428620 Chr.7: 1093453-1093453 on Build GRCh38
rs987011951 Chr.7: 1093522-1093522 on Build GRCh38
rs547965950 Chr.7: 1093372-1093372 on Build GRCh38
rs374524593 Chr.7: 1093334-1093334 on Build GRCh38
rs142248487 Chr.7: 1093448-1093448 on Build GRCh38
rs1032888775 Chr.7: 1093389-1093389 on Build GRCh38
rs765525830 Chr.7: 1093484-1093484 on Build GRCh38
rs758430418 Chr.7: 1093324-1093324 on Build GRCh38
rs750305816 Chr.7: 1093506-1093506 on Build GRCh38
rs763109751 Chr.7: 1093496-1093496 on Build GRCh38
rs190336298 Chr.7: 1093551-1093551 on Build GRCh38
rs528332561 Chr.7: 1093582-1093582 on Build GRCh38
rs116150092 Chr.7: 1093294-1093294 on Build GRCh38
rs139275763 Chr.7: 1093501-1093501 on Build GRCh38
rs577487581 Chr.7: 1093230-1093230 on Build GRCh38
rs563147729 Chr.7: 1093519-1093519 on Build GRCh38
rs918813989 Chr.7: 1093363-1093363 on Build GRCh38
rs570783178 Chr.7: 1093405-1093405 on Build GRCh38
rs150824398 Chr.7: 1093467-1093467 on Build GRCh38
rs900278444 Chr.7: 1093375-1093375 on Build GRCh38
rs553678090 Chr.7: 1093411-1093411 on Build GRCh38
rs759682528 Chr.7: 1093218-1093218 on Build GRCh38
rs1003315054 Chr.7: 1093304-1093304 on Build GRCh38
rs982170628 Chr.7: 1093618-1093618 on Build GRCh38
rs770338788 Chr.7: 1093663-1093663 on Build GRCh38
rs3808354 Chr.7: 1093535-1093535 on Build GRCh38
rs759754980 Chr.7: 1093472-1093472 on Build GRCh38
rs752310975 Chr.7: 1093660-1093660 on Build GRCh38
rs552483485 Chr.7: 1093335-1093335 on Build GRCh38
rs753550268 Chr.7: 1093538-1093538 on Build GRCh38
rs550700139 Chr.7: 1093387-1093387 on Build GRCh38
rs559257087 Chr.7: 1093578-1093578 on Build GRCh38
rs777481200 Chr.7: 1093232-1093232 on Build GRCh38
rs758589940 Chr.7: 1093245-1093245 on Build GRCh38
rs561498549 Chr.7: 1093626-1093626 on Build GRCh38
rs1056301239 Chr.7: 1093604-1093604 on Build GRCh38
rs777272204 Chr.7: 1093583-1093583 on Build GRCh38
rs978657584 Chr.7: 1093263-1093263 on Build GRCh38
rs180911837 Chr.7: 1093325-1093325 on Build GRCh38
rs1032611708 Chr.7: 1093561-1093561 on Build GRCh38
rs1047655080 Chr.7: 1093444-1093444 on Build GRCh38
rs902167159 Chr.7: 1093573-1093573 on Build GRCh38
rs767245027 Chr.7: 1093389-1093391 on Build GRCh38
rs967015911 Chr.7: 1093251-1093251 on Build GRCh38
rs748928065 Chr.7: 1093409-1093409 on Build GRCh38
rs866526563 Chr.7: 1093546-1093546 on Build GRCh38
rs766798392 Chr.7: 1093428-1093428 on Build GRCh38
rs775920733 Chr.7: 1093490-1093490 on Build GRCh38
rs773496959 Chr.7: 1093373-1093373 on Build GRCh38
rs925513526 Chr.7: 1093275-1093275 on Build GRCh38
rs191002661 Chr.7: 1093240-1093240 on Build GRCh38
rs963024744 Chr.7: 1093347-1093347 on Build GRCh38
rs747630633 Chr.7: 1093415-1093415 on Build GRCh38
rs574568453 Chr.7: 1093302-1093302 on Build GRCh38
rs939276455 Chr.7: 1093577-1093577 on Build GRCh38
rs1042332309 Chr.7: 1093417-1093417 on Build GRCh38
rs973045522 Chr.7: 1093355-1093355 on Build GRCh38
rs114986355 Chr.7: 1093614-1093614 on Build GRCh38
rs754846225 Chr.7: 1093539-1093539 on Build GRCh38
rs573519884 Chr.7: 1093280-1093280 on Build GRCh38
rs542442054 Chr.7: 1093290-1093290 on Build GRCh38
rs757092660 Chr.7: 1093279-1093279 on Build GRCh38
rs748219734 Chr.7: 1093351-1093351 on Build GRCh38
rs776856922 Chr.7: 1093217-1093217 on Build GRCh38
rs1030036784 Chr.7: 1093378-1093378 on Build GRCh38
rs776892613 Chr.7: 1093466-1093466 on Build GRCh38

View Full Product Details