Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1470755-1471255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS1
Entrez Gene ID
26173
Gene Name
integrator complex subunit 1
Gene Aliases
INT1, NET28
UniGene
Hs.532188
Gene Chromosome Location
Chr.7: 1470277-1504382 on Build GRCh38
Transcripts
NM_001080453.2 XM_011515260.1 XM_017011959.1 XM_017011960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3879455 c.6915C>T p.N2305N Chr.7: 1471177-1471177 on Build GRCh38
6300023 c.7029G>A p.T2343T Chr.7: 1470886-1470886 on Build GRCh38
4851891 c.6968C>T p.A2323V Chr.7: 1470947-1470947 on Build GRCh38
6362351 c.6911G>A p.R2304H Chr.7: 1471181-1471181 on Build GRCh38
3262084 c.7053C>T p.Y2351Y Chr.7: 1470862-1470862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530027193 Chr.7: 1471061-1471061 on Build GRCh38
rs766337276 Chr.7: 1471137-1471137 on Build GRCh38
rs759782397 Chr.7: 1470928-1470928 on Build GRCh38
rs923450203 Chr.7: 1470873-1470873 on Build GRCh38
rs368143838 Chr.7: 1470892-1470892 on Build GRCh38
rs757898200 Chr.7: 1470934-1470934 on Build GRCh38
rs778125850 Chr.7: 1470890-1470890 on Build GRCh38
rs534944777 Chr.7: 1470790-1470790 on Build GRCh38
rs549771523 Chr.7: 1471063-1471063 on Build GRCh38
rs756539915 Chr.7: 1470798-1470798 on Build GRCh38
rs559625340 Chr.7: 1471016-1471016 on Build GRCh38
rs774504860 Chr.7: 1471121-1471121 on Build GRCh38
rs920564883 Chr.7: 1470858-1470858 on Build GRCh38
rs111764173 Chr.7: 1471024-1471024 on Build GRCh38
rs537368704 Chr.7: 1471173-1471173 on Build GRCh38
rs777144764 Chr.7: 1470923-1470923 on Build GRCh38
rs569692953 Chr.7: 1471070-1471070 on Build GRCh38
rs758080475 Chr.7: 1471171-1471171 on Build GRCh38
rs767202557 Chr.7: 1471234-1471234 on Build GRCh38
rs765347552 Chr.7: 1470840-1470840 on Build GRCh38
rs369867585 Chr.7: 1471210-1471210 on Build GRCh38
rs1132987 Chr.7: 1470944-1470944 on Build GRCh38
rs977295455 Chr.7: 1471107-1471107 on Build GRCh38
rs776987164 Chr.7: 1470833-1470833 on Build GRCh38
rs535171698 Chr.7: 1471091-1471091 on Build GRCh38
rs758335937 Chr.7: 1471142-1471142 on Build GRCh38
rs769413708 Chr.7: 1471215-1471215 on Build GRCh38
rs1018923942 Chr.7: 1471199-1471199 on Build GRCh38
rs1027660373 Chr.7: 1471040-1471040 on Build GRCh38
rs770330700 Chr.7: 1471103-1471103 on Build GRCh38
rs73265901 Chr.7: 1470774-1470774 on Build GRCh38
rs566245687 Chr.7: 1470785-1470785 on Build GRCh38
rs73267806 Chr.7: 1471195-1471195 on Build GRCh38
rs1002331952 Chr.7: 1470783-1470783 on Build GRCh38
rs1030615628 Chr.7: 1471025-1471025 on Build GRCh38
rs149670463 Chr.7: 1471198-1471198 on Build GRCh38
rs761770199 Chr.7: 1470849-1470849 on Build GRCh38
rs532175813 Chr.7: 1471078-1471078 on Build GRCh38
rs73267803 Chr.7: 1471021-1471021 on Build GRCh38
rs765080450 Chr.7: 1471155-1471155 on Build GRCh38
rs1041509864 Chr.7: 1470954-1470954 on Build GRCh38
rs759839020 Chr.7: 1470838-1470838 on Build GRCh38
rs377062473 Chr.7: 1470887-1470887 on Build GRCh38
rs1004044257 Chr.7: 1470828-1470828 on Build GRCh38
rs763340360 Chr.7: 1471231-1471231 on Build GRCh38
rs879454088 Chr.7: 1471228-1471228 on Build GRCh38
rs779660617 Chr.7: 1470799-1470799 on Build GRCh38
rs142451049 Chr.7: 1470777-1470777 on Build GRCh38
rs573260428 Chr.7: 1471211-1471211 on Build GRCh38
rs757838613 Chr.7: 1470981-1470981 on Build GRCh38
rs751303536 Chr.7: 1471230-1471230 on Build GRCh38
rs575032979 Chr.7: 1470886-1470886 on Build GRCh38
rs568701941 Chr.7: 1471122-1471122 on Build GRCh38
rs776593164 Chr.7: 1471138-1471138 on Build GRCh38
rs373264112 Chr.7: 1471216-1471216 on Build GRCh38
rs1043410770 Chr.7: 1471002-1471002 on Build GRCh38
rs376165628 Chr.7: 1471218-1471218 on Build GRCh38
rs777337356 Chr.7: 1470987-1470987 on Build GRCh38
rs953543404 Chr.7: 1471047-1471047 on Build GRCh38
rs376669537 Chr.7: 1470965-1470965 on Build GRCh38
rs904822263 Chr.7: 1471006-1471006 on Build GRCh38
rs944909609 Chr.7: 1470925-1470925 on Build GRCh38
rs748634098 Chr.7: 1470824-1470824 on Build GRCh38
rs541180800 Chr.7: 1471212-1471212 on Build GRCh38
rs752221459 Chr.7: 1471158-1471158 on Build GRCh38
rs150543082 Chr.7: 1471081-1471081 on Build GRCh38
rs761832765 Chr.7: 1471230-1471231 on Build GRCh38
rs773020115 Chr.7: 1470949-1470949 on Build GRCh38
rs750422522 Chr.7: 1470862-1470862 on Build GRCh38
rs767324323 Chr.7: 1470853-1470853 on Build GRCh38
rs1015946350 Chr.7: 1471017-1471017 on Build GRCh38
rs753422925 Chr.7: 1470878-1470878 on Build GRCh38
rs762917637 Chr.7: 1470945-1470945 on Build GRCh38
rs754589182 Chr.7: 1470968-1470968 on Build GRCh38
rs768551421 Chr.7: 1471119-1471119 on Build GRCh38
rs750775913 Chr.7: 1470789-1470789 on Build GRCh38
rs764840537 Chr.7: 1470969-1470969 on Build GRCh38
rs953498915 Chr.7: 1470879-1470879 on Build GRCh38
rs755849785 Chr.7: 1470874-1470874 on Build GRCh38
rs946977768 Chr.7: 1471104-1471104 on Build GRCh38
rs897481324 Chr.7: 1471189-1471189 on Build GRCh38
rs914042324 Chr.7: 1471104-1471107 on Build GRCh38
rs1033991380 Chr.7: 1470782-1470782 on Build GRCh38
rs889697431 Chr.7: 1471197-1471197 on Build GRCh38
rs749592045 Chr.7: 1471118-1471118 on Build GRCh38
rs759308683 Chr.7: 1471140-1471140 on Build GRCh38
rs928593625 Chr.7: 1471120-1471120 on Build GRCh38
rs754640136 Chr.7: 1470884-1470884 on Build GRCh38
rs545871067 Chr.7: 1470972-1470972 on Build GRCh38
rs752846485 Chr.7: 1470841-1470841 on Build GRCh38
rs771382458 Chr.7: 1470831-1470831 on Build GRCh38
rs555238349 Chr.7: 1470829-1470829 on Build GRCh38
rs756865735 Chr.7: 1471182-1471182 on Build GRCh38
rs775819627 Chr.7: 1470932-1470932 on Build GRCh38
rs150394409 Chr.7: 1471121-1471122 on Build GRCh38
rs561190996 Chr.7: 1471028-1471028 on Build GRCh38
rs753542479 Chr.7: 1470931-1470931 on Build GRCh38
rs972194496 Chr.7: 1470855-1470855 on Build GRCh38
rs543370673 Chr.7: 1470906-1470906 on Build GRCh38
rs200067270 Chr.7: 1470950-1470950 on Build GRCh38
rs1018096809 Chr.7: 1471072-1471072 on Build GRCh38
rs781654674 Chr.7: 1471097-1471097 on Build GRCh38
rs763566173 Chr.7: 1471014-1471014 on Build GRCh38
rs370540456 Chr.7: 1471229-1471229 on Build GRCh38
rs946945189 Chr.7: 1470947-1470947 on Build GRCh38
rs771656306 Chr.7: 1470889-1470889 on Build GRCh38
rs894183649 Chr.7: 1471160-1471160 on Build GRCh38
rs1055935674 Chr.7: 1471001-1471001 on Build GRCh38
rs772482806 Chr.7: 1470825-1470825 on Build GRCh38
rs1036875282 Chr.7: 1471184-1471184 on Build GRCh38
rs372260855 Chr.7: 1470806-1470806 on Build GRCh38
rs766105593 Chr.7: 1470962-1470962 on Build GRCh38
rs557258850 Chr.7: 1471194-1471194 on Build GRCh38
rs768150928 Chr.7: 1470807-1470807 on Build GRCh38
rs995391837 Chr.7: 1471034-1471034 on Build GRCh38
rs11974848 Chr.7: 1471089-1471089 on Build GRCh38
rs187219281 Chr.7: 1470946-1470946 on Build GRCh38
rs369044953 Chr.7: 1471106-1471106 on Build GRCh38
rs771350527 Chr.7: 1470902-1470902 on Build GRCh38
rs768059275 Chr.7: 1470778-1470778 on Build GRCh38
rs986354998 Chr.7: 1471071-1471071 on Build GRCh38
rs775786204 Chr.7: 1470810-1470810 on Build GRCh38
rs774258959 Chr.7: 1471206-1471206 on Build GRCh38
rs554871901 Chr.7: 1471098-1471098 on Build GRCh38
rs751110317 Chr.7: 1471181-1471181 on Build GRCh38
rs779893699 Chr.7: 1470876-1470876 on Build GRCh38
rs896259583 Chr.7: 1471150-1471150 on Build GRCh38

View Full Product Details