Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2362553-2363042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3B
Entrez Gene ID
8662
Gene Name
eukaryotic translation initiation factor 3 subunit B
Gene Aliases
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1
UniGene
Hs.371001
Gene Chromosome Location
Chr.7: 2354540-2380745 on Build GRCh38
Transcripts
NM_001037283.1 NM_003751.3 XM_011515599.1 XM_011515600.1 XM_011515601.1 XM_017012752.1 XM_017012753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736428 c.758A>G p.Y253C Chr.7: 2362710-2362710 on Build GRCh38
4506352 c.717C>T p.S239S Chr.7: 2362669-2362669 on Build GRCh38
3923415 c.702C>T p.F234F Chr.7: 2362654-2362654 on Build GRCh38
7285329 c.771G>T p.K257N Chr.7: 2362723-2362723 on Build GRCh38
3880300 c.695A>G p.Y232C Chr.7: 2362647-2362647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768254136 Chr.7: 2362641-2362641 on Build GRCh38
rs776816526 Chr.7: 2362732-2362732 on Build GRCh38
rs781151737 Chr.7: 2362634-2362634 on Build GRCh38
rs754528550 Chr.7: 2362699-2362699 on Build GRCh38
rs545631871 Chr.7: 2362927-2362927 on Build GRCh38
rs748255445 Chr.7: 2362711-2362711 on Build GRCh38
rs761291355 Chr.7: 2363012-2363012 on Build GRCh38
rs201852385 Chr.7: 2362599-2362599 on Build GRCh38
rs181640069 Chr.7: 2362580-2362580 on Build GRCh38
rs575674802 Chr.7: 2362827-2362827 on Build GRCh38
rs181856373 Chr.7: 2362792-2362792 on Build GRCh38
rs555746352 Chr.7: 2362594-2362594 on Build GRCh38
rs373392664 Chr.7: 2362736-2362736 on Build GRCh38
rs778485654 Chr.7: 2362772-2362772 on Build GRCh38
rs766652294 Chr.7: 2362621-2362621 on Build GRCh38
rs892472286 Chr.7: 2362596-2362596 on Build GRCh38
rs200218143 Chr.7: 2362685-2362685 on Build GRCh38
rs774972775 Chr.7: 2362603-2362603 on Build GRCh38
rs570068196 Chr.7: 2362700-2362700 on Build GRCh38
rs890011082 Chr.7: 2362989-2362989 on Build GRCh38
rs775147509 Chr.7: 2362598-2362598 on Build GRCh38
rs373465954 Chr.7: 2362798-2362798 on Build GRCh38
rs768040027 Chr.7: 2362601-2362601 on Build GRCh38
rs774939354 Chr.7: 2362604-2362606 on Build GRCh38
rs944833342 Chr.7: 2362783-2362783 on Build GRCh38
rs202013695 Chr.7: 2362803-2362803 on Build GRCh38
rs184289390 Chr.7: 2362631-2362631 on Build GRCh38
rs765195711 Chr.7: 2362628-2362628 on Build GRCh38
rs774652838 Chr.7: 2362702-2362702 on Build GRCh38
rs148466865 Chr.7: 2362998-2362998 on Build GRCh38
rs140288121 Chr.7: 2362663-2362663 on Build GRCh38
rs553946799 Chr.7: 2362796-2362796 on Build GRCh38
rs762464365 Chr.7: 2362799-2362809 on Build GRCh38
rs914053845 Chr.7: 2362984-2362984 on Build GRCh38
rs745594871 Chr.7: 2362636-2362636 on Build GRCh38
rs535162743 Chr.7: 2362665-2362665 on Build GRCh38
rs112539906 Chr.7: 2362604-2362604 on Build GRCh38
rs764248115 Chr.7: 2362749-2362749 on Build GRCh38
rs1051266937 Chr.7: 2362971-2362971 on Build GRCh38
rs781626198 Chr.7: 2362797-2362797 on Build GRCh38
rs746288409 Chr.7: 2362802-2362802 on Build GRCh38
rs372385087 Chr.7: 2362611-2362611 on Build GRCh38
rs905096445 Chr.7: 2362626-2362626 on Build GRCh38
rs769516465 Chr.7: 2362597-2362597 on Build GRCh38
rs756024372 Chr.7: 2362687-2362687 on Build GRCh38
rs186986516 Chr.7: 2362824-2362824 on Build GRCh38
rs773506091 Chr.7: 2362609-2362609 on Build GRCh38
rs776874073 Chr.7: 2362666-2362666 on Build GRCh38
rs766704173 Chr.7: 2362891-2362891 on Build GRCh38
rs1029905119 Chr.7: 2362739-2362739 on Build GRCh38
rs191057443 Chr.7: 2362892-2362892 on Build GRCh38
rs754796959 Chr.7: 2362770-2362770 on Build GRCh38
rs200489277 Chr.7: 2362706-2362706 on Build GRCh38
rs748871874 Chr.7: 2362640-2362640 on Build GRCh38
rs762171100 Chr.7: 2362843-2362843 on Build GRCh38
rs771555375 Chr.7: 2362705-2362705 on Build GRCh38
rs1035345451 Chr.7: 2362859-2362859 on Build GRCh38
rs201931717 Chr.7: 2362639-2362639 on Build GRCh38
rs755971349 Chr.7: 2362638-2362638 on Build GRCh38
rs116770364 Chr.7: 2362753-2362753 on Build GRCh38
rs763185237 Chr.7: 2362745-2362745 on Build GRCh38
rs150739455 Chr.7: 2362680-2362680 on Build GRCh38
rs954086025 Chr.7: 2362572-2362572 on Build GRCh38
rs1015365260 Chr.7: 2362872-2362872 on Build GRCh38
rs1009010642 Chr.7: 2362581-2362581 on Build GRCh38
rs536347353 Chr.7: 2362819-2362819 on Build GRCh38
rs369741313 Chr.7: 2362696-2362696 on Build GRCh38
rs1028020144 Chr.7: 2362920-2362920 on Build GRCh38
rs747479647 Chr.7: 2362708-2362708 on Build GRCh38
rs748701494 Chr.7: 2362660-2362660 on Build GRCh38
rs908878341 Chr.7: 2362940-2362940 on Build GRCh38
rs779965692 Chr.7: 2362689-2362689 on Build GRCh38
rs542321302 Chr.7: 2362588-2362588 on Build GRCh38
rs745541864 Chr.7: 2362595-2362595 on Build GRCh38
rs757941589 Chr.7: 2362781-2362781 on Build GRCh38
rs752665303 Chr.7: 2362679-2362679 on Build GRCh38
rs953834192 Chr.7: 2362932-2362932 on Build GRCh38
rs988247119 Chr.7: 2362974-2362974 on Build GRCh38
rs759633167 Chr.7: 2362670-2362670 on Build GRCh38
rs926086805 Chr.7: 2362582-2362582 on Build GRCh38
rs986404101 Chr.7: 2362939-2362939 on Build GRCh38
rs896322206 Chr.7: 2362865-2362865 on Build GRCh38
rs897322836 Chr.7: 2362961-2362961 on Build GRCh38
rs1002162323 Chr.7: 2362848-2362848 on Build GRCh38
rs538234524 Chr.7: 2362844-2362844 on Build GRCh38
rs777221361 Chr.7: 2362791-2362791 on Build GRCh38
rs767631913 Chr.7: 2362684-2362684 on Build GRCh38
rs769844132 Chr.7: 2362737-2362737 on Build GRCh38
rs557656982 Chr.7: 2362949-2362949 on Build GRCh38
rs767486333 Chr.7: 2362866-2362866 on Build GRCh38
rs973797277 Chr.7: 2362911-2362911 on Build GRCh38
rs927220452 Chr.7: 2362880-2362880 on Build GRCh38
rs879262626 Chr.7: 2362760-2362760 on Build GRCh38
rs998336343 Chr.7: 2362963-2362963 on Build GRCh38
rs759751241 Chr.7: 2362972-2362972 on Build GRCh38
rs767697558 Chr.7: 2363021-2363022 on Build GRCh38
rs940415477 Chr.7: 2362946-2362946 on Build GRCh38
rs912105440 Chr.7: 2362758-2362758 on Build GRCh38
rs753509340 Chr.7: 2362767-2362767 on Build GRCh38
rs775778858 Chr.7: 2362743-2362743 on Build GRCh38
rs778237987 Chr.7: 2362647-2362647 on Build GRCh38
rs146230401 Chr.7: 2362678-2362678 on Build GRCh38
rs541329801 Chr.7: 2362997-2362997 on Build GRCh38
rs751551651 Chr.7: 2362629-2362629 on Build GRCh38
rs551734577 Chr.7: 2362752-2362752 on Build GRCh38
rs754043032 Chr.7: 2362624-2362624 on Build GRCh38
rs200969617 Chr.7: 2362693-2362693 on Build GRCh38
rs752369523 Chr.7: 2362776-2362776 on Build GRCh38
rs561298082 Chr.7: 2362664-2362664 on Build GRCh38
rs755131262 Chr.7: 2362627-2362627 on Build GRCh38
rs946831984 Chr.7: 2362996-2362996 on Build GRCh38

View Full Product Details