Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75534398-75534944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1
Entrez Gene ID
3092
Gene Name
huntingtin interacting protein 1
Gene Aliases
HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
UniGene
Hs.329266
Gene Chromosome Location
Chr.7: 75533298-75738976 on Build GRCh38
Transcripts
NM_001243198.2 NM_005338.6 XM_005250304.2 XM_005250305.2 XM_011516116.2 XM_017012099.1

SNP Detail

SNP ID Chromosome Location
rs367974587 Chr.7: 75534898-75534898 on Build GRCh38
rs371553830 Chr.7: 75534882-75534882 on Build GRCh38
rs919652879 Chr.7: 75534883-75534883 on Build GRCh38
rs905064769 Chr.7: 75534675-75534675 on Build GRCh38
rs587654896 Chr.7: 75534578-75534578 on Build GRCh38
rs190784856 Chr.7: 75534622-75534622 on Build GRCh38
rs942462492 Chr.7: 75534552-75534552 on Build GRCh38
rs587648670 Chr.7: 75534487-75534487 on Build GRCh38
rs75801816 Chr.7: 75534644-75534644 on Build GRCh38
rs782147618 Chr.7: 75534660-75534660 on Build GRCh38
rs988166130 Chr.7: 75534859-75534859 on Build GRCh38
rs910710205 Chr.7: 75534862-75534862 on Build GRCh38
rs587764392 Chr.7: 75534844-75534844 on Build GRCh38
rs587724850 Chr.7: 75534651-75534651 on Build GRCh38
rs910733925 Chr.7: 75534543-75534543 on Build GRCh38
rs958113265 Chr.7: 75534751-75534751 on Build GRCh38
rs587639183 Chr.7: 75534623-75534623 on Build GRCh38
rs769092802 Chr.7: 75534608-75534608 on Build GRCh38
rs1039460074 Chr.7: 75534554-75534554 on Build GRCh38
rs1002283341 Chr.7: 75534681-75534681 on Build GRCh38
rs587760001 Chr.7: 75534526-75534526 on Build GRCh38
rs6953853 Chr.7: 75534496-75534496 on Build GRCh38
rs782284084 Chr.7: 75534439-75534440 on Build GRCh38
rs782544422 Chr.7: 75534832-75534832 on Build GRCh38
rs931075980 Chr.7: 75534893-75534893 on Build GRCh38
rs183300075 Chr.7: 75534688-75534688 on Build GRCh38
rs587623153 Chr.7: 75534758-75534758 on Build GRCh38
rs587678659 Chr.7: 75534807-75534807 on Build GRCh38
rs782156967 Chr.7: 75534889-75534889 on Build GRCh38
rs587599252 Chr.7: 75534664-75534664 on Build GRCh38
rs995093311 Chr.7: 75534579-75534579 on Build GRCh38
rs933579828 Chr.7: 75534516-75534516 on Build GRCh38
rs587695482 Chr.7: 75534525-75534525 on Build GRCh38
rs988117656 Chr.7: 75534770-75534770 on Build GRCh38
rs943506444 Chr.7: 75534871-75534871 on Build GRCh38
rs148560744 Chr.7: 75534475-75534475 on Build GRCh38
rs1021401055 Chr.7: 75534837-75534837 on Build GRCh38
rs1051924277 Chr.7: 75534527-75534527 on Build GRCh38
rs587672350 Chr.7: 75534665-75534665 on Build GRCh38
rs78407086 Chr.7: 75534635-75534635 on Build GRCh38

View Full Product Details