Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87934418-87934909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
UniGene
Hs.256398
Gene Chromosome Location
Chr.7: 87934143-88202889 on Build GRCh38
Transcripts
NM_001324417.1 NM_001324418.1 NM_001324419.1 NM_001324420.1 NM_001324421.1 NM_004194.4 NM_016351.5 NM_021721.4 NM_021722.5 NM_021723.4 XM_005250445.3 XM_006716028.3 XM_006716029.3 XM_011516318.1 XM_011516319.2 XM_011516320.2 XM_011516321.2 XM_011516322.1 XM_011516323.1 XM_017012329.1 XM_017012330.1 XM_017012331.1 XM_017012332.1 XM_017012333.1 XM_017012334.1 XM_017012335.1 XM_017012336.1 XM_017012337.1 XM_017012338.1 XM_017012339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7224728 c.85+9G>A p.? Chr.7: 87934559-87934559 on Build GRCh38
393806 c.78C>A p.G26G Chr.7: 87934543-87934543 on Build GRCh38
5737987 c.3G>A p.M1I Chr.7: 87934468-87934468 on Build GRCh38
7248407 c.6G>T p.Q2H Chr.7: 87934471-87934471 on Build GRCh38
26542 c.7G>A p.A3T Chr.7: 87934472-87934472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894179398 Chr.7: 87934732-87934732 on Build GRCh38
rs532748835 Chr.7: 87934542-87934542 on Build GRCh38
rs548912375 Chr.7: 87934821-87934821 on Build GRCh38
rs772855169 Chr.7: 87934545-87934545 on Build GRCh38
rs748835811 Chr.7: 87934854-87934854 on Build GRCh38
rs555816821 Chr.7: 87934661-87934661 on Build GRCh38
rs185661659 Chr.7: 87934489-87934489 on Build GRCh38
rs928443899 Chr.7: 87934610-87934610 on Build GRCh38
rs750517248 Chr.7: 87934886-87934886 on Build GRCh38
rs775345905 Chr.7: 87934661-87934663 on Build GRCh38
rs1039006553 Chr.7: 87934671-87934671 on Build GRCh38
rs754929186 Chr.7: 87934600-87934600 on Build GRCh38
rs777885105 Chr.7: 87934490-87934490 on Build GRCh38
rs769586574 Chr.7: 87934824-87934824 on Build GRCh38
rs760970406 Chr.7: 87934868-87934868 on Build GRCh38
rs756394824 Chr.7: 87934564-87934564 on Build GRCh38
rs766928591 Chr.7: 87934456-87934456 on Build GRCh38
rs931080581 Chr.7: 87934689-87934689 on Build GRCh38
rs916290088 Chr.7: 87934554-87934554 on Build GRCh38
rs1048620752 Chr.7: 87934810-87934810 on Build GRCh38
rs753092752 Chr.7: 87934563-87934563 on Build GRCh38
rs767014451 Chr.7: 87934553-87934553 on Build GRCh38
rs746855564 Chr.7: 87934526-87934526 on Build GRCh38
rs373981946 Chr.7: 87934473-87934473 on Build GRCh38
rs764333345 Chr.7: 87934871-87934871 on Build GRCh38
rs779920389 Chr.7: 87934523-87934523 on Build GRCh38
rs548023517 Chr.7: 87934486-87934486 on Build GRCh38
rs953368102 Chr.7: 87934875-87934875 on Build GRCh38
rs1055634256 Chr.7: 87934654-87934654 on Build GRCh38
rs893190534 Chr.7: 87934656-87934656 on Build GRCh38
rs57360340 Chr.7: 87934680-87934680 on Build GRCh38
rs1053555368 Chr.7: 87934594-87934594 on Build GRCh38
rs776468673 Chr.7: 87934534-87934534 on Build GRCh38
rs201972241 Chr.7: 87934479-87934479 on Build GRCh38
rs1041521163 Chr.7: 87934585-87934585 on Build GRCh38
rs746969143 Chr.7: 87934593-87934593 on Build GRCh38
rs879033031 Chr.7: 87934661-87934662 on Build GRCh38
rs1005014232 Chr.7: 87934783-87934783 on Build GRCh38
rs1015610589 Chr.7: 87934507-87934507 on Build GRCh38
rs1029242605 Chr.7: 87934869-87934869 on Build GRCh38
rs773931477 Chr.7: 87934753-87934753 on Build GRCh38
rs760053360 Chr.7: 87934561-87934561 on Build GRCh38
rs757450527 Chr.7: 87934874-87934874 on Build GRCh38
rs900523579 Chr.7: 87934677-87934677 on Build GRCh38
rs1016447864 Chr.7: 87934788-87934788 on Build GRCh38
rs201363585 Chr.7: 87934582-87934582 on Build GRCh38
rs768379580 Chr.7: 87934531-87934531 on Build GRCh38
rs747812997 Chr.7: 87934536-87934536 on Build GRCh38
rs867229265 Chr.7: 87934518-87934518 on Build GRCh38
rs770602297 Chr.7: 87934856-87934856 on Build GRCh38
rs776355468 Chr.7: 87934474-87934479 on Build GRCh38
rs966292793 Chr.7: 87934522-87934522 on Build GRCh38
rs938622253 Chr.7: 87934611-87934611 on Build GRCh38
rs567405608 Chr.7: 87934840-87934840 on Build GRCh38
rs2279541 Chr.7: 87934864-87934864 on Build GRCh38
rs886836982 Chr.7: 87934816-87934816 on Build GRCh38
rs984902284 Chr.7: 87934693-87934693 on Build GRCh38
rs751965106 Chr.7: 87934464-87934464 on Build GRCh38
rs951015345 Chr.7: 87934779-87934779 on Build GRCh38
rs368425480 Chr.7: 87934474-87934474 on Build GRCh38
rs990451039 Chr.7: 87934546-87934546 on Build GRCh38
rs921951954 Chr.7: 87934535-87934535 on Build GRCh38
rs762496835 Chr.7: 87934547-87934547 on Build GRCh38
rs376585097 Chr.7: 87934579-87934579 on Build GRCh38
rs749362618 Chr.7: 87934519-87934519 on Build GRCh38
rs148253397 Chr.7: 87934559-87934559 on Build GRCh38
rs372487928 Chr.7: 87934451-87934451 on Build GRCh38
rs769468264 Chr.7: 87934662-87934663 on Build GRCh38
rs760008751 Chr.7: 87934483-87934484 on Build GRCh38
rs367685028 Chr.7: 87934589-87934589 on Build GRCh38
rs997809410 Chr.7: 87934866-87934866 on Build GRCh38
rs764233161 Chr.7: 87934565-87934565 on Build GRCh38
rs1011652309 Chr.7: 87934660-87934660 on Build GRCh38
rs765291661 Chr.7: 87934885-87934885 on Build GRCh38
rs781507898 Chr.7: 87934480-87934480 on Build GRCh38
rs997764026 Chr.7: 87934687-87934687 on Build GRCh38
rs771998986 Chr.7: 87934521-87934521 on Build GRCh38
rs760019519 Chr.7: 87934860-87934860 on Build GRCh38
rs1005011966 Chr.7: 87934492-87934492 on Build GRCh38
rs368718490 Chr.7: 87934530-87934530 on Build GRCh38
rs189387539 Chr.7: 87934752-87934752 on Build GRCh38
rs773885594 Chr.7: 87934859-87934859 on Build GRCh38
rs776056320 Chr.7: 87934757-87934757 on Build GRCh38

View Full Product Details