Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90899203-90899714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK14
Entrez Gene ID
5218
Gene Name
cyclin dependent kinase 14
Gene Aliases
PFTAIRE1, PFTK1
UniGene
Hs.258576
Gene Chromosome Location
Chr.7: 90595994-91210590 on Build GRCh38
Transcripts
NM_001287135.1 NM_001287136.1 NM_001287137.1 NM_012395.3 XM_005250436.3 XM_005250438.3 XM_005250439.2 XM_011516306.2 XM_011516307.2 XM_017012320.1 XM_017012321.1 XM_017012322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1452717 c.628C>A p.L210M Chr.7: 90899333-90899333 on Build GRCh38
4671100 c.667A>C p.K223Q Chr.7: 90899318-90899318 on Build GRCh38
6434893 c.662T>A p.M221K Chr.7: 90899313-90899313 on Build GRCh38
3778668 c.689C>T p.P230L Chr.7: 90899340-90899340 on Build GRCh38
4555179 c.643G>T p.V215L Chr.7: 90899348-90899348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370776469 Chr.7: 90899308-90899308 on Build GRCh38
rs962016644 Chr.7: 90899433-90899433 on Build GRCh38
rs767470602 Chr.7: 90899380-90899380 on Build GRCh38
rs761968001 Chr.7: 90899383-90899383 on Build GRCh38
rs757328135 Chr.7: 90899259-90899259 on Build GRCh38
rs760122649 Chr.7: 90899339-90899339 on Build GRCh38
rs144476335 Chr.7: 90899674-90899674 on Build GRCh38
rs773939489 Chr.7: 90899337-90899337 on Build GRCh38
rs993848328 Chr.7: 90899519-90899519 on Build GRCh38
rs955048360 Chr.7: 90899547-90899547 on Build GRCh38
rs769531582 Chr.7: 90899328-90899328 on Build GRCh38
rs754083023 Chr.7: 90899436-90899436 on Build GRCh38
rs534530782 Chr.7: 90899581-90899581 on Build GRCh38
rs187569768 Chr.7: 90899488-90899488 on Build GRCh38
rs765222047 Chr.7: 90899590-90899590 on Build GRCh38
rs141395840 Chr.7: 90899347-90899347 on Build GRCh38
rs181488348 Chr.7: 90899456-90899456 on Build GRCh38
rs967913829 Chr.7: 90899630-90899630 on Build GRCh38
rs946186011 Chr.7: 90899289-90899289 on Build GRCh38
rs184702498 Chr.7: 90899335-90899335 on Build GRCh38
rs373874746 Chr.7: 90899340-90899340 on Build GRCh38
rs566859740 Chr.7: 90899396-90899396 on Build GRCh38
rs754802356 Chr.7: 90899276-90899276 on Build GRCh38
rs772914218 Chr.7: 90899424-90899424 on Build GRCh38
rs993709180 Chr.7: 90899444-90899444 on Build GRCh38
rs111539926 Chr.7: 90899450-90899450 on Build GRCh38
rs896917240 Chr.7: 90899497-90899497 on Build GRCh38
rs979262726 Chr.7: 90899641-90899641 on Build GRCh38
rs751531255 Chr.7: 90899267-90899267 on Build GRCh38
rs545118306 Chr.7: 90899629-90899629 on Build GRCh38
rs894140153 Chr.7: 90899360-90899360 on Build GRCh38
rs988890384 Chr.7: 90899679-90899679 on Build GRCh38
rs950370752 Chr.7: 90899453-90899453 on Build GRCh38
rs923737884 Chr.7: 90899664-90899664 on Build GRCh38
rs764370281 Chr.7: 90899381-90899381 on Build GRCh38
rs926025814 Chr.7: 90899428-90899428 on Build GRCh38
rs972373326 Chr.7: 90899291-90899291 on Build GRCh38
rs747559535 Chr.7: 90899675-90899675 on Build GRCh38
rs906072924 Chr.7: 90899475-90899475 on Build GRCh38
rs1029760371 Chr.7: 90899521-90899521 on Build GRCh38
rs553489524 Chr.7: 90899568-90899568 on Build GRCh38
rs1005671048 Chr.7: 90899487-90899487 on Build GRCh38
rs777536906 Chr.7: 90899331-90899331 on Build GRCh38
rs547416380 Chr.7: 90899443-90899443 on Build GRCh38
rs760921761 Chr.7: 90899366-90899366 on Build GRCh38
rs55664580 Chr.7: 90899332-90899332 on Build GRCh38
rs554827965 Chr.7: 90899471-90899471 on Build GRCh38
rs754961863 Chr.7: 90899387-90899387 on Build GRCh38
rs1044637165 Chr.7: 90899464-90899464 on Build GRCh38
rs572808509 Chr.7: 90899627-90899627 on Build GRCh38
rs142742845 Chr.7: 90899385-90899385 on Build GRCh38
rs3779569 Chr.7: 90899608-90899608 on Build GRCh38
rs747845379 Chr.7: 90899323-90899323 on Build GRCh38
rs569104347 Chr.7: 90899451-90899451 on Build GRCh38
rs914792849 Chr.7: 90899546-90899546 on Build GRCh38
rs1005830201 Chr.7: 90899689-90899689 on Build GRCh38
rs758768737 Chr.7: 90899613-90899613 on Build GRCh38
rs914866025 Chr.7: 90899257-90899257 on Build GRCh38
rs917264268 Chr.7: 90899687-90899687 on Build GRCh38
rs190268211 Chr.7: 90899420-90899420 on Build GRCh38
rs753962793 Chr.7: 90899243-90899243 on Build GRCh38
rs558734494 Chr.7: 90899650-90899650 on Build GRCh38
rs1042240915 Chr.7: 90899585-90899586 on Build GRCh38

View Full Product Details