Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90917326-90917832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK14
Entrez Gene ID
5218
Gene Name
cyclin dependent kinase 14
Gene Aliases
PFTAIRE1, PFTK1
UniGene
Hs.258576
Gene Chromosome Location
Chr.7: 90595994-91210590 on Build GRCh38
Transcripts
NM_001287135.1 NM_001287136.1 NM_001287137.1 NM_012395.3 XM_005250436.3 XM_005250438.3 XM_005250439.2 XM_011516306.2 XM_011516307.2 XM_017012320.1 XM_017012321.1 XM_017012322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6291600 c.705G>C p.L235F Chr.7: 90917603-90917603 on Build GRCh38
1496725 c.800C>A p.T267K Chr.7: 90917698-90917698 on Build GRCh38
5004904 c.747G>A p.T249T Chr.7: 90917699-90917699 on Build GRCh38
3725223 c.796G>C p.D266H Chr.7: 90917694-90917694 on Build GRCh38
7230913 c.826G>T p.G276C Chr.7: 90917724-90917724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776916826 Chr.7: 90917651-90917651 on Build GRCh38
rs762051742 Chr.7: 90917659-90917659 on Build GRCh38
rs778399581 Chr.7: 90917745-90917745 on Build GRCh38
rs757836661 Chr.7: 90917760-90917760 on Build GRCh38
rs972198687 Chr.7: 90917452-90917452 on Build GRCh38
rs981179721 Chr.7: 90917383-90917383 on Build GRCh38
rs937756719 Chr.7: 90917801-90917801 on Build GRCh38
rs919062069 Chr.7: 90917509-90917509 on Build GRCh38
rs753641002 Chr.7: 90917739-90917739 on Build GRCh38
rs745349146 Chr.7: 90917599-90917599 on Build GRCh38
rs748960391 Chr.7: 90917590-90917590 on Build GRCh38
rs779561344 Chr.7: 90917761-90917761 on Build GRCh38
rs757021007 Chr.7: 90917744-90917744 on Build GRCh38
rs140888577 Chr.7: 90917382-90917382 on Build GRCh38
rs756057096 Chr.7: 90917724-90917724 on Build GRCh38
rs181847836 Chr.7: 90917554-90917554 on Build GRCh38
rs762907025 Chr.7: 90917699-90917699 on Build GRCh38
rs954948336 Chr.7: 90917482-90917482 on Build GRCh38
rs904211229 Chr.7: 90917636-90917636 on Build GRCh38
rs933168558 Chr.7: 90917537-90917537 on Build GRCh38
rs2286696 Chr.7: 90917449-90917449 on Build GRCh38
rs982285891 Chr.7: 90917762-90917762 on Build GRCh38
rs749910601 Chr.7: 90917516-90917516 on Build GRCh38
rs376270298 Chr.7: 90917698-90917698 on Build GRCh38
rs769024611 Chr.7: 90917647-90917647 on Build GRCh38
rs1007680269 Chr.7: 90917543-90917543 on Build GRCh38
rs78194207 Chr.7: 90917804-90917804 on Build GRCh38
rs746727809 Chr.7: 90917589-90917591 on Build GRCh38
rs1055191956 Chr.7: 90917373-90917373 on Build GRCh38
rs747522323 Chr.7: 90917768-90917768 on Build GRCh38
rs267601623 Chr.7: 90917628-90917628 on Build GRCh38
rs943323405 Chr.7: 90917630-90917630 on Build GRCh38
rs775861080 Chr.7: 90917644-90917644 on Build GRCh38
rs770669675 Chr.7: 90917592-90917592 on Build GRCh38
rs763984267 Chr.7: 90917733-90917733 on Build GRCh38
rs150860380 Chr.7: 90917632-90917632 on Build GRCh38
rs889029403 Chr.7: 90917614-90917614 on Build GRCh38
rs928189479 Chr.7: 90917790-90917790 on Build GRCh38
rs202149959 Chr.7: 90917723-90917723 on Build GRCh38
rs997993766 Chr.7: 90917448-90917448 on Build GRCh38
rs745453618 Chr.7: 90917751-90917751 on Build GRCh38
rs368500952 Chr.7: 90917717-90917717 on Build GRCh38
rs764769505 Chr.7: 90917453-90917453 on Build GRCh38
rs139294759 Chr.7: 90917681-90917681 on Build GRCh38
rs539919776 Chr.7: 90917381-90917381 on Build GRCh38
rs769108373 Chr.7: 90917772-90917772 on Build GRCh38
rs755969118 Chr.7: 90917571-90917571 on Build GRCh38
rs761163212 Chr.7: 90917645-90917645 on Build GRCh38
rs726303 Chr.7: 90917438-90917438 on Build GRCh38
rs1013762112 Chr.7: 90917802-90917802 on Build GRCh38
rs778508638 Chr.7: 90917598-90917598 on Build GRCh38
rs752577694 Chr.7: 90917560-90917560 on Build GRCh38
rs758302947 Chr.7: 90917646-90917646 on Build GRCh38

View Full Product Details