Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93219219-93219729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPACAM2
Entrez Gene ID
253012
Gene Name
HEPACAM family member 2
Gene Aliases
MIKI
UniGene
Hs.443169
Gene Chromosome Location
Chr.7: 93188586-93232297 on Build GRCh38
Transcripts
NM_001039372.2 NM_001288804.1 NM_001288810.1 NM_198151.2 XM_011516000.2 XM_011516001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447525 c.144A>C p.P48P Chr.7: 93219351-93219351 on Build GRCh38
5873389 c.155C>T p.P52L Chr.7: 93219376-93219376 on Build GRCh38
1163478 c.83C>T p.T28I Chr.7: 93219412-93219412 on Build GRCh38
5721004 c.32C>T p.S11F Chr.7: 93219463-93219463 on Build GRCh38
2864661 c.71T>G p.V24G Chr.7: 93219424-93219424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750096728 Chr.7: 93219422-93219422 on Build GRCh38
rs183808462 Chr.7: 93219601-93219601 on Build GRCh38
rs774470334 Chr.7: 93219298-93219298 on Build GRCh38
rs193921119 Chr.7: 93219492-93219492 on Build GRCh38
rs779135164 Chr.7: 93219378-93219378 on Build GRCh38
rs551796395 Chr.7: 93219657-93219657 on Build GRCh38
rs563747542 Chr.7: 93219438-93219438 on Build GRCh38
rs750057761 Chr.7: 93219350-93219350 on Build GRCh38
rs769990226 Chr.7: 93219266-93219266 on Build GRCh38
rs775904906 Chr.7: 93219329-93219329 on Build GRCh38
rs747334017 Chr.7: 93219472-93219472 on Build GRCh38
rs777461135 Chr.7: 93219409-93219410 on Build GRCh38
rs61733337 Chr.7: 93219520-93219520 on Build GRCh38
rs1013510625 Chr.7: 93219637-93219637 on Build GRCh38
rs751530261 Chr.7: 93219369-93219369 on Build GRCh38
rs1055160459 Chr.7: 93219569-93219569 on Build GRCh38
rs893247512 Chr.7: 93219439-93219439 on Build GRCh38
rs762666268 Chr.7: 93219523-93219523 on Build GRCh38
rs568671793 Chr.7: 93219656-93219656 on Build GRCh38
rs139778626 Chr.7: 93219397-93219397 on Build GRCh38
rs772419396 Chr.7: 93219386-93219386 on Build GRCh38
rs777035702 Chr.7: 93219406-93219406 on Build GRCh38
rs769243743 Chr.7: 93219401-93219401 on Build GRCh38
rs1040090189 Chr.7: 93219423-93219423 on Build GRCh38
rs914134985 Chr.7: 93219454-93219454 on Build GRCh38
rs780364371 Chr.7: 93219459-93219459 on Build GRCh38
rs766091916 Chr.7: 93219544-93219544 on Build GRCh38
rs962757991 Chr.7: 93219247-93219247 on Build GRCh38
rs777968754 Chr.7: 93219272-93219272 on Build GRCh38
rs1011938743 Chr.7: 93219591-93219591 on Build GRCh38
rs758202094 Chr.7: 93219352-93219352 on Build GRCh38
rs1043868133 Chr.7: 93219483-93219483 on Build GRCh38
rs746074655 Chr.7: 93219453-93219453 on Build GRCh38
rs550374974 Chr.7: 93219491-93219491 on Build GRCh38
rs986479773 Chr.7: 93219448-93219448 on Build GRCh38
rs146241924 Chr.7: 93219381-93219381 on Build GRCh38
rs763580579 Chr.7: 93219332-93219332 on Build GRCh38
rs762753952 Chr.7: 93219426-93219426 on Build GRCh38
rs772900144 Chr.7: 93219521-93219521 on Build GRCh38
rs368471128 Chr.7: 93219455-93219455 on Build GRCh38
rs761146462 Chr.7: 93219395-93219395 on Build GRCh38
rs565412217 Chr.7: 93219669-93219669 on Build GRCh38
rs1004999742 Chr.7: 93219320-93219320 on Build GRCh38
rs529480032 Chr.7: 93219703-93219703 on Build GRCh38
rs150604467 Chr.7: 93219375-93219375 on Build GRCh38
rs770940799 Chr.7: 93219376-93219376 on Build GRCh38
rs759848399 Chr.7: 93219300-93219300 on Build GRCh38
rs142784565 Chr.7: 93219374-93219374 on Build GRCh38
rs755018349 Chr.7: 93219371-93219371 on Build GRCh38
rs542955475 Chr.7: 93219577-93219577 on Build GRCh38
rs772437645 Chr.7: 93219314-93219314 on Build GRCh38
rs761336938 Chr.7: 93219409-93219409 on Build GRCh38
rs747390368 Chr.7: 93219534-93219535 on Build GRCh38
rs371808142 Chr.7: 93219432-93219432 on Build GRCh38
rs10281525 Chr.7: 93219274-93219274 on Build GRCh38
rs775888411 Chr.7: 93219389-93219389 on Build GRCh38
rs768923425 Chr.7: 93219512-93219512 on Build GRCh38
rs557473967 Chr.7: 93219358-93219358 on Build GRCh38
rs771229311 Chr.7: 93219295-93219295 on Build GRCh38
rs375176875 Chr.7: 93219441-93219441 on Build GRCh38
rs764659725 Chr.7: 93219421-93219421 on Build GRCh38
rs371594356 Chr.7: 93219502-93219502 on Build GRCh38
rs1016757912 Chr.7: 93219704-93219704 on Build GRCh38
rs751442611 Chr.7: 93219431-93219431 on Build GRCh38
rs148519653 Chr.7: 93219338-93219338 on Build GRCh38
rs368882066 Chr.7: 93219342-93219342 on Build GRCh38
rs748669977 Chr.7: 93219517-93219517 on Build GRCh38
rs752593668 Chr.7: 93219437-93219437 on Build GRCh38
rs930229382 Chr.7: 93219373-93219373 on Build GRCh38

View Full Product Details