Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93991842-93992391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BET1
Entrez Gene ID
10282
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
HBET1
UniGene
Hs.489132
Gene Chromosome Location
Chr.7: 93962770-94004382 on Build GRCh38
Transcripts
NR_133908.1 NR_133909.1

SNP Detail

SNP ID Chromosome Location
rs1007324954 Chr.7: 93992346-93992346 on Build GRCh38
rs186127297 Chr.7: 93992164-93992164 on Build GRCh38
rs910455711 Chr.7: 93992245-93992245 on Build GRCh38
rs1052367043 Chr.7: 93991957-93991957 on Build GRCh38
rs976307893 Chr.7: 93992304-93992304 on Build GRCh38
rs558175441 Chr.7: 93992249-93992249 on Build GRCh38
rs6945362 Chr.7: 93992081-93992081 on Build GRCh38
rs1027292875 Chr.7: 93992204-93992204 on Build GRCh38
rs574765420 Chr.7: 93992335-93992335 on Build GRCh38
rs1030073881 Chr.7: 93991914-93991914 on Build GRCh38
rs548653071 Chr.7: 93992088-93992088 on Build GRCh38
rs1020293397 Chr.7: 93992047-93992047 on Build GRCh38
rs1011129624 Chr.7: 93991970-93991970 on Build GRCh38
rs967403760 Chr.7: 93992080-93992080 on Build GRCh38
rs1036613317 Chr.7: 93992189-93992189 on Build GRCh38
rs549519963 Chr.7: 93991980-93991980 on Build GRCh38
rs958179813 Chr.7: 93992031-93992031 on Build GRCh38
rs180672382 Chr.7: 93991892-93991892 on Build GRCh38
rs1032195963 Chr.7: 93992011-93992011 on Build GRCh38
rs145225837 Chr.7: 93992161-93992161 on Build GRCh38
rs961778772 Chr.7: 93992151-93992151 on Build GRCh38
rs935394070 Chr.7: 93992367-93992367 on Build GRCh38
rs534601623 Chr.7: 93992168-93992168 on Build GRCh38
rs951808212 Chr.7: 93992206-93992206 on Build GRCh38
rs1004119243 Chr.7: 93992112-93992112 on Build GRCh38
rs937460322 Chr.7: 93992156-93992156 on Build GRCh38
rs893827180 Chr.7: 93991969-93991969 on Build GRCh38
rs897489189 Chr.7: 93992194-93992194 on Build GRCh38
rs544201377 Chr.7: 93991944-93991944 on Build GRCh38
rs751631473 Chr.7: 93992310-93992310 on Build GRCh38
rs538238292 Chr.7: 93991896-93991896 on Build GRCh38
rs766482825 Chr.7: 93992235-93992235 on Build GRCh38
rs928670238 Chr.7: 93992138-93992138 on Build GRCh38
rs1035222994 Chr.7: 93992139-93992139 on Build GRCh38
rs988078146 Chr.7: 93992037-93992037 on Build GRCh38
rs571862143 Chr.7: 93992250-93992250 on Build GRCh38
rs569436232 Chr.7: 93992040-93992040 on Build GRCh38

View Full Product Details