Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95495680-95496183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB4
Entrez Gene ID
51666
Gene Name
ankyrin repeat and SOCS box containing 4
Gene Aliases
ASB-4
UniGene
Hs.602765
Gene Chromosome Location
Chr.7: 95485439-95544738 on Build GRCh38
Transcripts
NM_016116.2 NM_145872.2 XM_017012303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365039 c.464G>A p.C155Y Chr.7: 95496034-95496034 on Build GRCh38
6605562 c.337G>A p.V113M Chr.7: 95495907-95495907 on Build GRCh38
6297640 c.358G>T p.V120L Chr.7: 95495928-95495928 on Build GRCh38
5884609 c.233G>A p.G78D Chr.7: 95495803-95495803 on Build GRCh38
4542608 c.321G>A p.G107G Chr.7: 95495891-95495891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757208378 Chr.7: 95496087-95496087 on Build GRCh38
rs754642300 Chr.7: 95496027-95496027 on Build GRCh38
rs777640563 Chr.7: 95495960-95495960 on Build GRCh38
rs371005051 Chr.7: 95495739-95495739 on Build GRCh38
rs763421536 Chr.7: 95496004-95496004 on Build GRCh38
rs547148697 Chr.7: 95495883-95495883 on Build GRCh38
rs760930817 Chr.7: 95495739-95495742 on Build GRCh38
rs755923726 Chr.7: 95496063-95496063 on Build GRCh38
rs533791334 Chr.7: 95496104-95496104 on Build GRCh38
rs771574376 Chr.7: 95495721-95495721 on Build GRCh38
rs761635646 Chr.7: 95495754-95495754 on Build GRCh38
rs773582169 Chr.7: 95495737-95495738 on Build GRCh38
rs1031541136 Chr.7: 95495799-95495799 on Build GRCh38
rs763927661 Chr.7: 95495738-95495739 on Build GRCh38
rs898160085 Chr.7: 95496073-95496073 on Build GRCh38
rs778508330 Chr.7: 95495808-95495808 on Build GRCh38
rs866367600 Chr.7: 95495756-95495756 on Build GRCh38
rs372932698 Chr.7: 95495961-95495961 on Build GRCh38
rs996212800 Chr.7: 95496129-95496129 on Build GRCh38
rs745565907 Chr.7: 95495717-95495717 on Build GRCh38
rs777589865 Chr.7: 95496080-95496080 on Build GRCh38
rs147091900 Chr.7: 95495981-95495981 on Build GRCh38
rs754585089 Chr.7: 95495940-95495940 on Build GRCh38
rs765595046 Chr.7: 95495910-95495910 on Build GRCh38
rs759469934 Chr.7: 95495873-95495873 on Build GRCh38
rs767015489 Chr.7: 95496030-95496030 on Build GRCh38
rs1047191434 Chr.7: 95495921-95495921 on Build GRCh38
rs749211498 Chr.7: 95496085-95496085 on Build GRCh38
rs747253499 Chr.7: 95495978-95495978 on Build GRCh38
rs761176544 Chr.7: 95495786-95495786 on Build GRCh38
rs779517506 Chr.7: 95495927-95495927 on Build GRCh38
rs373101368 Chr.7: 95495887-95495887 on Build GRCh38
rs757660898 Chr.7: 95495806-95495806 on Build GRCh38
rs147798502 Chr.7: 95495856-95495856 on Build GRCh38
rs879170749 Chr.7: 95495740-95495741 on Build GRCh38
rs764581157 Chr.7: 95495898-95495898 on Build GRCh38
rs759829404 Chr.7: 95495776-95495776 on Build GRCh38
rs773643647 Chr.7: 95496003-95496003 on Build GRCh38
rs757962480 Chr.7: 95495926-95495926 on Build GRCh38
rs780991883 Chr.7: 95495955-95495955 on Build GRCh38
rs768490367 Chr.7: 95495845-95495845 on Build GRCh38
rs779836593 Chr.7: 95495731-95495731 on Build GRCh38
rs776609639 Chr.7: 95495748-95495748 on Build GRCh38
rs527430788 Chr.7: 95495769-95495769 on Build GRCh38
rs767916695 Chr.7: 95495723-95495724 on Build GRCh38
rs556456967 Chr.7: 95495906-95495906 on Build GRCh38
rs777057452 Chr.7: 95495987-95495987 on Build GRCh38
rs758073916 Chr.7: 95495813-95495813 on Build GRCh38
rs754432763 Chr.7: 95495805-95495805 on Build GRCh38
rs749844932 Chr.7: 95495812-95495812 on Build GRCh38
rs141186023 Chr.7: 95495956-95495956 on Build GRCh38
rs751022766 Chr.7: 95496015-95496015 on Build GRCh38
rs774506865 Chr.7: 95495868-95495868 on Build GRCh38
rs114296603 Chr.7: 95495781-95495781 on Build GRCh38
rs539458950 Chr.7: 95495896-95495896 on Build GRCh38
rs544099785 Chr.7: 95495752-95495754 on Build GRCh38
rs376814974 Chr.7: 95495916-95495916 on Build GRCh38
rs762150698 Chr.7: 95495988-95495988 on Build GRCh38
rs752392005 Chr.7: 95496047-95496047 on Build GRCh38
rs114951738 Chr.7: 95496150-95496150 on Build GRCh38
rs1047954353 Chr.7: 95496009-95496009 on Build GRCh38
rs1038009951 Chr.7: 95496036-95496036 on Build GRCh38
rs200892706 Chr.7: 95495764-95495764 on Build GRCh38
rs758281370 Chr.7: 95495739-95495741 on Build GRCh38
rs370663407 Chr.7: 95495917-95495917 on Build GRCh38
rs751253040 Chr.7: 95495939-95495939 on Build GRCh38
rs765619625 Chr.7: 95495997-95495997 on Build GRCh38
rs898866612 Chr.7: 95496084-95496084 on Build GRCh38
rs765905478 Chr.7: 95496012-95496012 on Build GRCh38
rs756768517 Chr.7: 95495873-95495874 on Build GRCh38
rs768437307 Chr.7: 95495740-95495740 on Build GRCh38
rs570326706 Chr.7: 95495821-95495821 on Build GRCh38
rs762395018 Chr.7: 95495907-95495907 on Build GRCh38
rs769573705 Chr.7: 95495863-95495863 on Build GRCh38
rs753118507 Chr.7: 95495785-95495785 on Build GRCh38
rs140701495 Chr.7: 95495739-95495740 on Build GRCh38
rs202221772 Chr.7: 95495971-95495971 on Build GRCh38
rs746653735 Chr.7: 95495837-95495837 on Build GRCh38
rs879018852 Chr.7: 95495740-95495740 on Build GRCh38
rs746737726 Chr.7: 95495737-95495737 on Build GRCh38
rs756838332 Chr.7: 95495707-95495707 on Build GRCh38
rs35506436 Chr.7: 95495739-95495739 on Build GRCh38
rs748149300 Chr.7: 95495860-95495860 on Build GRCh38
rs764528090 Chr.7: 95495791-95495791 on Build GRCh38
rs142138079 Chr.7: 95495796-95495796 on Build GRCh38
rs778250717 Chr.7: 95495712-95495712 on Build GRCh38
rs780291223 Chr.7: 95496101-95496101 on Build GRCh38
rs942276713 Chr.7: 95496033-95496033 on Build GRCh38
rs566827194 Chr.7: 95495754-95495754 on Build GRCh38
rs746441233 Chr.7: 95495740-95495741 on Build GRCh38
rs1055916901 Chr.7: 95495847-95495847 on Build GRCh38

View Full Product Details