Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95810300-95810803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
UniGene
Hs.440364
Gene Chromosome Location
Chr.7: 95772506-96110322 on Build GRCh38
Transcripts
NM_001135556.1 NM_001135557.1 NM_001278421.1 NM_001278422.1 NM_004411.4 XM_011515861.1 XM_011515862.1 XM_011515863.2 XM_017011804.1 XM_017011805.1 XM_017011806.1 XM_017011807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884611 c.145G>A p.D49N Chr.7: 95810428-95810428 on Build GRCh38
5562852 c.209C>T p.P70L Chr.7: 95810492-95810492 on Build GRCh38
6650586 c.135C>T p.P45P Chr.7: 95810418-95810418 on Build GRCh38
160619 c.204C>T p.I68I Chr.7: 95810487-95810487 on Build GRCh38
5826439 c.160C>T p.R54C Chr.7: 95810443-95810443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558895871 Chr.7: 95810711-95810712 on Build GRCh38
rs397789374 Chr.7: 95810695-95810696 on Build GRCh38
rs760856913 Chr.7: 95810498-95810498 on Build GRCh38
rs772941982 Chr.7: 95810489-95810489 on Build GRCh38
rs930958508 Chr.7: 95810708-95810708 on Build GRCh38
rs116148088 Chr.7: 95810754-95810754 on Build GRCh38
rs757671459 Chr.7: 95810419-95810419 on Build GRCh38
rs746321086 Chr.7: 95810345-95810345 on Build GRCh38
rs577002460 Chr.7: 95810367-95810367 on Build GRCh38
rs886743576 Chr.7: 95810744-95810744 on Build GRCh38
rs917531688 Chr.7: 95810654-95810654 on Build GRCh38
rs764900772 Chr.7: 95810371-95810371 on Build GRCh38
rs774923117 Chr.7: 95810348-95810348 on Build GRCh38
rs771573932 Chr.7: 95810347-95810347 on Build GRCh38
rs963808908 Chr.7: 95810519-95810519 on Build GRCh38
rs895549165 Chr.7: 95810706-95810706 on Build GRCh38
rs375639863 Chr.7: 95810380-95810380 on Build GRCh38
rs772623334 Chr.7: 95810428-95810428 on Build GRCh38
rs780509560 Chr.7: 95810443-95810443 on Build GRCh38
rs184726667 Chr.7: 95810427-95810427 on Build GRCh38
rs758740767 Chr.7: 95810551-95810551 on Build GRCh38
rs778186339 Chr.7: 95810412-95810412 on Build GRCh38
rs532789204 Chr.7: 95810716-95810716 on Build GRCh38
rs544620187 Chr.7: 95810433-95810433 on Build GRCh38
rs78122113 Chr.7: 95810499-95810499 on Build GRCh38
rs926885544 Chr.7: 95810659-95810659 on Build GRCh38
rs779181893 Chr.7: 95810546-95810546 on Build GRCh38
rs547349982 Chr.7: 95810580-95810580 on Build GRCh38
rs752893858 Chr.7: 95810400-95810400 on Build GRCh38
rs938130623 Chr.7: 95810676-95810676 on Build GRCh38
rs1056675215 Chr.7: 95810686-95810686 on Build GRCh38
rs1047162525 Chr.7: 95810743-95810743 on Build GRCh38
rs776377255 Chr.7: 95810457-95810457 on Build GRCh38
rs757363632 Chr.7: 95810516-95810516 on Build GRCh38
rs567029171 Chr.7: 95810684-95810684 on Build GRCh38
rs776126904 Chr.7: 95810362-95810362 on Build GRCh38
rs749417050 Chr.7: 95810418-95810418 on Build GRCh38
rs762840556 Chr.7: 95810491-95810491 on Build GRCh38
rs200845264 Chr.7: 95810392-95810392 on Build GRCh38
rs756229765 Chr.7: 95810406-95810406 on Build GRCh38
rs946713877 Chr.7: 95810606-95810606 on Build GRCh38
rs371217689 Chr.7: 95810469-95810469 on Build GRCh38
rs191170660 Chr.7: 95810361-95810361 on Build GRCh38
rs35308603 Chr.7: 95810694-95810695 on Build GRCh38
rs201811117 Chr.7: 95810694-95810694 on Build GRCh38
rs768106213 Chr.7: 95810453-95810453 on Build GRCh38
rs76250658 Chr.7: 95810423-95810423 on Build GRCh38
rs1048147907 Chr.7: 95810720-95810720 on Build GRCh38
rs149830072 Chr.7: 95810470-95810470 on Build GRCh38
rs754094624 Chr.7: 95810514-95810514 on Build GRCh38
rs979667058 Chr.7: 95810651-95810651 on Build GRCh38
rs113047449 Chr.7: 95810344-95810344 on Build GRCh38
rs565417352 Chr.7: 95810425-95810425 on Build GRCh38
rs780562849 Chr.7: 95810552-95810552 on Build GRCh38
rs949820790 Chr.7: 95810734-95810734 on Build GRCh38
rs201786441 Chr.7: 95810555-95810555 on Build GRCh38
rs377495263 Chr.7: 95810444-95810444 on Build GRCh38
rs988086591 Chr.7: 95810581-95810581 on Build GRCh38
rs144180774 Chr.7: 95810676-95810676 on Build GRCh38
rs1013914335 Chr.7: 95810397-95810397 on Build GRCh38
rs375116771 Chr.7: 95810493-95810493 on Build GRCh38
rs540109367 Chr.7: 95810332-95810332 on Build GRCh38
rs913945343 Chr.7: 95810605-95810605 on Build GRCh38
rs918268296 Chr.7: 95810696-95810696 on Build GRCh38

View Full Product Details