Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95869770-95870259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
UniGene
Hs.440364
Gene Chromosome Location
Chr.7: 95772506-96110322 on Build GRCh38
Transcripts
NM_001135556.1 NM_001135557.1 NM_001278421.1 NM_001278422.1 NM_004411.4 XM_011515861.1 XM_011515862.1 XM_011515863.2 XM_017011804.1 XM_017011805.1 XM_017011806.1 XM_017011807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400874 c.426-1G>A p.? Chr.7: 95869882-95869882 on Build GRCh38
160618 c.499T>G p.S167A Chr.7: 95869956-95869956 on Build GRCh38
7429574 c.503A>C p.K168T Chr.7: 95869960-95869960 on Build GRCh38
1093373 c.456G>T p.K152N Chr.7: 95869913-95869913 on Build GRCh38
7400875 c.315-1G>A p.? Chr.7: 95869882-95869882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777564616 Chr.7: 95870033-95870033 on Build GRCh38
rs1040786176 Chr.7: 95870154-95870154 on Build GRCh38
rs3757697 Chr.7: 95869922-95869922 on Build GRCh38
rs916159604 Chr.7: 95869825-95869825 on Build GRCh38
rs144939564 Chr.7: 95869985-95869985 on Build GRCh38
rs1053717908 Chr.7: 95870176-95870176 on Build GRCh38
rs535934899 Chr.7: 95870087-95870087 on Build GRCh38
rs369627655 Chr.7: 95869984-95869984 on Build GRCh38
rs750057492 Chr.7: 95869904-95869904 on Build GRCh38
rs777283494 Chr.7: 95870004-95870004 on Build GRCh38
rs760284766 Chr.7: 95870092-95870093 on Build GRCh38
rs759734484 Chr.7: 95869841-95869841 on Build GRCh38
rs745998715 Chr.7: 95870035-95870035 on Build GRCh38
rs769093121 Chr.7: 95870001-95870001 on Build GRCh38
rs754748364 Chr.7: 95869925-95869925 on Build GRCh38
rs1045965292 Chr.7: 95870108-95870108 on Build GRCh38
rs1019737725 Chr.7: 95869840-95869840 on Build GRCh38
rs371885884 Chr.7: 95870016-95870016 on Build GRCh38
rs1023735396 Chr.7: 95870003-95870003 on Build GRCh38
rs999880172 Chr.7: 95869848-95869848 on Build GRCh38
rs762351980 Chr.7: 95870006-95870006 on Build GRCh38
rs888200544 Chr.7: 95870170-95870170 on Build GRCh38
rs932158504 Chr.7: 95870090-95870090 on Build GRCh38
rs191899121 Chr.7: 95869828-95869828 on Build GRCh38
rs779248114 Chr.7: 95870124-95870124 on Build GRCh38
rs74419990 Chr.7: 95870026-95870026 on Build GRCh38
rs267601651 Chr.7: 95869931-95869931 on Build GRCh38
rs201114371 Chr.7: 95869905-95869905 on Build GRCh38
rs142483580 Chr.7: 95869929-95869929 on Build GRCh38
rs760942472 Chr.7: 95869874-95869874 on Build GRCh38
rs749534280 Chr.7: 95869851-95869852 on Build GRCh38
rs552010203 Chr.7: 95870015-95870015 on Build GRCh38
rs748035438 Chr.7: 95869983-95869983 on Build GRCh38
rs912058258 Chr.7: 95870115-95870115 on Build GRCh38
rs370302761 Chr.7: 95870017-95870017 on Build GRCh38
rs957572417 Chr.7: 95870049-95870049 on Build GRCh38
rs775544140 Chr.7: 95869987-95869987 on Build GRCh38
rs752639824 Chr.7: 95870030-95870030 on Build GRCh38
rs552599263 Chr.7: 95870132-95870132 on Build GRCh38
rs79162063 Chr.7: 95869871-95869871 on Build GRCh38
rs34435934 Chr.7: 95869968-95869969 on Build GRCh38
rs1039569718 Chr.7: 95870185-95870185 on Build GRCh38
rs768432721 Chr.7: 95870099-95870099 on Build GRCh38
rs999177340 Chr.7: 95870169-95870169 on Build GRCh38
rs374468238 Chr.7: 95870010-95870010 on Build GRCh38
rs764775393 Chr.7: 95869895-95869895 on Build GRCh38
rs146559643 Chr.7: 95869911-95869911 on Build GRCh38
rs369400455 Chr.7: 95870028-95870028 on Build GRCh38
rs755268981 Chr.7: 95869945-95869945 on Build GRCh38
rs761440788 Chr.7: 95869887-95869887 on Build GRCh38
rs755162354 Chr.7: 95870011-95870011 on Build GRCh38
rs950369366 Chr.7: 95870101-95870101 on Build GRCh38
rs373047379 Chr.7: 95870046-95870046 on Build GRCh38
rs761170308 Chr.7: 95869997-95869997 on Build GRCh38
rs940682062 Chr.7: 95870044-95870044 on Build GRCh38
rs754373412 Chr.7: 95869879-95869879 on Build GRCh38
rs756098858 Chr.7: 95870032-95870032 on Build GRCh38
rs774286184 Chr.7: 95870168-95870168 on Build GRCh38
rs765827683 Chr.7: 95870009-95870009 on Build GRCh38
rs774584206 Chr.7: 95869832-95869832 on Build GRCh38
rs557109521 Chr.7: 95870045-95870045 on Build GRCh38
rs1018082393 Chr.7: 95869942-95869942 on Build GRCh38
rs772404533 Chr.7: 95869986-95869986 on Build GRCh38
rs377010996 Chr.7: 95869878-95869878 on Build GRCh38
rs906176957 Chr.7: 95869872-95869872 on Build GRCh38
rs184039866 Chr.7: 95870216-95870216 on Build GRCh38

View Full Product Details