Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97020310-97020821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX5
Entrez Gene ID
1749
Gene Name
distal-less homeobox 5
Gene Aliases
SHFM1D
UniGene
Hs.99348
Gene Chromosome Location
Chr.7: 97020390-97024831 on Build GRCh38
Transcripts
NM_005221.5 XM_005250185.3 XM_017011803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250206 c.819_820GC>AA p.P274T Chr.7: 97020786-97020787 on Build GRCh38
3642615 c.814C>T p.P272S Chr.7: 97020792-97020792 on Build GRCh38
7426811 c.830T>C p.L277S Chr.7: 97020776-97020776 on Build GRCh38
3642616 c.806C>T p.S269F Chr.7: 97020800-97020800 on Build GRCh38
5408335 c.822G>A p.P274P Chr.7: 97020784-97020784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779237377 Chr.7: 97020742-97020742 on Build GRCh38
rs763609484 Chr.7: 97020703-97020703 on Build GRCh38
rs397692728 Chr.7: 97020579-97020580 on Build GRCh38
rs750016914 Chr.7: 97020719-97020719 on Build GRCh38
rs929373566 Chr.7: 97020767-97020767 on Build GRCh38
rs1045102452 Chr.7: 97020594-97020594 on Build GRCh38
rs751332130 Chr.7: 97020785-97020785 on Build GRCh38
rs35646168 Chr.7: 97020355-97020355 on Build GRCh38
rs928010283 Chr.7: 97020602-97020602 on Build GRCh38
rs977110173 Chr.7: 97020648-97020648 on Build GRCh38
rs547131001 Chr.7: 97020744-97020744 on Build GRCh38
rs963873117 Chr.7: 97020625-97020625 on Build GRCh38
rs1207732 Chr.7: 97020356-97020356 on Build GRCh38
rs926170797 Chr.7: 97020389-97020389 on Build GRCh38
rs138650319 Chr.7: 97020771-97020771 on Build GRCh38
rs576971046 Chr.7: 97020620-97020620 on Build GRCh38
rs747524363 Chr.7: 97020759-97020759 on Build GRCh38
rs562464172 Chr.7: 97020651-97020651 on Build GRCh38
rs1027944435 Chr.7: 97020363-97020363 on Build GRCh38
rs764890966 Chr.7: 97020717-97020717 on Build GRCh38
rs991751279 Chr.7: 97020684-97020684 on Build GRCh38
rs761160497 Chr.7: 97020700-97020700 on Build GRCh38
rs869079594 Chr.7: 97020354-97020354 on Build GRCh38
rs948001862 Chr.7: 97020587-97020587 on Build GRCh38
rs977166129 Chr.7: 97020449-97020449 on Build GRCh38
rs990468095 Chr.7: 97020576-97020576 on Build GRCh38
rs541457727 Chr.7: 97020674-97020674 on Build GRCh38
rs527440729 Chr.7: 97020720-97020720 on Build GRCh38
rs1018622775 Chr.7: 97020394-97020394 on Build GRCh38
rs1007157810 Chr.7: 97020680-97020680 on Build GRCh38
rs5886001 Chr.7: 97020357-97020357 on Build GRCh38
rs762498461 Chr.7: 97020782-97020782 on Build GRCh38
rs776445505 Chr.7: 97020699-97020700 on Build GRCh38
rs761431045 Chr.7: 97020779-97020779 on Build GRCh38
rs751602574 Chr.7: 97020727-97020727 on Build GRCh38
rs3214218 Chr.7: 97020572-97020573 on Build GRCh38
rs564721565 Chr.7: 97020385-97020385 on Build GRCh38
rs889381870 Chr.7: 97020677-97020677 on Build GRCh38
rs575908695 Chr.7: 97020653-97020653 on Build GRCh38
rs765735593 Chr.7: 97020718-97020719 on Build GRCh38
rs757262628 Chr.7: 97020734-97020734 on Build GRCh38
rs776056313 Chr.7: 97020707-97020707 on Build GRCh38
rs772981631 Chr.7: 97020781-97020781 on Build GRCh38
rs950215549 Chr.7: 97020535-97020535 on Build GRCh38
rs554983217 Chr.7: 97020357-97020357 on Build GRCh38
rs202152755 Chr.7: 97020354-97020357 on Build GRCh38
rs534291396 Chr.7: 97020503-97020503 on Build GRCh38
rs3028342 Chr.7: 97020351-97020354 on Build GRCh38
rs967055712 Chr.7: 97020638-97020638 on Build GRCh38
rs574872037 Chr.7: 97020480-97020480 on Build GRCh38
rs866381684 Chr.7: 97020675-97020675 on Build GRCh38
rs778003408 Chr.7: 97020728-97020728 on Build GRCh38
rs1007190310 Chr.7: 97020375-97020375 on Build GRCh38
rs752650484 Chr.7: 97020797-97020797 on Build GRCh38
rs915832816 Chr.7: 97020584-97020584 on Build GRCh38
rs924425162 Chr.7: 97020486-97020486 on Build GRCh38
rs77382615 Chr.7: 97020572-97020572 on Build GRCh38
rs563982917 Chr.7: 97020791-97020791 on Build GRCh38
rs1046985054 Chr.7: 97020605-97020605 on Build GRCh38
rs759221585 Chr.7: 97020712-97020713 on Build GRCh38
rs775776434 Chr.7: 97020747-97020747 on Build GRCh38
rs998353912 Chr.7: 97020626-97020626 on Build GRCh38
rs553730476 Chr.7: 97020511-97020511 on Build GRCh38
rs532759180 Chr.7: 97020798-97020798 on Build GRCh38
rs200468189 Chr.7: 97020715-97020715 on Build GRCh38
rs1045776322 Chr.7: 97020546-97020546 on Build GRCh38
rs953010518 Chr.7: 97020365-97020365 on Build GRCh38
rs766080014 Chr.7: 97020721-97020721 on Build GRCh38
rs965787602 Chr.7: 97020443-97020443 on Build GRCh38
rs765748732 Chr.7: 97020694-97020695 on Build GRCh38
rs545645231 Chr.7: 97020649-97020649 on Build GRCh38
rs939405084 Chr.7: 97020614-97020614 on Build GRCh38
rs772549093 Chr.7: 97020746-97020746 on Build GRCh38
rs887777137 Chr.7: 97020776-97020776 on Build GRCh38
rs199678276 Chr.7: 97020786-97020786 on Build GRCh38
rs749333020 Chr.7: 97020730-97020730 on Build GRCh38
rs550399986 Chr.7: 97020751-97020751 on Build GRCh38
rs571835755 Chr.7: 97020565-97020566 on Build GRCh38
rs888394234 Chr.7: 97020607-97020607 on Build GRCh38
rs1018564643 Chr.7: 97020623-97020623 on Build GRCh38
rs776137912 Chr.7: 97020778-97020778 on Build GRCh38

View Full Product Details