Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99408374-99408880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUD31
Entrez Gene ID
8896
Gene Name
BUD31 homolog
Gene Aliases
Cwc14, EDG-2, EDG2, G10, YCR063W, fSAP17
UniGene
Hs.380233
Gene Chromosome Location
Chr.7: 99408620-99419616 on Build GRCh38
Transcripts
XM_005250671.4 XM_005250674.3 NM_014891.6

Gene Symbol
PDAP1
Entrez Gene ID
11333
Gene Name
PDGFA associated protein 1
Gene Aliases
HASPP28, PAP, PAP1
UniGene
Hs.632296
Gene Chromosome Location
Chr.7: 99394673-99408682 on Build GRCh38
Transcripts
XM_005250671.4 XM_005250674.3 NM_014891.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5548037 c.13+8delG p.? Chr.7: 99408528-99408528 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780824260 Chr.7: 99408523-99408523 on Build GRCh38
rs114782359 Chr.7: 99408853-99408853 on Build GRCh38
rs994904529 Chr.7: 99408580-99408581 on Build GRCh38
rs1055025003 Chr.7: 99408647-99408647 on Build GRCh38
rs971637059 Chr.7: 99408630-99408630 on Build GRCh38
rs1054907806 Chr.7: 99408408-99408408 on Build GRCh38
rs28411391 Chr.7: 99408754-99408754 on Build GRCh38
rs902156401 Chr.7: 99408677-99408677 on Build GRCh38
rs926903434 Chr.7: 99408702-99408702 on Build GRCh38
rs938237243 Chr.7: 99408723-99408723 on Build GRCh38
rs753335575 Chr.7: 99408565-99408567 on Build GRCh38
rs1017052159 Chr.7: 99408604-99408604 on Build GRCh38
rs1036150367 Chr.7: 99408790-99408790 on Build GRCh38
rs927533097 Chr.7: 99408634-99408634 on Build GRCh38
rs1056523586 Chr.7: 99408551-99408551 on Build GRCh38
rs981639151 Chr.7: 99408632-99408632 on Build GRCh38
rs868721168 Chr.7: 99408794-99408794 on Build GRCh38
rs1018858703 Chr.7: 99408851-99408851 on Build GRCh38
rs373984450 Chr.7: 99408850-99408850 on Build GRCh38
rs897568641 Chr.7: 99408791-99408791 on Build GRCh38
rs750838605 Chr.7: 99408396-99408401 on Build GRCh38
rs1007502583 Chr.7: 99408842-99408842 on Build GRCh38
rs994517167 Chr.7: 99408797-99408797 on Build GRCh38
rs1044016594 Chr.7: 99408469-99408469 on Build GRCh38
rs746961922 Chr.7: 99408808-99408808 on Build GRCh38
rs1010606438 Chr.7: 99408441-99408441 on Build GRCh38
rs1007122275 Chr.7: 99408570-99408570 on Build GRCh38
rs1002329777 Chr.7: 99408489-99408489 on Build GRCh38
rs915037555 Chr.7: 99408651-99408651 on Build GRCh38
rs889054192 Chr.7: 99408838-99408838 on Build GRCh38
rs953382221 Chr.7: 99408616-99408616 on Build GRCh38
rs1029520448 Chr.7: 99408561-99408561 on Build GRCh38
rs1017139221 Chr.7: 99408756-99408756 on Build GRCh38
rs893534735 Chr.7: 99408428-99408428 on Build GRCh38
rs963114720 Chr.7: 99408611-99408611 on Build GRCh38
rs11537951 Chr.7: 99408620-99408620 on Build GRCh38
rs115544138 Chr.7: 99408614-99408614 on Build GRCh38
rs563901216 Chr.7: 99408465-99408465 on Build GRCh38
rs946694350 Chr.7: 99408419-99408419 on Build GRCh38
rs1056576965 Chr.7: 99408730-99408730 on Build GRCh38
rs905440365 Chr.7: 99408471-99408471 on Build GRCh38
rs1042084504 Chr.7: 99408671-99408671 on Build GRCh38
rs988253086 Chr.7: 99408637-99408637 on Build GRCh38
rs1004327063 Chr.7: 99408783-99408783 on Build GRCh38
rs374549266 Chr.7: 99408582-99408583 on Build GRCh38
rs902434151 Chr.7: 99408543-99408543 on Build GRCh38
rs540283597 Chr.7: 99408731-99408731 on Build GRCh38
rs918551473 Chr.7: 99408622-99408622 on Build GRCh38
rs946419261 Chr.7: 99408659-99408659 on Build GRCh38
rs895584326 Chr.7: 99408557-99408557 on Build GRCh38
rs779995727 Chr.7: 99408564-99408565 on Build GRCh38
rs1051036027 Chr.7: 99408729-99408729 on Build GRCh38
rs998068209 Chr.7: 99408558-99408558 on Build GRCh38
rs140677404 Chr.7: 99408857-99408857 on Build GRCh38
rs937555314 Chr.7: 99408641-99408641 on Build GRCh38
rs529278717 Chr.7: 99408805-99408805 on Build GRCh38
rs997833896 Chr.7: 99408682-99408682 on Build GRCh38
rs756428065 Chr.7: 99408657-99408665 on Build GRCh38
rs752408460 Chr.7: 99408564-99408564 on Build GRCh38

View Full Product Details