Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128677207-128677745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71F2
Entrez Gene ID
346653
Gene Name
family with sequence similarity 71 member F2
Gene Aliases
FAM137B
UniGene
Hs.445236
Gene Chromosome Location
Chr.7: 128671628-128702262 on Build GRCh38
Transcripts
NM_001012454.5 NM_001128926.3 NM_001290254.1 NM_001290255.1 XM_011516128.2 XM_011516129.2 XM_011516130.2 XM_011516131.2 XM_011516132.2 XM_011516134.2 XM_011516135.2 XM_011516136.2 XM_017012147.1 XM_017012149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2768964 c.525G>A p.Q175Q Chr.7: 128677723-128677723 on Build GRCh38
5494874 c.448A>T p.I150F Chr.7: 128677646-128677646 on Build GRCh38
3949915 c.522G>T p.L174L Chr.7: 128677720-128677720 on Build GRCh38
4004288 c.406C>T p.R136W Chr.7: 128677604-128677604 on Build GRCh38
3633033 c.493C>T p.Q165* Chr.7: 128677691-128677691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201240369 Chr.7: 128677582-128677582 on Build GRCh38
rs149261565 Chr.7: 128677434-128677434 on Build GRCh38
rs762191267 Chr.7: 128677532-128677532 on Build GRCh38
rs1024888234 Chr.7: 128677476-128677476 on Build GRCh38
rs553880583 Chr.7: 128677667-128677667 on Build GRCh38
rs752098817 Chr.7: 128677618-128677618 on Build GRCh38
rs905360754 Chr.7: 128677364-128677364 on Build GRCh38
rs764712714 Chr.7: 128677548-128677548 on Build GRCh38
rs992589481 Chr.7: 128677660-128677660 on Build GRCh38
rs531576408 Chr.7: 128677614-128677614 on Build GRCh38
rs910308253 Chr.7: 128677277-128677277 on Build GRCh38
rs928959162 Chr.7: 128677309-128677309 on Build GRCh38
rs780040728 Chr.7: 128677586-128677586 on Build GRCh38
rs745872836 Chr.7: 128677528-128677528 on Build GRCh38
rs181109862 Chr.7: 128677599-128677599 on Build GRCh38
rs746847099 Chr.7: 128677528-128677532 on Build GRCh38
rs769910752 Chr.7: 128677583-128677583 on Build GRCh38
rs776048661 Chr.7: 128677529-128677529 on Build GRCh38
rs954751399 Chr.7: 128677424-128677424 on Build GRCh38
rs750686783 Chr.7: 128677515-128677516 on Build GRCh38
rs764781019 Chr.7: 128677616-128677616 on Build GRCh38
rs777677949 Chr.7: 128677525-128677525 on Build GRCh38
rs918944597 Chr.7: 128677289-128677289 on Build GRCh38
rs757118745 Chr.7: 128677578-128677578 on Build GRCh38
rs537680903 Chr.7: 128677245-128677245 on Build GRCh38
rs368780631 Chr.7: 128677521-128677521 on Build GRCh38
rs200938500 Chr.7: 128677640-128677640 on Build GRCh38
rs576794223 Chr.7: 128677490-128677490 on Build GRCh38
rs762556424 Chr.7: 128677635-128677635 on Build GRCh38
rs1014236879 Chr.7: 128677645-128677645 on Build GRCh38
rs916916903 Chr.7: 128677515-128677515 on Build GRCh38
rs185046903 Chr.7: 128677436-128677436 on Build GRCh38
rs936794444 Chr.7: 128677533-128677533 on Build GRCh38
rs766585684 Chr.7: 128677379-128677379 on Build GRCh38
rs571067980 Chr.7: 128677662-128677662 on Build GRCh38
rs763750831 Chr.7: 128677717-128677717 on Build GRCh38
rs6467208 Chr.7: 128677351-128677351 on Build GRCh38
rs774632019 Chr.7: 128677531-128677531 on Build GRCh38
rs941818353 Chr.7: 128677279-128677279 on Build GRCh38
rs6467210 Chr.7: 128677604-128677604 on Build GRCh38
rs747210660 Chr.7: 128677525-128677525 on Build GRCh38
rs560186199 Chr.7: 128677504-128677505 on Build GRCh38
rs769291032 Chr.7: 128677694-128677694 on Build GRCh38
rs577340396 Chr.7: 128677335-128677335 on Build GRCh38
rs753494341 Chr.7: 128677535-128677535 on Build GRCh38
rs777788075 Chr.7: 128677570-128677570 on Build GRCh38
rs762308334 Chr.7: 128677708-128677708 on Build GRCh38
rs10674238 Chr.7: 128677520-128677521 on Build GRCh38
rs532657795 Chr.7: 128677546-128677546 on Build GRCh38
rs1053892099 Chr.7: 128677542-128677542 on Build GRCh38
rs749291100 Chr.7: 128677655-128677655 on Build GRCh38
rs768787544 Chr.7: 128677666-128677666 on Build GRCh38
rs377562102 Chr.7: 128677704-128677704 on Build GRCh38
rs918975712 Chr.7: 128677466-128677466 on Build GRCh38
rs770884216 Chr.7: 128677537-128677539 on Build GRCh38
rs752647419 Chr.7: 128677552-128677552 on Build GRCh38
rs1009961262 Chr.7: 128677387-128677387 on Build GRCh38
rs748714129 Chr.7: 128677672-128677672 on Build GRCh38
rs1052231589 Chr.7: 128677592-128677592 on Build GRCh38
rs781197315 Chr.7: 128677650-128677650 on Build GRCh38
rs372599605 Chr.7: 128677653-128677653 on Build GRCh38
rs898082540 Chr.7: 128677294-128677294 on Build GRCh38
rs1045305460 Chr.7: 128677519-128677519 on Build GRCh38
rs758314723 Chr.7: 128677556-128677556 on Build GRCh38
rs950429948 Chr.7: 128677478-128677478 on Build GRCh38
rs549567409 Chr.7: 128677651-128677651 on Build GRCh38
rs755374158 Chr.7: 128677417-128677417 on Build GRCh38
rs764490027 Chr.7: 128677530-128677530 on Build GRCh38
rs776255301 Chr.7: 128677613-128677613 on Build GRCh38
rs538444441 Chr.7: 128677663-128677663 on Build GRCh38
rs749783143 Chr.7: 128677593-128677593 on Build GRCh38
rs757102292 Chr.7: 128677648-128677648 on Build GRCh38
rs745551542 Chr.7: 128677575-128677575 on Build GRCh38
rs1005618805 Chr.7: 128677303-128677303 on Build GRCh38
rs772423422 Chr.7: 128677682-128677682 on Build GRCh38
rs896705514 Chr.7: 128677344-128677344 on Build GRCh38
rs971073999 Chr.7: 128677484-128677484 on Build GRCh38
rs773614211 Chr.7: 128677685-128677685 on Build GRCh38
rs949709417 Chr.7: 128677517-128677517 on Build GRCh38
rs143509068 Chr.7: 128677435-128677435 on Build GRCh38
rs751358688 Chr.7: 128677573-128677573 on Build GRCh38
rs189850912 Chr.7: 128677500-128677500 on Build GRCh38
rs369042232 Chr.7: 128677283-128677283 on Build GRCh38
rs913661005 Chr.7: 128677563-128677563 on Build GRCh38
rs892515978 Chr.7: 128677554-128677554 on Build GRCh38
rs6467209 Chr.7: 128677389-128677389 on Build GRCh38
rs777021802 Chr.7: 128677526-128677526 on Build GRCh38
rs986328259 Chr.7: 128677250-128677250 on Build GRCh38
rs567199544 Chr.7: 128677229-128677229 on Build GRCh38
rs765976815 Chr.7: 128677534-128677534 on Build GRCh38
rs993723820 Chr.7: 128677376-128677376 on Build GRCh38
rs992863725 Chr.7: 128677483-128677483 on Build GRCh38
rs767678655 Chr.7: 128677514-128677515 on Build GRCh38
rs1007467333 Chr.7: 128677433-128677433 on Build GRCh38
rs138918252 Chr.7: 128677639-128677639 on Build GRCh38
rs1045675551 Chr.7: 128677363-128677363 on Build GRCh38
rs186458943 Chr.7: 128677605-128677605 on Build GRCh38
rs532060264 Chr.7: 128677517-128677518 on Build GRCh38
rs1000976688 Chr.7: 128677371-128677371 on Build GRCh38
rs758424308 Chr.7: 128677519-128677520 on Build GRCh38
rs761122140 Chr.7: 128677690-128677690 on Build GRCh38
rs555905658 Chr.7: 128677282-128677282 on Build GRCh38
rs756080314 Chr.7: 128677516-128677517 on Build GRCh38
rs954587881 Chr.7: 128677235-128677235 on Build GRCh38
rs780929015 Chr.7: 128677541-128677542 on Build GRCh38
rs1046174394 Chr.7: 128677646-128677646 on Build GRCh38

View Full Product Details