Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129400153-129400652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL2
Entrez Gene ID
23382
Gene Name
adenosylhomocysteinase like 2
Gene Aliases
ADOHCYASE3, IRBIT2
UniGene
Hs.600789
Gene Chromosome Location
Chr.7: 129225014-129430211 on Build GRCh38
Transcripts
NM_001130720.2 NM_001130722.2 NM_001130723.2 NM_015328.3 XM_005250237.1 XM_011515987.2 XM_011515988.2 XM_017011904.1 XM_017011905.1 XM_017011906.1 XM_017011907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6458197 c.917_918+1delTGG p.? Chr.7: 129400383-129400385 on Build GRCh38
6596616 c.637G>A p.D213N Chr.7: 129400346-129400346 on Build GRCh38
3411577 c.566G>A p.C189Y Chr.7: 129400341-129400341 on Build GRCh38
4660477 c.524delT p.F176fs*19 Chr.7: 129400299-129400299 on Build GRCh38
2154622 c.824-2A>G p.? Chr.7: 129400288-129400288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028374297 Chr.7: 129400267-129400267 on Build GRCh38
rs925695885 Chr.7: 129400544-129400544 on Build GRCh38
rs375799335 Chr.7: 129400250-129400250 on Build GRCh38
rs117550824 Chr.7: 129400312-129400312 on Build GRCh38
rs775907121 Chr.7: 129400268-129400268 on Build GRCh38
rs1004271912 Chr.7: 129400346-129400346 on Build GRCh38
rs899782071 Chr.7: 129400259-129400259 on Build GRCh38
rs1008850681 Chr.7: 129400192-129400192 on Build GRCh38
rs762229398 Chr.7: 129400435-129400435 on Build GRCh38
rs777466409 Chr.7: 129400348-129400348 on Build GRCh38
rs571310434 Chr.7: 129400285-129400285 on Build GRCh38
rs776758767 Chr.7: 129400246-129400246 on Build GRCh38
rs776001082 Chr.7: 129400417-129400417 on Build GRCh38
rs891501471 Chr.7: 129400565-129400565 on Build GRCh38
rs866355965 Chr.7: 129400194-129400194 on Build GRCh38
rs774646261 Chr.7: 129400433-129400433 on Build GRCh38
rs891717335 Chr.7: 129400182-129400182 on Build GRCh38
rs573704028 Chr.7: 129400581-129400581 on Build GRCh38
rs200049707 Chr.7: 129400425-129400425 on Build GRCh38
rs370421807 Chr.7: 129400393-129400393 on Build GRCh38
rs979825337 Chr.7: 129400538-129400538 on Build GRCh38
rs879107931 Chr.7: 129400203-129400203 on Build GRCh38
rs367651937 Chr.7: 129400408-129400408 on Build GRCh38
rs1053191596 Chr.7: 129400560-129400560 on Build GRCh38
rs770527352 Chr.7: 129400256-129400257 on Build GRCh38
rs996871875 Chr.7: 129400263-129400263 on Build GRCh38
rs78414944 Chr.7: 129400572-129400572 on Build GRCh38
rs932490005 Chr.7: 129400275-129400275 on Build GRCh38
rs758331503 Chr.7: 129400345-129400345 on Build GRCh38
rs906495532 Chr.7: 129400404-129400404 on Build GRCh38
rs144881015 Chr.7: 129400318-129400318 on Build GRCh38
rs866617749 Chr.7: 129400238-129400238 on Build GRCh38
rs989765165 Chr.7: 129400507-129400507 on Build GRCh38
rs763408622 Chr.7: 129400269-129400269 on Build GRCh38
rs1033920769 Chr.7: 129400422-129400422 on Build GRCh38
rs1022855932 Chr.7: 129400509-129400509 on Build GRCh38
rs776348538 Chr.7: 129400284-129400285 on Build GRCh38
rs779163039 Chr.7: 129400190-129400190 on Build GRCh38
rs935648244 Chr.7: 129400557-129400557 on Build GRCh38
rs1040318677 Chr.7: 129400597-129400597 on Build GRCh38
rs770298182 Chr.7: 129400260-129400260 on Build GRCh38
rs374395207 Chr.7: 129400406-129400406 on Build GRCh38
rs377182906 Chr.7: 129400240-129400240 on Build GRCh38
rs943709330 Chr.7: 129400239-129400239 on Build GRCh38
rs746810785 Chr.7: 129400368-129400368 on Build GRCh38
rs769176172 Chr.7: 129400431-129400431 on Build GRCh38
rs924402943 Chr.7: 129400562-129400562 on Build GRCh38
rs1040965271 Chr.7: 129400255-129400255 on Build GRCh38
rs917409648 Chr.7: 129400456-129400456 on Build GRCh38
rs200412891 Chr.7: 129400295-129400295 on Build GRCh38
rs977102094 Chr.7: 129400550-129400550 on Build GRCh38
rs771199865 Chr.7: 129400378-129400378 on Build GRCh38
rs556226587 Chr.7: 129400284-129400284 on Build GRCh38
rs1015697422 Chr.7: 129400387-129400387 on Build GRCh38
rs754482082 Chr.7: 129400292-129400292 on Build GRCh38
rs17552411 Chr.7: 129400177-129400177 on Build GRCh38
rs753461146 Chr.7: 129400291-129400291 on Build GRCh38
rs959912448 Chr.7: 129400466-129400466 on Build GRCh38
rs572035291 Chr.7: 129400382-129400382 on Build GRCh38
rs752880786 Chr.7: 129400403-129400403 on Build GRCh38
rs775413182 Chr.7: 129400543-129400543 on Build GRCh38
rs138763581 Chr.7: 129400379-129400379 on Build GRCh38
rs191105300 Chr.7: 129400277-129400277 on Build GRCh38
rs759598800 Chr.7: 129400246-129400247 on Build GRCh38
rs770897195 Chr.7: 129400527-129400527 on Build GRCh38

View Full Product Details