Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129403088-129403585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL2
Entrez Gene ID
23382
Gene Name
adenosylhomocysteinase like 2
Gene Aliases
ADOHCYASE3, IRBIT2
UniGene
Hs.600789
Gene Chromosome Location
Chr.7: 129225014-129430211 on Build GRCh38
Transcripts
NM_001130720.2 NM_001130722.2 NM_001130723.2 NM_015328.3 XM_005250237.1 XM_011515987.2 XM_011515988.2 XM_017011904.1 XM_017011905.1 XM_017011906.1 XM_017011907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4359671 c.754G>A p.V252I Chr.7: 129403457-129403457 on Build GRCh38
2769746 c.997G>A p.V333I Chr.7: 129403457-129403457 on Build GRCh38
6176185 c.693G>C p.G231G Chr.7: 129403396-129403396 on Build GRCh38
7285977 c.713A>G p.H238R Chr.7: 129403482-129403482 on Build GRCh38
5628348 c.757G>A p.E253K Chr.7: 129403460-129403460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372863954 Chr.7: 129403345-129403345 on Build GRCh38
rs190243017 Chr.7: 129403236-129403236 on Build GRCh38
rs956544942 Chr.7: 129403153-129403153 on Build GRCh38
rs977501262 Chr.7: 129403165-129403165 on Build GRCh38
rs537396397 Chr.7: 129403134-129403134 on Build GRCh38
rs910799467 Chr.7: 129403265-129403265 on Build GRCh38
rs986565254 Chr.7: 129403205-129403205 on Build GRCh38
rs902374379 Chr.7: 129403549-129403549 on Build GRCh38
rs566650083 Chr.7: 129403454-129403454 on Build GRCh38
rs111443622 Chr.7: 129403529-129403529 on Build GRCh38
rs565458540 Chr.7: 129403462-129403462 on Build GRCh38
rs923263861 Chr.7: 129403171-129403171 on Build GRCh38
rs571576020 Chr.7: 129403177-129403177 on Build GRCh38
rs755218460 Chr.7: 129403155-129403155 on Build GRCh38
rs752216290 Chr.7: 129403365-129403365 on Build GRCh38
rs963600605 Chr.7: 129403278-129403278 on Build GRCh38
rs756307302 Chr.7: 129403433-129403433 on Build GRCh38
rs372445200 Chr.7: 129403503-129403503 on Build GRCh38
rs370189414 Chr.7: 129403281-129403281 on Build GRCh38
rs903810858 Chr.7: 129403118-129403118 on Build GRCh38
rs781422539 Chr.7: 129403413-129403413 on Build GRCh38
rs1000639013 Chr.7: 129403120-129403120 on Build GRCh38
rs766146866 Chr.7: 129403330-129403330 on Build GRCh38
rs759136648 Chr.7: 129403342-129403342 on Build GRCh38
rs752809890 Chr.7: 129403473-129403473 on Build GRCh38
rs558338381 Chr.7: 129403133-129403133 on Build GRCh38
rs746042375 Chr.7: 129403416-129403416 on Build GRCh38
rs576566297 Chr.7: 129403393-129403393 on Build GRCh38
rs919391540 Chr.7: 129403359-129403359 on Build GRCh38
rs376109589 Chr.7: 129403372-129403372 on Build GRCh38
rs546349293 Chr.7: 129403257-129403257 on Build GRCh38
rs751323577 Chr.7: 129403396-129403396 on Build GRCh38
rs750365342 Chr.7: 129403305-129403305 on Build GRCh38
rs141705758 Chr.7: 129403474-129403474 on Build GRCh38
rs111915343 Chr.7: 129403156-129403156 on Build GRCh38
rs536824641 Chr.7: 129403296-129403296 on Build GRCh38
rs572598779 Chr.7: 129403262-129403262 on Build GRCh38
rs375465332 Chr.7: 129403508-129403508 on Build GRCh38
rs368385003 Chr.7: 129403481-129403481 on Build GRCh38
rs780038461 Chr.7: 129403446-129403446 on Build GRCh38
rs1030869868 Chr.7: 129403162-129403162 on Build GRCh38
rs758498494 Chr.7: 129403470-129403470 on Build GRCh38
rs539049951 Chr.7: 129403198-129403198 on Build GRCh38
rs776357973 Chr.7: 129403338-129403338 on Build GRCh38
rs540653855 Chr.7: 129403523-129403523 on Build GRCh38
rs756978852 Chr.7: 129403401-129403401 on Build GRCh38
rs929028385 Chr.7: 129403358-129403358 on Build GRCh38
rs202238607 Chr.7: 129403457-129403457 on Build GRCh38

View Full Product Details