Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129439893-129440388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP2
Entrez Gene ID
57464
Gene Name
striatin interacting protein 2
Gene Aliases
FAM40B, FAR11B
UniGene
Hs.489988
Gene Chromosome Location
Chr.7: 129434433-129488399 on Build GRCh38
Transcripts
NM_001134336.1 NM_020704.2 XM_006716069.3 XM_011516435.1 XM_017012470.1 XM_017012472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871808 c.179G>A p.G60E Chr.7: 129440071-129440071 on Build GRCh38
1085458 c.132C>T p.G44G Chr.7: 129440024-129440024 on Build GRCh38
5501622 c.160G>C p.E54Q Chr.7: 129440052-129440052 on Build GRCh38
5499733 c.130G>T p.G44C Chr.7: 129440022-129440022 on Build GRCh38
5499732 c.130G>T p.G44C Chr.7: 129440022-129440022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746855917 Chr.7: 129440052-129440052 on Build GRCh38
rs748990113 Chr.7: 129440023-129440023 on Build GRCh38
rs775395019 Chr.7: 129440250-129440250 on Build GRCh38
rs770125907 Chr.7: 129440067-129440067 on Build GRCh38
rs776533288 Chr.7: 129440056-129440056 on Build GRCh38
rs62479488 Chr.7: 129440160-129440160 on Build GRCh38
rs111256650 Chr.7: 129440317-129440317 on Build GRCh38
rs142815895 Chr.7: 129440318-129440318 on Build GRCh38
rs779778578 Chr.7: 129440019-129440019 on Build GRCh38
rs1006947888 Chr.7: 129440267-129440267 on Build GRCh38
rs879278029 Chr.7: 129440146-129440146 on Build GRCh38
rs562714648 Chr.7: 129440014-129440014 on Build GRCh38
rs987457681 Chr.7: 129440162-129440162 on Build GRCh38
rs79040720 Chr.7: 129440276-129440276 on Build GRCh38
rs750784182 Chr.7: 129440112-129440112 on Build GRCh38
rs754595296 Chr.7: 129440027-129440027 on Build GRCh38
rs762360308 Chr.7: 129440096-129440096 on Build GRCh38
rs754685257 Chr.7: 129440116-129440116 on Build GRCh38
rs1031407303 Chr.7: 129440080-129440080 on Build GRCh38
rs778482790 Chr.7: 129440124-129440124 on Build GRCh38
rs752210670 Chr.7: 129440131-129440131 on Build GRCh38
rs767812903 Chr.7: 129440106-129440106 on Build GRCh38
rs757810145 Chr.7: 129440143-129440143 on Build GRCh38
rs141063717 Chr.7: 129440077-129440077 on Build GRCh38
rs779192222 Chr.7: 129440048-129440049 on Build GRCh38
rs891210483 Chr.7: 129440186-129440186 on Build GRCh38
rs759902300 Chr.7: 129440063-129440063 on Build GRCh38
rs746275880 Chr.7: 129439991-129439991 on Build GRCh38
rs764691340 Chr.7: 129440078-129440078 on Build GRCh38
rs775724444 Chr.7: 129440070-129440070 on Build GRCh38
rs974256079 Chr.7: 129440252-129440253 on Build GRCh38
rs751956854 Chr.7: 129439982-129439982 on Build GRCh38
rs1050954842 Chr.7: 129440169-129440169 on Build GRCh38
rs979730076 Chr.7: 129439954-129439954 on Build GRCh38
rs942804926 Chr.7: 129440192-129440192 on Build GRCh38
rs151084809 Chr.7: 129440046-129440046 on Build GRCh38
rs929695569 Chr.7: 129440290-129440290 on Build GRCh38
rs374038661 Chr.7: 129439939-129439939 on Build GRCh38
rs781502397 Chr.7: 129439929-129439929 on Build GRCh38
rs751962026 Chr.7: 129440082-129440082 on Build GRCh38
rs61744175 Chr.7: 129440040-129440040 on Build GRCh38
rs911484879 Chr.7: 129440170-129440170 on Build GRCh38
rs367830416 Chr.7: 129440037-129440037 on Build GRCh38
rs778464419 Chr.7: 129440029-129440029 on Build GRCh38
rs371488277 Chr.7: 129440079-129440079 on Build GRCh38
rs971159407 Chr.7: 129439951-129439951 on Build GRCh38

View Full Product Details