Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135164711-135165212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
UniGene
Hs.643113
Gene Chromosome Location
Chr.7: 135092303-135170826 on Build GRCh38
Transcripts
NM_001305630.1 XM_017012589.1 XM_017012590.1 XM_017012591.1 XM_017012592.1 XM_017012593.1 XM_017012594.1 XM_017012595.1 NM_018295.4

Gene Symbol
TMEM140
Entrez Gene ID
55281
Gene Name
transmembrane protein 140
Gene Aliases
-
UniGene
Hs.521213
Gene Chromosome Location
Chr.7: 135148014-135166215 on Build GRCh38
Transcripts
NM_001305630.1 XM_017012589.1 XM_017012590.1 XM_017012591.1 XM_017012592.1 XM_017012593.1 XM_017012594.1 XM_017012595.1 NM_018295.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5772559 c.383C>G p.A128G Chr.7: 135164824-135164824 on Build GRCh38
1448237 c.388G>A p.G130S Chr.7: 135164829-135164829 on Build GRCh38
5782168 c.491T>C p.L164S Chr.7: 135164932-135164932 on Build GRCh38
5503320 c.330G>T p.E110D Chr.7: 135164771-135164771 on Build GRCh38
5676280 c.542A>C p.K181T Chr.7: 135164983-135164983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368957588 Chr.7: 135165036-135165036 on Build GRCh38
rs772997979 Chr.7: 135164972-135164972 on Build GRCh38
rs765977165 Chr.7: 135164978-135164978 on Build GRCh38
rs945624202 Chr.7: 135165060-135165060 on Build GRCh38
rs773882052 Chr.7: 135164959-135164960 on Build GRCh38
rs201469043 Chr.7: 135165016-135165016 on Build GRCh38
rs201284142 Chr.7: 135164910-135164910 on Build GRCh38
rs939012718 Chr.7: 135164794-135164794 on Build GRCh38
rs977821233 Chr.7: 135164943-135164943 on Build GRCh38
rs557742539 Chr.7: 135165096-135165096 on Build GRCh38
rs754366575 Chr.7: 135164985-135164985 on Build GRCh38
rs1049962814 Chr.7: 135165101-135165101 on Build GRCh38
rs762557232 Chr.7: 135164754-135164754 on Build GRCh38
rs368287615 Chr.7: 135165011-135165011 on Build GRCh38
rs758716406 Chr.7: 135165126-135165126 on Build GRCh38
rs755157807 Chr.7: 135165023-135165023 on Build GRCh38
rs150916088 Chr.7: 135164829-135164829 on Build GRCh38
rs762258689 Chr.7: 135164995-135165001 on Build GRCh38
rs918638772 Chr.7: 135165082-135165082 on Build GRCh38
rs1037174973 Chr.7: 135165064-135165064 on Build GRCh38
rs292502 Chr.7: 135164776-135164776 on Build GRCh38
rs764481357 Chr.7: 135164909-135164909 on Build GRCh38
rs181583668 Chr.7: 135165085-135165085 on Build GRCh38
rs867512582 Chr.7: 135164944-135164944 on Build GRCh38
rs762677084 Chr.7: 135164769-135164770 on Build GRCh38
rs746806447 Chr.7: 135165034-135165034 on Build GRCh38
rs370468370 Chr.7: 135164920-135164920 on Build GRCh38
rs771090349 Chr.7: 135164734-135164734 on Build GRCh38
rs756322177 Chr.7: 135164932-135164932 on Build GRCh38
rs923581131 Chr.7: 135164770-135164770 on Build GRCh38
rs370150432 Chr.7: 135164733-135164733 on Build GRCh38
rs755249689 Chr.7: 135164890-135164890 on Build GRCh38
rs375824469 Chr.7: 135164913-135164913 on Build GRCh38
rs1046374424 Chr.7: 135165098-135165098 on Build GRCh38
rs372768615 Chr.7: 135165018-135165018 on Build GRCh38
rs11972838 Chr.7: 135165148-135165148 on Build GRCh38
rs145425395 Chr.7: 135165095-135165095 on Build GRCh38
rs199612642 Chr.7: 135164882-135164882 on Build GRCh38
rs555560730 Chr.7: 135165074-135165074 on Build GRCh38
rs914808725 Chr.7: 135164779-135164779 on Build GRCh38
rs1048380947 Chr.7: 135165086-135165086 on Build GRCh38
rs374720338 Chr.7: 135165015-135165015 on Build GRCh38
rs2288237 Chr.7: 135164825-135164825 on Build GRCh38
rs926402398 Chr.7: 135164784-135164784 on Build GRCh38
rs754564466 Chr.7: 135164777-135164777 on Build GRCh38
rs559385983 Chr.7: 135164782-135164782 on Build GRCh38
rs767560540 Chr.7: 135164771-135164771 on Build GRCh38
rs774234090 Chr.7: 135164761-135164761 on Build GRCh38
rs775925118 Chr.7: 135164828-135164828 on Build GRCh38
rs770162914 Chr.7: 135164757-135164757 on Build GRCh38
rs750434845 Chr.7: 135164773-135164773 on Build GRCh38
rs974032437 Chr.7: 135165103-135165103 on Build GRCh38
rs148014348 Chr.7: 135164736-135164736 on Build GRCh38
rs151231260 Chr.7: 135164879-135164879 on Build GRCh38
rs375469494 Chr.7: 135165014-135165014 on Build GRCh38
rs374108885 Chr.7: 135164987-135164987 on Build GRCh38
rs377098001 Chr.7: 135164774-135164774 on Build GRCh38
rs765869187 Chr.7: 135164775-135164775 on Build GRCh38
rs753056412 Chr.7: 135164802-135164802 on Build GRCh38
rs369115443 Chr.7: 135164906-135164906 on Build GRCh38
rs1027403769 Chr.7: 135164884-135164884 on Build GRCh38
rs138263623 Chr.7: 135164881-135164881 on Build GRCh38
rs577709401 Chr.7: 135165147-135165147 on Build GRCh38
rs376210094 Chr.7: 135165001-135165001 on Build GRCh38
rs766237152 Chr.7: 135164871-135164871 on Build GRCh38
rs749390569 Chr.7: 135165019-135165019 on Build GRCh38
rs370885555 Chr.7: 135165007-135165007 on Build GRCh38
rs535270843 Chr.7: 135165045-135165045 on Build GRCh38
rs747593448 Chr.7: 135164813-135164813 on Build GRCh38
rs759022678 Chr.7: 135164995-135164995 on Build GRCh38
rs769019760 Chr.7: 135164967-135164967 on Build GRCh38
rs760609346 Chr.7: 135164851-135164851 on Build GRCh38
rs961995152 Chr.7: 135164767-135164767 on Build GRCh38
rs758818338 Chr.7: 135164805-135164805 on Build GRCh38
rs776136485 Chr.7: 135164873-135164873 on Build GRCh38
rs770078926 Chr.7: 135164826-135164826 on Build GRCh38
rs76023601 Chr.7: 135164938-135164938 on Build GRCh38
rs776490873 Chr.7: 135164990-135164990 on Build GRCh38
rs778229565 Chr.7: 135164806-135164806 on Build GRCh38
rs891176888 Chr.7: 135165120-135165120 on Build GRCh38
rs186256537 Chr.7: 135164778-135164778 on Build GRCh38
rs958047538 Chr.7: 135164981-135164981 on Build GRCh38
rs761708082 Chr.7: 135164769-135164769 on Build GRCh38
rs781284369 Chr.7: 135164821-135164821 on Build GRCh38
rs772909323 Chr.7: 135164835-135164835 on Build GRCh38
rs551320074 Chr.7: 135165008-135165008 on Build GRCh38
rs543835844 Chr.7: 135164755-135164755 on Build GRCh38
rs575323338 Chr.7: 135164749-135164749 on Build GRCh38
rs560752107 Chr.7: 135165156-135165156 on Build GRCh38
rs142088210 Chr.7: 135164752-135164752 on Build GRCh38
rs749478586 Chr.7: 135164966-135164966 on Build GRCh38
rs767980880 Chr.7: 135164960-135164968 on Build GRCh38
rs771374160 Chr.7: 135164819-135164819 on Build GRCh38
rs745763383 Chr.7: 135165041-135165041 on Build GRCh38
rs533977284 Chr.7: 135165031-135165031 on Build GRCh38
rs1024808998 Chr.7: 135164914-135164914 on Build GRCh38
rs190331994 Chr.7: 135165035-135165035 on Build GRCh38
rs755357266 Chr.7: 135164999-135164999 on Build GRCh38

View Full Product Details