Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39011565-39012063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM9
Entrez Gene ID
8754
Gene Name
ADAM metallopeptidase domain 9
Gene Aliases
CORD9, MCMP, MDC9, Mltng
UniGene
Hs.591852
Gene Chromosome Location
Chr.8: 38996771-39105261 on Build GRCh38
Transcripts
NM_003816.2 NR_027638.1 NR_027639.1 NR_027878.1 XM_011544682.2 XM_017013942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1457000 c.220G>T p.G74* Chr.8: 39011682-39011682 on Build GRCh38
3367349 c.247A>T p.R83W Chr.8: 39011709-39011709 on Build GRCh38
7145194 c.225A>G p.K75K Chr.8: 39011687-39011687 on Build GRCh38
4659410 c.235A>G p.I79V Chr.8: 39011697-39011697 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34091706 Chr.8: 39011814-39011815 on Build GRCh38
rs751707329 Chr.8: 39011970-39011970 on Build GRCh38
rs1011131922 Chr.8: 39011610-39011610 on Build GRCh38
rs778552330 Chr.8: 39011768-39011768 on Build GRCh38
rs1006575427 Chr.8: 39011993-39011993 on Build GRCh38
rs560310834 Chr.8: 39011981-39011981 on Build GRCh38
rs757633652 Chr.8: 39012035-39012035 on Build GRCh38
rs755006543 Chr.8: 39011622-39011622 on Build GRCh38
rs948007443 Chr.8: 39011773-39011773 on Build GRCh38
rs750570805 Chr.8: 39011693-39011693 on Build GRCh38
rs756200261 Chr.8: 39011917-39011917 on Build GRCh38
rs780833500 Chr.8: 39011625-39011625 on Build GRCh38
rs755884246 Chr.8: 39011729-39011729 on Build GRCh38
rs1009180685 Chr.8: 39011916-39011916 on Build GRCh38
rs758419449 Chr.8: 39011696-39011696 on Build GRCh38
rs906588804 Chr.8: 39011690-39011690 on Build GRCh38
rs895135730 Chr.8: 39011841-39011841 on Build GRCh38
rs1029253898 Chr.8: 39011637-39011637 on Build GRCh38
rs765400681 Chr.8: 39011692-39011692 on Build GRCh38
rs981899412 Chr.8: 39011790-39011790 on Build GRCh38
rs929787927 Chr.8: 39011623-39011623 on Build GRCh38
rs1050455144 Chr.8: 39011902-39011902 on Build GRCh38
rs748992321 Chr.8: 39011652-39011652 on Build GRCh38
rs777588861 Chr.8: 39011735-39011735 on Build GRCh38
rs752601927 Chr.8: 39011724-39011724 on Build GRCh38
rs1057518208 Chr.8: 39011657-39011657 on Build GRCh38
rs370116751 Chr.8: 39011921-39011921 on Build GRCh38
rs562802367 Chr.8: 39011588-39011588 on Build GRCh38
rs769510020 Chr.8: 39011635-39011635 on Build GRCh38
rs1022541689 Chr.8: 39011892-39011892 on Build GRCh38
rs983964382 Chr.8: 39011739-39011739 on Build GRCh38
rs752792636 Chr.8: 39011831-39011831 on Build GRCh38
rs1029379626 Chr.8: 39011952-39011952 on Build GRCh38
rs747916148 Chr.8: 39011626-39011626 on Build GRCh38
rs770629380 Chr.8: 39011761-39011761 on Build GRCh38
rs767418357 Chr.8: 39011698-39011698 on Build GRCh38
rs889247394 Chr.8: 39011661-39011661 on Build GRCh38
rs1009064949 Chr.8: 39011681-39011681 on Build GRCh38
rs372808162 Chr.8: 39011751-39011751 on Build GRCh38
rs1011610370 Chr.8: 39011868-39011868 on Build GRCh38
rs746885401 Chr.8: 39011612-39011612 on Build GRCh38
rs775867251 Chr.8: 39011667-39011667 on Build GRCh38
rs532976167 Chr.8: 39011923-39011923 on Build GRCh38
rs893819296 Chr.8: 39011592-39011592 on Build GRCh38
rs576667792 Chr.8: 39011589-39011589 on Build GRCh38
rs757592897 Chr.8: 39011691-39011691 on Build GRCh38
rs545687342 Chr.8: 39011638-39011638 on Build GRCh38
rs191369261 Chr.8: 39011912-39011912 on Build GRCh38
rs796870440 Chr.8: 39011916-39011918 on Build GRCh38
rs770400649 Chr.8: 39011656-39011656 on Build GRCh38
rs376543947 Chr.8: 39011836-39011836 on Build GRCh38
rs866415221 Chr.8: 39011951-39011951 on Build GRCh38
rs927813079 Chr.8: 39011792-39011792 on Build GRCh38
rs61753672 Chr.8: 39011688-39011688 on Build GRCh38
rs374171809 Chr.8: 39011757-39011757 on Build GRCh38
rs893772038 Chr.8: 39011834-39011834 on Build GRCh38
rs529062251 Chr.8: 39012007-39012007 on Build GRCh38
rs546695553 Chr.8: 39011962-39011962 on Build GRCh38
rs761058825 Chr.8: 39011670-39011670 on Build GRCh38
rs759947958 Chr.8: 39011660-39011660 on Build GRCh38
rs773918513 Chr.8: 39011658-39011658 on Build GRCh38
rs768011201 Chr.8: 39011662-39011662 on Build GRCh38
rs1055153828 Chr.8: 39011793-39011793 on Build GRCh38
rs935409048 Chr.8: 39011812-39011812 on Build GRCh38
rs772805392 Chr.8: 39011648-39011648 on Build GRCh38

View Full Product Details