Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39013470-39014013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM9
Entrez Gene ID
8754
Gene Name
ADAM metallopeptidase domain 9
Gene Aliases
CORD9, MCMP, MDC9, Mltng
UniGene
Hs.591852
Gene Chromosome Location
Chr.8: 38996771-39105261 on Build GRCh38
Transcripts
NM_003816.2 NR_027638.1 NR_027639.1 NR_027878.1 XM_011544682.2 XM_017013942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4779835 c.257A>G p.D86G Chr.8: 39013967-39013967 on Build GRCh38
4659411 c.260delT p.L88fs*16 Chr.8: 39013970-39013970 on Build GRCh38
6593750 c.255-1G>A p.? Chr.8: 39013964-39013964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs980338370 Chr.8: 39013569-39013569 on Build GRCh38
rs901632026 Chr.8: 39013814-39013814 on Build GRCh38
rs540187906 Chr.8: 39013929-39013929 on Build GRCh38
rs1043393113 Chr.8: 39013802-39013802 on Build GRCh38
rs764448169 Chr.8: 39013870-39013870 on Build GRCh38
rs766421346 Chr.8: 39013963-39013963 on Build GRCh38
rs964087195 Chr.8: 39013638-39013638 on Build GRCh38
rs769111658 Chr.8: 39013934-39013934 on Build GRCh38
rs186316131 Chr.8: 39013694-39013694 on Build GRCh38
rs75029712 Chr.8: 39013919-39013919 on Build GRCh38
rs992580371 Chr.8: 39013784-39013784 on Build GRCh38
rs943237690 Chr.8: 39013808-39013808 on Build GRCh38
rs370238974 Chr.8: 39013506-39013506 on Build GRCh38
rs745320048 Chr.8: 39013922-39013922 on Build GRCh38
rs756115024 Chr.8: 39013969-39013969 on Build GRCh38
rs970264176 Chr.8: 39013545-39013545 on Build GRCh38
rs997354012 Chr.8: 39013816-39013816 on Build GRCh38
rs776015736 Chr.8: 39013928-39013928 on Build GRCh38
rs763041780 Chr.8: 39013944-39013944 on Build GRCh38
rs752653261 Chr.8: 39013965-39013965 on Build GRCh38
rs62504420 Chr.8: 39013876-39013876 on Build GRCh38
rs960193214 Chr.8: 39013783-39013783 on Build GRCh38
rs1038976068 Chr.8: 39013810-39013810 on Build GRCh38
rs778560318 Chr.8: 39013607-39013607 on Build GRCh38
rs960244338 Chr.8: 39013595-39013595 on Build GRCh38
rs745421812 Chr.8: 39013646-39013646 on Build GRCh38
rs1006287529 Chr.8: 39013906-39013906 on Build GRCh38
rs997225222 Chr.8: 39013702-39013702 on Build GRCh38
rs1006168123 Chr.8: 39013505-39013507 on Build GRCh38
rs752599940 Chr.8: 39013946-39013946 on Build GRCh38
rs77781038 Chr.8: 39013918-39013918 on Build GRCh38
rs112177358 Chr.8: 39013717-39013717 on Build GRCh38
rs7840044 Chr.8: 39013806-39013806 on Build GRCh38
rs573652293 Chr.8: 39013507-39013507 on Build GRCh38
rs970046146 Chr.8: 39013927-39013927 on Build GRCh38
rs886514573 Chr.8: 39013899-39013899 on Build GRCh38
rs548354013 Chr.8: 39013609-39013609 on Build GRCh38
rs576090749 Chr.8: 39013760-39013760 on Build GRCh38
rs533862332 Chr.8: 39013907-39013907 on Build GRCh38
rs191016085 Chr.8: 39013942-39013942 on Build GRCh38
rs928768569 Chr.8: 39013585-39013585 on Build GRCh38
rs983016530 Chr.8: 39013763-39013763 on Build GRCh38
rs777199628 Chr.8: 39013941-39013941 on Build GRCh38
rs1008308290 Chr.8: 39013509-39013509 on Build GRCh38
rs1048058769 Chr.8: 39013856-39013856 on Build GRCh38
rs992279577 Chr.8: 39013611-39013611 on Build GRCh38
rs76099010 Chr.8: 39013943-39013943 on Build GRCh38
rs565082549 Chr.8: 39013822-39013822 on Build GRCh38
rs773303455 Chr.8: 39013948-39013948 on Build GRCh38
rs771512707 Chr.8: 39013925-39013925 on Build GRCh38
rs908741708 Chr.8: 39013772-39013772 on Build GRCh38
rs1027255375 Chr.8: 39013708-39013708 on Build GRCh38
rs181978584 Chr.8: 39013637-39013637 on Build GRCh38

View Full Product Details