Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39013960-39014503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM9
Entrez Gene ID
8754
Gene Name
ADAM metallopeptidase domain 9
Gene Aliases
CORD9, MCMP, MDC9, Mltng
UniGene
Hs.591852
Gene Chromosome Location
Chr.8: 38996771-39105261 on Build GRCh38
Transcripts
NM_003816.2 NR_027638.1 NR_027639.1 NR_027878.1 XM_011544682.2 XM_017013942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4779835 c.257A>G p.D86G Chr.8: 39013967-39013967 on Build GRCh38
750434 c.312C>G p.I104M Chr.8: 39014022-39014022 on Build GRCh38
4659411 c.260delT p.L88fs*16 Chr.8: 39013970-39013970 on Build GRCh38
6593750 c.255-1G>A p.? Chr.8: 39013964-39013964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771587211 Chr.8: 39014025-39014025 on Build GRCh38
rs759627385 Chr.8: 39014079-39014079 on Build GRCh38
rs550158378 Chr.8: 39014036-39014036 on Build GRCh38
rs138058399 Chr.8: 39014026-39014026 on Build GRCh38
rs1038864705 Chr.8: 39014365-39014365 on Build GRCh38
rs372386835 Chr.8: 39014069-39014069 on Build GRCh38
rs570001592 Chr.8: 39014111-39014111 on Build GRCh38
rs888443895 Chr.8: 39014124-39014124 on Build GRCh38
rs758053201 Chr.8: 39014122-39014122 on Build GRCh38
rs896045547 Chr.8: 39014468-39014468 on Build GRCh38
rs970242418 Chr.8: 39014211-39014211 on Build GRCh38
rs535255617 Chr.8: 39014197-39014197 on Build GRCh38
rs746733892 Chr.8: 39014379-39014379 on Build GRCh38
rs751497733 Chr.8: 39014280-39014282 on Build GRCh38
rs566260372 Chr.8: 39014153-39014153 on Build GRCh38
rs748717115 Chr.8: 39014138-39014138 on Build GRCh38
rs911782894 Chr.8: 39014320-39014320 on Build GRCh38
rs376641202 Chr.8: 39014294-39014294 on Build GRCh38
rs770307880 Chr.8: 39014038-39014038 on Build GRCh38
rs532633984 Chr.8: 39014131-39014131 on Build GRCh38
rs555269468 Chr.8: 39014198-39014198 on Build GRCh38
rs773519298 Chr.8: 39014442-39014442 on Build GRCh38
rs974424920 Chr.8: 39014362-39014362 on Build GRCh38
rs754537086 Chr.8: 39014132-39014132 on Build GRCh38
rs745528352 Chr.8: 39014016-39014016 on Build GRCh38
rs201485493 Chr.8: 39014003-39014003 on Build GRCh38
rs757052065 Chr.8: 39014002-39014002 on Build GRCh38
rs753572901 Chr.8: 39014001-39014001 on Build GRCh38
rs765200165 Chr.8: 39014135-39014149 on Build GRCh38
rs779598124 Chr.8: 39014127-39014127 on Build GRCh38
rs759432939 Chr.8: 39014126-39014127 on Build GRCh38
rs146980702 Chr.8: 39013990-39013990 on Build GRCh38
rs888447464 Chr.8: 39014451-39014451 on Build GRCh38
rs1023116366 Chr.8: 39014208-39014208 on Build GRCh38
rs143726709 Chr.8: 39014267-39014267 on Build GRCh38
rs763128480 Chr.8: 39014063-39014063 on Build GRCh38
rs1047540964 Chr.8: 39014113-39014113 on Build GRCh38
rs895817240 Chr.8: 39014480-39014480 on Build GRCh38
rs757922112 Chr.8: 39014132-39014133 on Build GRCh38
rs779347320 Chr.8: 39014194-39014194 on Build GRCh38
rs983788262 Chr.8: 39014431-39014431 on Build GRCh38
rs758709363 Chr.8: 39014358-39014358 on Build GRCh38
rs978527331 Chr.8: 39014213-39014213 on Build GRCh38
rs754060539 Chr.8: 39014169-39014169 on Build GRCh38
rs922955591 Chr.8: 39014388-39014388 on Build GRCh38
rs777287822 Chr.8: 39014033-39014033 on Build GRCh38
rs561769853 Chr.8: 39014068-39014068 on Build GRCh38
rs988699531 Chr.8: 39014297-39014297 on Build GRCh38
rs1010923120 Chr.8: 39014251-39014252 on Build GRCh38
rs1048469212 Chr.8: 39014445-39014445 on Build GRCh38
rs773464514 Chr.8: 39014049-39014049 on Build GRCh38
rs932760824 Chr.8: 39014395-39014395 on Build GRCh38
rs556122062 Chr.8: 39014457-39014457 on Build GRCh38
rs1037537599 Chr.8: 39014443-39014443 on Build GRCh38
rs763954582 Chr.8: 39014089-39014089 on Build GRCh38
rs191141761 Chr.8: 39014336-39014336 on Build GRCh38
rs907680582 Chr.8: 39014437-39014437 on Build GRCh38
rs747513455 Chr.8: 39014028-39014028 on Build GRCh38
rs1020421839 Chr.8: 39014252-39014252 on Build GRCh38
rs900319705 Chr.8: 39014383-39014383 on Build GRCh38
rs764988927 Chr.8: 39014115-39014115 on Build GRCh38
rs539344713 Chr.8: 39014461-39014461 on Build GRCh38
rs185252887 Chr.8: 39014137-39014137 on Build GRCh38
rs886293361 Chr.8: 39014117-39014117 on Build GRCh38
rs770166682 Chr.8: 39014145-39014145 on Build GRCh38
rs757543626 Chr.8: 39014173-39014173 on Build GRCh38
rs369082153 Chr.8: 39014470-39014470 on Build GRCh38
rs750059527 Chr.8: 39014121-39014121 on Build GRCh38
rs933230598 Chr.8: 39014412-39014412 on Build GRCh38
rs1015866039 Chr.8: 39014464-39014464 on Build GRCh38
rs769340668 Chr.8: 39014032-39014032 on Build GRCh38

View Full Product Details