Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39018765-39019275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM9
Entrez Gene ID
8754
Gene Name
ADAM metallopeptidase domain 9
Gene Aliases
CORD9, MCMP, MDC9, Mltng
UniGene
Hs.591852
Gene Chromosome Location
Chr.8: 38996771-39105261 on Build GRCh38
Transcripts
NM_003816.2 NR_027638.1 NR_027639.1 NR_027878.1 XM_011544682.2 XM_017013942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5583253 c.643G>A p.E215K Chr.8: 39018889-39018889 on Build GRCh38
1457002 c.659T>C p.V220A Chr.8: 39018905-39018905 on Build GRCh38
5563414 c.630G>C p.Q210H Chr.8: 39018876-39018876 on Build GRCh38
5564936 c.608G>C p.R203T Chr.8: 39018854-39018854 on Build GRCh38
6501309 c.614G>A p.R205K Chr.8: 39018860-39018860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138367650 Chr.8: 39018904-39018904 on Build GRCh38
rs776640071 Chr.8: 39018813-39018813 on Build GRCh38
rs569136200 Chr.8: 39018975-39018975 on Build GRCh38
rs188184271 Chr.8: 39018798-39018798 on Build GRCh38
rs745852661 Chr.8: 39018894-39018894 on Build GRCh38
rs181730840 Chr.8: 39019104-39019104 on Build GRCh38
rs200115954 Chr.8: 39018949-39018950 on Build GRCh38
rs941027470 Chr.8: 39019140-39019140 on Build GRCh38
rs966724749 Chr.8: 39019099-39019099 on Build GRCh38
rs771963841 Chr.8: 39018898-39018898 on Build GRCh38
rs775507590 Chr.8: 39018881-39018881 on Build GRCh38
rs200002472 Chr.8: 39019139-39019140 on Build GRCh38
rs754224538 Chr.8: 39019224-39019224 on Build GRCh38
rs184304860 Chr.8: 39018950-39018950 on Build GRCh38
rs890783079 Chr.8: 39018790-39018790 on Build GRCh38
rs372696732 Chr.8: 39018834-39018834 on Build GRCh38
rs200061745 Chr.8: 39018957-39018957 on Build GRCh38
rs1006939292 Chr.8: 39019179-39019179 on Build GRCh38
rs905141477 Chr.8: 39018972-39018972 on Build GRCh38
rs946948487 Chr.8: 39019030-39019030 on Build GRCh38
rs756633910 Chr.8: 39018935-39018935 on Build GRCh38
rs768293074 Chr.8: 39018925-39018925 on Build GRCh38
rs748106142 Chr.8: 39018826-39018826 on Build GRCh38
rs898444148 Chr.8: 39019183-39019183 on Build GRCh38
rs890805452 Chr.8: 39019127-39019127 on Build GRCh38
rs753146447 Chr.8: 39019214-39019214 on Build GRCh38
rs188978439 Chr.8: 39019055-39019055 on Build GRCh38
rs776149008 Chr.8: 39018932-39018932 on Build GRCh38
rs1025469787 Chr.8: 39018861-39018861 on Build GRCh38
rs748192481 Chr.8: 39018880-39018880 on Build GRCh38
rs386724609 Chr.8: 39018947-39018949 on Build GRCh38
rs991101909 Chr.8: 39018979-39018979 on Build GRCh38
rs751698128 Chr.8: 39018851-39018851 on Build GRCh38
rs929692826 Chr.8: 39019136-39019136 on Build GRCh38
rs6474519 Chr.8: 39018949-39018949 on Build GRCh38
rs766487146 Chr.8: 39019000-39019000 on Build GRCh38
rs1009670766 Chr.8: 39019043-39019043 on Build GRCh38
rs534074764 Chr.8: 39019180-39019180 on Build GRCh38
rs763505149 Chr.8: 39018807-39018807 on Build GRCh38
rs755286802 Chr.8: 39018857-39018857 on Build GRCh38
rs761272931 Chr.8: 39018938-39018938 on Build GRCh38
rs956754523 Chr.8: 39019040-39019040 on Build GRCh38
rs766814763 Chr.8: 39018839-39018839 on Build GRCh38
rs369963157 Chr.8: 39018859-39018859 on Build GRCh38
rs758712063 Chr.8: 39018835-39018835 on Build GRCh38
rs751895581 Chr.8: 39018958-39018958 on Build GRCh38
rs1025522076 Chr.8: 39019226-39019226 on Build GRCh38
rs764747978 Chr.8: 39018944-39018944 on Build GRCh38
rs910798366 Chr.8: 39019076-39019076 on Build GRCh38
rs775419284 Chr.8: 39018903-39018903 on Build GRCh38
rs951153960 Chr.8: 39019233-39019233 on Build GRCh38
rs1043899600 Chr.8: 39019048-39019048 on Build GRCh38
rs367824444 Chr.8: 39018822-39018822 on Build GRCh38
rs372304969 Chr.8: 39018960-39018960 on Build GRCh38
rs778948702 Chr.8: 39018892-39018892 on Build GRCh38
rs780532163 Chr.8: 39018950-39018960 on Build GRCh38
rs995414814 Chr.8: 39019217-39019217 on Build GRCh38
rs1046801401 Chr.8: 39019139-39019139 on Build GRCh38
rs553937288 Chr.8: 39019249-39019249 on Build GRCh38
rs199673299 Chr.8: 39018944-39018944 on Build GRCh38
rs900911390 Chr.8: 39018889-39018889 on Build GRCh38
rs5891051 Chr.8: 39019227-39019227 on Build GRCh38
rs935133146 Chr.8: 39019057-39019057 on Build GRCh38
rs573348452 Chr.8: 39018998-39018998 on Build GRCh38
rs190607349 Chr.8: 39018948-39018948 on Build GRCh38
rs6474518 Chr.8: 39018947-39018947 on Build GRCh38
rs968079296 Chr.8: 39019048-39019049 on Build GRCh38
rs538260048 Chr.8: 39019054-39019054 on Build GRCh38
rs929745090 Chr.8: 39018938-39018940 on Build GRCh38

View Full Product Details