Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 65602164-65602673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC1
Entrez Gene ID
55156
Gene Name
armadillo repeat containing 1
Gene Aliases
Arcp
UniGene
Hs.269542
Gene Chromosome Location
Chr.8: 65602456-65634217 on Build GRCh38
Transcripts
NM_001286702.1 NM_018120.5 XM_005251264.1 XM_017013605.1

SNP Detail

SNP ID Chromosome Location
rs1006758504 Chr.8: 65602333-65602333 on Build GRCh38
rs925014167 Chr.8: 65602419-65602419 on Build GRCh38
rs897260898 Chr.8: 65602255-65602255 on Build GRCh38
rs542906292 Chr.8: 65602341-65602341 on Build GRCh38
rs891517919 Chr.8: 65602231-65602231 on Build GRCh38
rs545912446 Chr.8: 65602470-65602470 on Build GRCh38
rs980941943 Chr.8: 65602431-65602431 on Build GRCh38
rs141051115 Chr.8: 65602380-65602380 on Build GRCh38
rs10957360 Chr.8: 65602271-65602271 on Build GRCh38
rs1010422815 Chr.8: 65602245-65602245 on Build GRCh38
rs918602855 Chr.8: 65602353-65602353 on Build GRCh38
rs34208962 Chr.8: 65602407-65602408 on Build GRCh38
rs58830657 Chr.8: 65602331-65602331 on Build GRCh38
rs780023057 Chr.8: 65602469-65602469 on Build GRCh38
rs1013422449 Chr.8: 65602622-65602622 on Build GRCh38
rs541323440 Chr.8: 65602553-65602553 on Build GRCh38
rs897293526 Chr.8: 65602536-65602536 on Build GRCh38
rs186290942 Chr.8: 65602619-65602619 on Build GRCh38
rs1048507349 Chr.8: 65602562-65602562 on Build GRCh38
rs1018107403 Chr.8: 65602334-65602334 on Build GRCh38
rs771938446 Chr.8: 65602425-65602425 on Build GRCh38
rs1034001277 Chr.8: 65602347-65602347 on Build GRCh38
rs970455348 Chr.8: 65602313-65602313 on Build GRCh38
rs925494287 Chr.8: 65602445-65602445 on Build GRCh38
rs541622125 Chr.8: 65602373-65602373 on Build GRCh38
rs945138791 Chr.8: 65602390-65602390 on Build GRCh38
rs563465716 Chr.8: 65602497-65602497 on Build GRCh38
rs189305244 Chr.8: 65602350-65602350 on Build GRCh38
rs571847434 Chr.8: 65602406-65602406 on Build GRCh38
rs1035672624 Chr.8: 65602580-65602580 on Build GRCh38
rs138126965 Chr.8: 65602370-65602370 on Build GRCh38
rs944245816 Chr.8: 65602486-65602486 on Build GRCh38
rs181727400 Chr.8: 65602577-65602577 on Build GRCh38
rs1020780485 Chr.8: 65602645-65602645 on Build GRCh38
rs995550824 Chr.8: 65602265-65602265 on Build GRCh38
rs376134133 Chr.8: 65602596-65602596 on Build GRCh38
rs1026994552 Chr.8: 65602302-65602302 on Build GRCh38
rs778698419 Chr.8: 65602382-65602382 on Build GRCh38
rs905887053 Chr.8: 65602568-65602568 on Build GRCh38
rs1029484788 Chr.8: 65602275-65602275 on Build GRCh38
rs34494846 Chr.8: 65602397-65602397 on Build GRCh38
rs1054541404 Chr.8: 65602198-65602198 on Build GRCh38
rs781629763 Chr.8: 65602195-65602195 on Build GRCh38
rs374231251 Chr.8: 65602267-65602267 on Build GRCh38
rs77152155 Chr.8: 65602545-65602545 on Build GRCh38
rs563723923 Chr.8: 65602534-65602534 on Build GRCh38

View Full Product Details