Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 65604060-65604594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC1
Entrez Gene ID
55156
Gene Name
armadillo repeat containing 1
Gene Aliases
Arcp
UniGene
Hs.269542
Gene Chromosome Location
Chr.8: 65602456-65634217 on Build GRCh38
Transcripts
NM_001286702.1 NM_018120.5 XM_005251264.1 XM_017013605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710996 c.663G>T p.L221F Chr.8: 65604580-65604580 on Build GRCh38
6477915 c.753G>T p.Q251H Chr.8: 65604490-65604490 on Build GRCh38
5624707 c.769C>G p.R257G Chr.8: 65604474-65604474 on Build GRCh38
5518840 c.809G>A p.S270N Chr.8: 65604434-65604434 on Build GRCh38
3650182 c.768C>T p.S256S Chr.8: 65604475-65604475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112262902 Chr.8: 65604220-65604220 on Build GRCh38
rs755302470 Chr.8: 65604474-65604474 on Build GRCh38
rs773589121 Chr.8: 65604354-65604354 on Build GRCh38
rs771277399 Chr.8: 65604535-65604535 on Build GRCh38
rs74618162 Chr.8: 65604427-65604427 on Build GRCh38
rs926025227 Chr.8: 65604306-65604306 on Build GRCh38
rs945807857 Chr.8: 65604196-65604196 on Build GRCh38
rs1025632319 Chr.8: 65604391-65604391 on Build GRCh38
rs752895613 Chr.8: 65604165-65604168 on Build GRCh38
rs767177622 Chr.8: 65604558-65604558 on Build GRCh38
rs751195581 Chr.8: 65604401-65604401 on Build GRCh38
rs1018076446 Chr.8: 65604324-65604324 on Build GRCh38
rs757591080 Chr.8: 65604465-65604465 on Build GRCh38
rs376698775 Chr.8: 65604381-65604381 on Build GRCh38
rs373766316 Chr.8: 65604278-65604278 on Build GRCh38
rs763968681 Chr.8: 65604466-65604466 on Build GRCh38
rs891483881 Chr.8: 65604170-65604170 on Build GRCh38
rs530545141 Chr.8: 65604158-65604158 on Build GRCh38
rs748211690 Chr.8: 65604481-65604481 on Build GRCh38
rs1024460738 Chr.8: 65604286-65604286 on Build GRCh38
rs747204678 Chr.8: 65604471-65604471 on Build GRCh38
rs1006645428 Chr.8: 65604292-65604292 on Build GRCh38
rs563957149 Chr.8: 65604468-65604468 on Build GRCh38
rs888223039 Chr.8: 65604523-65604523 on Build GRCh38
rs759248455 Chr.8: 65604421-65604421 on Build GRCh38
rs751496631 Chr.8: 65604274-65604274 on Build GRCh38
rs778919793 Chr.8: 65604469-65604469 on Build GRCh38
rs146747274 Chr.8: 65604503-65604503 on Build GRCh38
rs554927843 Chr.8: 65604277-65604277 on Build GRCh38
rs202230866 Chr.8: 65604532-65604532 on Build GRCh38
rs139310924 Chr.8: 65604467-65604467 on Build GRCh38
rs749860416 Chr.8: 65604484-65604484 on Build GRCh38
rs199552111 Chr.8: 65604426-65604426 on Build GRCh38
rs557545587 Chr.8: 65604114-65604114 on Build GRCh38
rs781374786 Chr.8: 65604480-65604480 on Build GRCh38
rs967514689 Chr.8: 65604300-65604300 on Build GRCh38
rs575855264 Chr.8: 65604513-65604513 on Build GRCh38
rs773610877 Chr.8: 65604482-65604482 on Build GRCh38
rs898396224 Chr.8: 65604358-65604358 on Build GRCh38
rs369417752 Chr.8: 65604270-65604270 on Build GRCh38
rs1005677095 Chr.8: 65604548-65604548 on Build GRCh38
rs770227460 Chr.8: 65604352-65604352 on Build GRCh38
rs993958365 Chr.8: 65604132-65604132 on Build GRCh38
rs199910042 Chr.8: 65604529-65604529 on Build GRCh38
rs546386774 Chr.8: 65604551-65604551 on Build GRCh38
rs879142486 Chr.8: 65604097-65604097 on Build GRCh38
rs774735268 Chr.8: 65604540-65604540 on Build GRCh38
rs1055469984 Chr.8: 65604561-65604561 on Build GRCh38
rs1040980770 Chr.8: 65604439-65604439 on Build GRCh38
rs765962440 Chr.8: 65604395-65604395 on Build GRCh38
rs775023934 Chr.8: 65604568-65604568 on Build GRCh38
rs141533227 Chr.8: 65604405-65604405 on Build GRCh38
rs375597098 Chr.8: 65604350-65604350 on Build GRCh38
rs573618954 Chr.8: 65604302-65604302 on Build GRCh38
rs3750184 Chr.8: 65604255-65604255 on Build GRCh38
rs1005234322 Chr.8: 65604272-65604272 on Build GRCh38
rs957656302 Chr.8: 65604335-65604337 on Build GRCh38
rs150868518 Chr.8: 65604461-65604461 on Build GRCh38

View Full Product Details