Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 65604480-65604980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC1
Entrez Gene ID
55156
Gene Name
armadillo repeat containing 1
Gene Aliases
Arcp
UniGene
Hs.269542
Gene Chromosome Location
Chr.8: 65602456-65634217 on Build GRCh38
Transcripts
NM_001286702.1 NM_018120.5 XM_005251264.1 XM_017013605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5710996 c.663G>T p.L221F Chr.8: 65604580-65604580 on Build GRCh38
6477915 c.753G>T p.Q251H Chr.8: 65604490-65604490 on Build GRCh38
3745637 c.695A>G p.E232G Chr.8: 65604548-65604548 on Build GRCh38
3745636 c.701A>G p.N234S Chr.8: 65604542-65604542 on Build GRCh38
107818 c.740C>T p.P247L Chr.8: 65604503-65604503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988329427 Chr.8: 65604927-65604927 on Build GRCh38
rs754861833 Chr.8: 65604777-65604777 on Build GRCh38
rs1050725837 Chr.8: 65604645-65604645 on Build GRCh38
rs894163678 Chr.8: 65604863-65604863 on Build GRCh38
rs529335872 Chr.8: 65604682-65604682 on Build GRCh38
rs771277399 Chr.8: 65604535-65604535 on Build GRCh38
rs967547192 Chr.8: 65604623-65604623 on Build GRCh38
rs765104254 Chr.8: 65604599-65604599 on Build GRCh38
rs188872769 Chr.8: 65604601-65604601 on Build GRCh38
rs138458929 Chr.8: 65604649-65604649 on Build GRCh38
rs536966475 Chr.8: 65604810-65604810 on Build GRCh38
rs533077855 Chr.8: 65604879-65604879 on Build GRCh38
rs1045342855 Chr.8: 65604931-65604931 on Build GRCh38
rs980879625 Chr.8: 65604867-65604867 on Build GRCh38
rs767177622 Chr.8: 65604558-65604558 on Build GRCh38
rs760107248 Chr.8: 65604573-65604573 on Build GRCh38
rs986612834 Chr.8: 65604678-65604678 on Build GRCh38
rs1017645047 Chr.8: 65604685-65604685 on Build GRCh38
rs201174215 Chr.8: 65604614-65604614 on Build GRCh38
rs573816843 Chr.8: 65604843-65604846 on Build GRCh38
rs115554303 Chr.8: 65604854-65604854 on Build GRCh38
rs1026023408 Chr.8: 65604684-65604684 on Build GRCh38
rs756139019 Chr.8: 65604635-65604635 on Build GRCh38
rs958486210 Chr.8: 65604900-65604900 on Build GRCh38
rs1039495097 Chr.8: 65604658-65604658 on Build GRCh38
rs779917112 Chr.8: 65604637-65604637 on Build GRCh38
rs540767388 Chr.8: 65604644-65604644 on Build GRCh38
rs748915269 Chr.8: 65604589-65604590 on Build GRCh38
rs888223039 Chr.8: 65604523-65604523 on Build GRCh38
rs945664323 Chr.8: 65604636-65604636 on Build GRCh38
rs550703598 Chr.8: 65604754-65604754 on Build GRCh38
rs146747274 Chr.8: 65604503-65604503 on Build GRCh38
rs202230866 Chr.8: 65604532-65604532 on Build GRCh38
rs781685916 Chr.8: 65604759-65604759 on Build GRCh38
rs757300643 Chr.8: 65604588-65604588 on Build GRCh38
rs575855264 Chr.8: 65604513-65604513 on Build GRCh38
rs757855581 Chr.8: 65604629-65604629 on Build GRCh38
rs753715249 Chr.8: 65604583-65604583 on Build GRCh38
rs914106270 Chr.8: 65604953-65604953 on Build GRCh38
rs1005677095 Chr.8: 65604548-65604548 on Build GRCh38
rs764034757 Chr.8: 65604574-65604574 on Build GRCh38
rs779678272 Chr.8: 65604585-65604586 on Build GRCh38
rs909671068 Chr.8: 65604826-65604826 on Build GRCh38
rs903463775 Chr.8: 65604956-65604956 on Build GRCh38
rs748204803 Chr.8: 65604628-65604628 on Build GRCh38
rs966018185 Chr.8: 65604732-65604732 on Build GRCh38
rs199910042 Chr.8: 65604529-65604529 on Build GRCh38
rs546386774 Chr.8: 65604551-65604551 on Build GRCh38
rs781572874 Chr.8: 65604626-65604626 on Build GRCh38
rs774735268 Chr.8: 65604540-65604540 on Build GRCh38
rs751254233 Chr.8: 65604646-65604646 on Build GRCh38
rs1013972464 Chr.8: 65604789-65604789 on Build GRCh38
rs1055469984 Chr.8: 65604561-65604561 on Build GRCh38
rs898352806 Chr.8: 65604665-65604665 on Build GRCh38
rs551139324 Chr.8: 65604892-65604892 on Build GRCh38
rs775023934 Chr.8: 65604568-65604568 on Build GRCh38
rs922528189 Chr.8: 65604767-65604767 on Build GRCh38

View Full Product Details