Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66677712-66678224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf44
Entrez Gene ID
56260
Gene Name
chromosome 8 open reading frame 44
Gene Aliases
-
UniGene
Hs.661238
Gene Chromosome Location
Chr.8: 66667552-66681149 on Build GRCh38
Transcripts
NM_019607.2 NM_001204173.1

Gene Symbol
C8orf44-SGK3
Entrez Gene ID
100533105
Gene Name
C8orf44-SGK3 readthrough
Gene Aliases
-
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66667552-66862022 on Build GRCh38
Transcripts
NM_019607.2 NM_001204173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2721103 c.70G>A p.G24R Chr.8: 66677778-66677778 on Build GRCh38
3834922 c.168A>C p.S56S Chr.8: 66677876-66677876 on Build GRCh38
3699142 c.130G>A p.V44I Chr.8: 66677838-66677838 on Build GRCh38
3901073 c.152C>T p.A51V Chr.8: 66677860-66677860 on Build GRCh38
6457872 c.115C>T p.R39W Chr.8: 66677823-66677823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986240680 Chr.8: 66678167-66678167 on Build GRCh38
rs1047700712 Chr.8: 66678142-66678142 on Build GRCh38
rs1002059388 Chr.8: 66678180-66678180 on Build GRCh38
rs894658485 Chr.8: 66678086-66678086 on Build GRCh38
rs755514202 Chr.8: 66677959-66677959 on Build GRCh38
rs917286333 Chr.8: 66678153-66678153 on Build GRCh38
rs746000809 Chr.8: 66677758-66677758 on Build GRCh38
rs750422103 Chr.8: 66677757-66677758 on Build GRCh38
rs920702070 Chr.8: 66678133-66678133 on Build GRCh38
rs143430246 Chr.8: 66678160-66678160 on Build GRCh38
rs377182380 Chr.8: 66677788-66677788 on Build GRCh38
rs954939863 Chr.8: 66678051-66678051 on Build GRCh38
rs568554290 Chr.8: 66677953-66677953 on Build GRCh38
rs773719511 Chr.8: 66677744-66677744 on Build GRCh38
rs767320717 Chr.8: 66677817-66677817 on Build GRCh38
rs757533517 Chr.8: 66678093-66678093 on Build GRCh38
rs758809503 Chr.8: 66677769-66677769 on Build GRCh38
rs201834734 Chr.8: 66677748-66677748 on Build GRCh38
rs769553702 Chr.8: 66677880-66677880 on Build GRCh38
rs755347706 Chr.8: 66677853-66677853 on Build GRCh38
rs957914945 Chr.8: 66677997-66677997 on Build GRCh38
rs183297940 Chr.8: 66677915-66677915 on Build GRCh38
rs1046303147 Chr.8: 66678170-66678170 on Build GRCh38
rs747567015 Chr.8: 66677850-66677850 on Build GRCh38
rs757438621 Chr.8: 66677819-66677819 on Build GRCh38
rs967394207 Chr.8: 66678102-66678102 on Build GRCh38
rs769150478 Chr.8: 66677778-66677778 on Build GRCh38
rs775244183 Chr.8: 66677917-66677917 on Build GRCh38
rs368997109 Chr.8: 66677774-66677774 on Build GRCh38
rs764233123 Chr.8: 66677745-66677745 on Build GRCh38
rs929408856 Chr.8: 66678140-66678140 on Build GRCh38
rs186267809 Chr.8: 66677740-66677740 on Build GRCh38
rs139248429 Chr.8: 66677776-66677776 on Build GRCh38
rs1002325148 Chr.8: 66677974-66677974 on Build GRCh38
rs558536617 Chr.8: 66678173-66678173 on Build GRCh38
rs1055936951 Chr.8: 66678030-66678030 on Build GRCh38
rs988776922 Chr.8: 66677767-66677767 on Build GRCh38
rs868626977 Chr.8: 66678192-66678192 on Build GRCh38
rs1047392233 Chr.8: 66677824-66677824 on Build GRCh38
rs758571831 Chr.8: 66677831-66677831 on Build GRCh38
rs760522794 Chr.8: 66677925-66677925 on Build GRCh38
rs148867765 Chr.8: 66677807-66677807 on Build GRCh38
rs546189778 Chr.8: 66677885-66677885 on Build GRCh38
rs3739317 Chr.8: 66677843-66677843 on Build GRCh38
rs548729011 Chr.8: 66677931-66677931 on Build GRCh38
rs944593591 Chr.8: 66677909-66677909 on Build GRCh38
rs762577335 Chr.8: 66677799-66677799 on Build GRCh38
rs914171990 Chr.8: 66677775-66677775 on Build GRCh38
rs1040281670 Chr.8: 66677910-66677910 on Build GRCh38
rs145557649 Chr.8: 66677798-66677798 on Build GRCh38
rs571129313 Chr.8: 66678112-66678112 on Build GRCh38
rs371977893 Chr.8: 66678178-66678178 on Build GRCh38
rs999298027 Chr.8: 66678102-66678102 on Build GRCh38
rs775510119 Chr.8: 66677820-66677820 on Build GRCh38
rs750457540 Chr.8: 66677827-66677827 on Build GRCh38
rs200113060 Chr.8: 66677818-66677818 on Build GRCh38
rs747829595 Chr.8: 66677788-66677788 on Build GRCh38
rs532301051 Chr.8: 66677812-66677812 on Build GRCh38
rs774105822 Chr.8: 66677896-66677896 on Build GRCh38
rs748677677 Chr.8: 66677889-66677889 on Build GRCh38
rs975954125 Chr.8: 66678182-66678182 on Build GRCh38
rs3739318 Chr.8: 66678011-66678011 on Build GRCh38
rs191530952 Chr.8: 66677787-66677787 on Build GRCh38
rs910690035 Chr.8: 66678091-66678091 on Build GRCh38
rs1024584086 Chr.8: 66678101-66678102 on Build GRCh38
rs781717034 Chr.8: 66677858-66677858 on Build GRCh38
rs751974342 Chr.8: 66677958-66677958 on Build GRCh38
rs1033097150 Chr.8: 66678156-66678156 on Build GRCh38
rs1020656848 Chr.8: 66678035-66678035 on Build GRCh38
rs780670614 Chr.8: 66677770-66677770 on Build GRCh38
rs1001962800 Chr.8: 66678028-66678028 on Build GRCh38
rs763770904 Chr.8: 66677750-66677750 on Build GRCh38
rs548490026 Chr.8: 66678190-66678190 on Build GRCh38
rs551195500 Chr.8: 66678056-66678056 on Build GRCh38
rs764021734 Chr.8: 66677747-66677747 on Build GRCh38
rs1024281284 Chr.8: 66677782-66677782 on Build GRCh38
rs762886637 Chr.8: 66677942-66677942 on Build GRCh38
rs770487157 Chr.8: 66677894-66677894 on Build GRCh38
rs781778066 Chr.8: 66677987-66677987 on Build GRCh38
rs148605675 Chr.8: 66677823-66677823 on Build GRCh38
rs887561892 Chr.8: 66677973-66677973 on Build GRCh38
rs10096544 Chr.8: 66677916-66677916 on Build GRCh38
rs765442327 Chr.8: 66677934-66677934 on Build GRCh38
rs750664285 Chr.8: 66677941-66677941 on Build GRCh38

View Full Product Details