Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17085028-17085576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU3
Entrez Gene ID
286097
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
UniGene
Hs.403594
Gene Chromosome Location
Chr.8: 17027238-17133069 on Build GRCh38
Transcripts
NM_181723.2 XM_005273477.2 XM_006716328.2 XM_006716329.2 XM_006716330.2 XM_006716333.3 XM_011544507.2 XM_011544508.2 XM_017013336.1 XM_017013337.1 XM_017013338.1 XM_017013339.1 XM_017013340.1 XM_017013341.1 XM_017013342.1 XM_017013343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603457 c.695-1G>A p.? Chr.8: 17085235-17085235 on Build GRCh38
6180451 c.729G>T p.M243I Chr.8: 17085270-17085270 on Build GRCh38
6180452 c.706G>T p.G236W Chr.8: 17085247-17085247 on Build GRCh38
454256 c.757G>A p.D253N Chr.8: 17085298-17085298 on Build GRCh38
750215 c.734A>T p.D245V Chr.8: 17085275-17085275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779957949 Chr.8: 17085362-17085365 on Build GRCh38
rs201647320 Chr.8: 17085334-17085334 on Build GRCh38
rs921121245 Chr.8: 17085111-17085111 on Build GRCh38
rs757828444 Chr.8: 17085207-17085207 on Build GRCh38
rs2285264 Chr.8: 17085328-17085328 on Build GRCh38
rs185376937 Chr.8: 17085421-17085421 on Build GRCh38
rs536832798 Chr.8: 17085448-17085448 on Build GRCh38
rs1012348833 Chr.8: 17085192-17085192 on Build GRCh38
rs898733950 Chr.8: 17085172-17085172 on Build GRCh38
rs755846418 Chr.8: 17085363-17085363 on Build GRCh38
rs200086342 Chr.8: 17085249-17085249 on Build GRCh38
rs556786470 Chr.8: 17085510-17085510 on Build GRCh38
rs756560755 Chr.8: 17085194-17085194 on Build GRCh38
rs373742559 Chr.8: 17085108-17085108 on Build GRCh38
rs767745847 Chr.8: 17085324-17085324 on Build GRCh38
rs774414897 Chr.8: 17085285-17085285 on Build GRCh38
rs768320920 Chr.8: 17085430-17085430 on Build GRCh38
rs866660906 Chr.8: 17085259-17085259 on Build GRCh38
rs977130764 Chr.8: 17085096-17085096 on Build GRCh38
rs1013773447 Chr.8: 17085329-17085329 on Build GRCh38
rs1051588743 Chr.8: 17085118-17085118 on Build GRCh38
rs79459174 Chr.8: 17085288-17085288 on Build GRCh38
rs765921021 Chr.8: 17085338-17085338 on Build GRCh38
rs201310335 Chr.8: 17085318-17085318 on Build GRCh38
rs781025156 Chr.8: 17085349-17085349 on Build GRCh38
rs548097761 Chr.8: 17085415-17085415 on Build GRCh38
rs1038380029 Chr.8: 17085150-17085150 on Build GRCh38
rs999282260 Chr.8: 17085278-17085278 on Build GRCh38
rs764571559 Chr.8: 17085332-17085332 on Build GRCh38
rs747979584 Chr.8: 17085229-17085229 on Build GRCh38
rs376261056 Chr.8: 17085198-17085198 on Build GRCh38
rs775790330 Chr.8: 17085325-17085325 on Build GRCh38
rs2285263 Chr.8: 17085102-17085102 on Build GRCh38
rs559129294 Chr.8: 17085166-17085166 on Build GRCh38
rs1024449033 Chr.8: 17085330-17085330 on Build GRCh38
rs770911769 Chr.8: 17085270-17085270 on Build GRCh38
rs764543325 Chr.8: 17085195-17085195 on Build GRCh38
rs989396855 Chr.8: 17085075-17085075 on Build GRCh38
rs940154083 Chr.8: 17085529-17085529 on Build GRCh38
rs146270779 Chr.8: 17085548-17085548 on Build GRCh38
rs939977299 Chr.8: 17085127-17085127 on Build GRCh38
rs1047918784 Chr.8: 17085132-17085132 on Build GRCh38
rs755942799 Chr.8: 17085314-17085316 on Build GRCh38
rs934064623 Chr.8: 17085116-17085116 on Build GRCh38
rs781078344 Chr.8: 17085222-17085222 on Build GRCh38
rs754629138 Chr.8: 17085342-17085342 on Build GRCh38
rs975215997 Chr.8: 17085109-17085109 on Build GRCh38
rs906199759 Chr.8: 17085532-17085533 on Build GRCh38
rs775344871 Chr.8: 17085398-17085398 on Build GRCh38
rs546963033 Chr.8: 17085357-17085357 on Build GRCh38
rs570403926 Chr.8: 17085153-17085153 on Build GRCh38
rs942248346 Chr.8: 17085171-17085171 on Build GRCh38
rs368688623 Chr.8: 17085120-17085120 on Build GRCh38
rs566761333 Chr.8: 17085384-17085384 on Build GRCh38
rs777689846 Chr.8: 17085365-17085365 on Build GRCh38
rs769668921 Chr.8: 17085242-17085242 on Build GRCh38
rs894796883 Chr.8: 17085173-17085173 on Build GRCh38
rs754682239 Chr.8: 17085218-17085218 on Build GRCh38
rs371588190 Chr.8: 17085370-17085370 on Build GRCh38
rs2285265 Chr.8: 17085340-17085340 on Build GRCh38
rs967417545 Chr.8: 17085088-17085088 on Build GRCh38
rs746656722 Chr.8: 17085215-17085215 on Build GRCh38
rs200859456 Chr.8: 17085241-17085241 on Build GRCh38
rs182112261 Chr.8: 17085119-17085119 on Build GRCh38
rs148666305 Chr.8: 17085257-17085257 on Build GRCh38
rs779676253 Chr.8: 17085212-17085212 on Build GRCh38
rs776372641 Chr.8: 17085499-17085499 on Build GRCh38
rs986654528 Chr.8: 17085449-17085449 on Build GRCh38
rs757853436 Chr.8: 17085335-17085335 on Build GRCh38
rs142186793 Chr.8: 17085385-17085385 on Build GRCh38
rs762861145 Chr.8: 17085435-17085435 on Build GRCh38
rs925831647 Chr.8: 17085099-17085099 on Build GRCh38
rs1002495700 Chr.8: 17085246-17085246 on Build GRCh38

View Full Product Details