Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22097585-22098089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
UniGene
Hs.491223
Gene Chromosome Location
Chr.8: 22089150-22104380 on Build GRCh38
Transcripts
NM_022749.5 XM_005273602.3 XM_006716381.2 XM_006716385.3 XM_017013732.1 XM_017013733.1 XM_017013734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2785646 c.438C>T p.S146S Chr.8: 22097752-22097752 on Build GRCh38
2785647 c.478T>G p.C160G Chr.8: 22097792-22097792 on Build GRCh38
6658739 c.516C>T p.Y172Y Chr.8: 22097830-22097830 on Build GRCh38
6891732 c.434G>A p.G145E Chr.8: 22097748-22097748 on Build GRCh38
6415233 c.479G>A p.C160Y Chr.8: 22097793-22097793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377355852 Chr.8: 22097854-22097854 on Build GRCh38
rs765346713 Chr.8: 22097724-22097724 on Build GRCh38
rs775211664 Chr.8: 22097786-22097786 on Build GRCh38
rs11352 Chr.8: 22097605-22097605 on Build GRCh38
rs776813466 Chr.8: 22097805-22097805 on Build GRCh38
rs1001271035 Chr.8: 22097677-22097677 on Build GRCh38
rs898635675 Chr.8: 22097727-22097727 on Build GRCh38
rs1018221350 Chr.8: 22097783-22097783 on Build GRCh38
rs771861790 Chr.8: 22097785-22097785 on Build GRCh38
rs532690794 Chr.8: 22097937-22097937 on Build GRCh38
rs769843390 Chr.8: 22097732-22097733 on Build GRCh38
rs536903609 Chr.8: 22097979-22097979 on Build GRCh38
rs763298686 Chr.8: 22097876-22097876 on Build GRCh38
rs1025807235 Chr.8: 22097975-22097975 on Build GRCh38
rs569952208 Chr.8: 22097615-22097615 on Build GRCh38
rs937008474 Chr.8: 22097909-22097909 on Build GRCh38
rs766676586 Chr.8: 22097878-22097878 on Build GRCh38
rs753816042 Chr.8: 22097707-22097707 on Build GRCh38
rs947020216 Chr.8: 22097644-22097644 on Build GRCh38
rs956810185 Chr.8: 22098033-22098033 on Build GRCh38
rs978379225 Chr.8: 22097920-22097920 on Build GRCh38
rs201277494 Chr.8: 22097771-22097771 on Build GRCh38
rs374830504 Chr.8: 22097632-22097632 on Build GRCh38
rs148519054 Chr.8: 22097659-22097659 on Build GRCh38
rs200052101 Chr.8: 22097673-22097673 on Build GRCh38
rs771667696 Chr.8: 22097857-22097857 on Build GRCh38
rs898250344 Chr.8: 22097940-22097940 on Build GRCh38
rs934404262 Chr.8: 22097946-22097946 on Build GRCh38
rs577309670 Chr.8: 22098059-22098059 on Build GRCh38
rs757532253 Chr.8: 22097721-22097721 on Build GRCh38
rs202006792 Chr.8: 22097651-22097651 on Build GRCh38
rs768438810 Chr.8: 22097800-22097800 on Build GRCh38
rs372658577 Chr.8: 22097803-22097803 on Build GRCh38
rs555270520 Chr.8: 22097650-22097650 on Build GRCh38
rs371809549 Chr.8: 22097612-22097612 on Build GRCh38
rs142809271 Chr.8: 22097866-22097866 on Build GRCh38
rs748712675 Chr.8: 22097759-22097759 on Build GRCh38
rs567319267 Chr.8: 22098009-22098009 on Build GRCh38
rs569323236 Chr.8: 22097972-22097972 on Build GRCh38
rs891326082 Chr.8: 22097994-22097994 on Build GRCh38
rs756714973 Chr.8: 22097839-22097839 on Build GRCh38
rs34061344 Chr.8: 22098004-22098005 on Build GRCh38
rs747529832 Chr.8: 22097747-22097747 on Build GRCh38
rs202029717 Chr.8: 22097735-22097735 on Build GRCh38
rs377182092 Chr.8: 22097865-22097865 on Build GRCh38
rs776783558 Chr.8: 22097682-22097683 on Build GRCh38
rs372519731 Chr.8: 22097859-22097859 on Build GRCh38
rs760633143 Chr.8: 22097797-22097797 on Build GRCh38
rs367620437 Chr.8: 22097703-22097703 on Build GRCh38
rs747438103 Chr.8: 22097736-22097736 on Build GRCh38
rs868202037 Chr.8: 22097637-22097637 on Build GRCh38
rs773375469 Chr.8: 22097868-22097868 on Build GRCh38
rs748311414 Chr.8: 22097680-22097680 on Build GRCh38
rs537394346 Chr.8: 22097631-22097631 on Build GRCh38
rs373744765 Chr.8: 22097867-22097867 on Build GRCh38
rs200042774 Chr.8: 22097855-22097855 on Build GRCh38
rs770307508 Chr.8: 22098018-22098018 on Build GRCh38
rs376667721 Chr.8: 22097831-22097831 on Build GRCh38
rs539578670 Chr.8: 22097836-22097836 on Build GRCh38
rs758824667 Chr.8: 22097737-22097737 on Build GRCh38
rs1973254 Chr.8: 22098041-22098041 on Build GRCh38
rs947072830 Chr.8: 22097916-22097916 on Build GRCh38
rs367901523 Chr.8: 22097664-22097664 on Build GRCh38
rs755545199 Chr.8: 22097829-22097829 on Build GRCh38
rs552781211 Chr.8: 22098058-22098058 on Build GRCh38
rs963986897 Chr.8: 22098001-22098001 on Build GRCh38
rs765555699 Chr.8: 22097815-22097815 on Build GRCh38
rs766871647 Chr.8: 22097824-22097824 on Build GRCh38
rs561283756 Chr.8: 22097830-22097830 on Build GRCh38
rs752014825 Chr.8: 22097885-22097885 on Build GRCh38
rs905507919 Chr.8: 22097893-22097893 on Build GRCh38
rs780497993 Chr.8: 22097746-22097746 on Build GRCh38
rs760541433 Chr.8: 22097697-22097697 on Build GRCh38
rs1018501574 Chr.8: 22097997-22097997 on Build GRCh38
rs377052952 Chr.8: 22097705-22097705 on Build GRCh38
rs1056953904 Chr.8: 22097691-22097692 on Build GRCh38
rs562984468 Chr.8: 22097675-22097675 on Build GRCh38
rs921486641 Chr.8: 22097899-22097899 on Build GRCh38
rs767406750 Chr.8: 22097604-22097604 on Build GRCh38
rs764292084 Chr.8: 22097613-22097613 on Build GRCh38
rs745963127 Chr.8: 22097683-22097683 on Build GRCh38
rs750677280 Chr.8: 22097821-22097821 on Build GRCh38
rs755391684 Chr.8: 22097752-22097752 on Build GRCh38
rs770661336 Chr.8: 22097764-22097764 on Build GRCh38
rs200785895 Chr.8: 22097670-22097670 on Build GRCh38
rs753147403 Chr.8: 22098032-22098032 on Build GRCh38
rs754098746 Chr.8: 22097614-22097614 on Build GRCh38
rs549272533 Chr.8: 22097982-22097982 on Build GRCh38
rs925760199 Chr.8: 22097621-22097621 on Build GRCh38
rs987051592 Chr.8: 22097774-22097774 on Build GRCh38
rs899061748 Chr.8: 22098013-22098013 on Build GRCh38
rs778421444 Chr.8: 22098049-22098049 on Build GRCh38
rs759015926 Chr.8: 22097636-22097636 on Build GRCh38
rs550988593 Chr.8: 22097942-22097942 on Build GRCh38
rs761778789 Chr.8: 22097813-22097813 on Build GRCh38
rs745726476 Chr.8: 22097784-22097784 on Build GRCh38
rs774165278 Chr.8: 22098058-22098059 on Build GRCh38
rs751932847 Chr.8: 22097825-22097825 on Build GRCh38
rs965888736 Chr.8: 22097877-22097877 on Build GRCh38
rs777373559 Chr.8: 22097753-22097753 on Build GRCh38
rs979467510 Chr.8: 22098011-22098011 on Build GRCh38
rs1008417774 Chr.8: 22097996-22097996 on Build GRCh38
rs78824473 Chr.8: 22097889-22097889 on Build GRCh38
rs772338201 Chr.8: 22097693-22097693 on Build GRCh38

View Full Product Details